Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 197328182-197328709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRB1
Entrez Gene ID
23418
Gene Name
crumbs 1, cell polarity complex component
Gene Aliases
LCA8, RP12
UniGene
Hs.126135
Gene Chromosome Location
Chr.1: 197201462-197478455 on Build GRCh38
Transcripts
NM_001193640.1 NM_001257965.1 NM_001257966.1 NM_201253.2 NR_047563.1 NR_047564.1 XM_011509365.2 XM_011509367.1 XM_017000851.1 XM_017000852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3976784 c.28T>A p.C10S Chr.1: 197328586-197328586 on Build GRCh38
4822651 c.107C>G p.S36* Chr.1: 197328458-197328458 on Build GRCh38
2125514 c.214G>C p.D72H Chr.1: 197328565-197328565 on Build GRCh38
4228501 c.7G>C p.D3H Chr.1: 197328565-197328565 on Build GRCh38
3976783 c.235T>A p.C79S Chr.1: 197328586-197328586 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893763475 Chr.1: 197328556-197328556 on Build GRCh38
rs548468838 Chr.1: 197328351-197328351 on Build GRCh38
rs972854308 Chr.1: 197328567-197328567 on Build GRCh38
rs74406466 Chr.1: 197328526-197328526 on Build GRCh38
rs753898127 Chr.1: 197328636-197328636 on Build GRCh38
rs748811426 Chr.1: 197328359-197328359 on Build GRCh38
rs116254396 Chr.1: 197328659-197328659 on Build GRCh38
rs777099127 Chr.1: 197328660-197328660 on Build GRCh38
rs1019383677 Chr.1: 197328398-197328398 on Build GRCh38
rs777954761 Chr.1: 197328514-197328514 on Build GRCh38
rs909976161 Chr.1: 197328255-197328255 on Build GRCh38
rs999558466 Chr.1: 197328282-197328282 on Build GRCh38
rs947867411 Chr.1: 197328457-197328457 on Build GRCh38
rs760083289 Chr.1: 197328689-197328689 on Build GRCh38
rs778903526 Chr.1: 197328655-197328655 on Build GRCh38
rs761295842 Chr.1: 197328470-197328470 on Build GRCh38
rs1050641209 Chr.1: 197328677-197328677 on Build GRCh38
rs750294087 Chr.1: 197328529-197328529 on Build GRCh38
rs368816923 Chr.1: 197328620-197328620 on Build GRCh38
rs754810781 Chr.1: 197328642-197328642 on Build GRCh38
rs62636260 Chr.1: 197328608-197328609 on Build GRCh38
rs916428055 Chr.1: 197328452-197328452 on Build GRCh38
rs1011122138 Chr.1: 197328298-197328298 on Build GRCh38
rs116067724 Chr.1: 197328667-197328667 on Build GRCh38
rs980680937 Chr.1: 197328539-197328539 on Build GRCh38
rs755942444 Chr.1: 197328629-197328629 on Build GRCh38
rs113433549 Chr.1: 197328356-197328356 on Build GRCh38
rs764563530 Chr.1: 197328267-197328267 on Build GRCh38
rs532723434 Chr.1: 197328236-197328236 on Build GRCh38
rs12042924 Chr.1: 197328287-197328287 on Build GRCh38
rs759889286 Chr.1: 197328612-197328612 on Build GRCh38
rs748905381 Chr.1: 197328417-197328417 on Build GRCh38
rs757150006 Chr.1: 197328405-197328405 on Build GRCh38
rs375702596 Chr.1: 197328531-197328531 on Build GRCh38
rs569940169 Chr.1: 197328402-197328402 on Build GRCh38
rs774842265 Chr.1: 197328440-197328440 on Build GRCh38
rs560271761 Chr.1: 197328582-197328582 on Build GRCh38
rs547557527 Chr.1: 197328277-197328277 on Build GRCh38
rs766622294 Chr.1: 197328474-197328474 on Build GRCh38
rs35050379 Chr.1: 197328220-197328221 on Build GRCh38
rs1051479044 Chr.1: 197328301-197328301 on Build GRCh38
rs780148505 Chr.1: 197328634-197328634 on Build GRCh38
rs772914182 Chr.1: 197328404-197328405 on Build GRCh38
rs773786080 Chr.1: 197328665-197328666 on Build GRCh38
rs748239943 Chr.1: 197328404-197328404 on Build GRCh38
rs754866121 Chr.1: 197328378-197328378 on Build GRCh38
rs761047352 Chr.1: 197328586-197328586 on Build GRCh38
rs962778860 Chr.1: 197328561-197328561 on Build GRCh38
rs754232939 Chr.1: 197328483-197328483 on Build GRCh38
rs751678963 Chr.1: 197328627-197328627 on Build GRCh38
rs142514137 Chr.1: 197328680-197328680 on Build GRCh38
rs771491213 Chr.1: 197328600-197328600 on Build GRCh38
rs145141811 Chr.1: 197328486-197328486 on Build GRCh38
rs755856515 Chr.1: 197328297-197328297 on Build GRCh38
rs190292660 Chr.1: 197328283-197328283 on Build GRCh38
rs145525029 Chr.1: 197328616-197328616 on Build GRCh38
rs754837698 Chr.1: 197328356-197328357 on Build GRCh38
rs374653244 Chr.1: 197328408-197328408 on Build GRCh38
rs140428156 Chr.1: 197328512-197328512 on Build GRCh38
rs59691602 Chr.1: 197328450-197328450 on Build GRCh38
rs889615715 Chr.1: 197328312-197328312 on Build GRCh38
rs768441650 Chr.1: 197328426-197328426 on Build GRCh38
rs143046320 Chr.1: 197328507-197328507 on Build GRCh38
rs571591819 Chr.1: 197328413-197328413 on Build GRCh38
rs747895417 Chr.1: 197328555-197328555 on Build GRCh38
rs765448599 Chr.1: 197328493-197328493 on Build GRCh38
rs773556156 Chr.1: 197328294-197328294 on Build GRCh38
rs12042179 Chr.1: 197328410-197328410 on Build GRCh38
rs770494438 Chr.1: 197328679-197328679 on Build GRCh38
rs747726898 Chr.1: 197328656-197328656 on Build GRCh38
rs934010730 Chr.1: 197328661-197328661 on Build GRCh38
rs778707167 Chr.1: 197328552-197328552 on Build GRCh38
rs553875922 Chr.1: 197328641-197328641 on Build GRCh38
rs201893755 Chr.1: 197328614-197328614 on Build GRCh38
rs779941462 Chr.1: 197328541-197328541 on Build GRCh38
rs1030616952 Chr.1: 197328443-197328443 on Build GRCh38
rs772819260 Chr.1: 197328573-197328573 on Build GRCh38
rs779570030 Chr.1: 197328310-197328310 on Build GRCh38
rs894943596 Chr.1: 197328207-197328207 on Build GRCh38
rs550098 Chr.1: 197328278-197328278 on Build GRCh38
rs1046198745 Chr.1: 197328473-197328473 on Build GRCh38
rs62645750 Chr.1: 197328462-197328462 on Build GRCh38
rs749112809 Chr.1: 197328551-197328551 on Build GRCh38
rs776914636 Chr.1: 197328608-197328608 on Build GRCh38
rs763274428 Chr.1: 197328619-197328619 on Build GRCh38
rs771462053 Chr.1: 197328559-197328559 on Build GRCh38
rs755234058 Chr.1: 197328396-197328396 on Build GRCh38
rs918243631 Chr.1: 197328416-197328416 on Build GRCh38
rs369741574 Chr.1: 197328410-197328410 on Build GRCh38

View Full Product Details