Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56271536-56272125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CS
Entrez Gene ID
1431
Gene Name
citrate synthase
Gene Aliases
-
UniGene
Hs.743252
Gene Chromosome Location
Chr.12: 56271699-56300391 on Build GRCh38
Transcripts
NM_004077.2

SNP Detail

SNP ID Chromosome Location
rs762717610 Chr.12: 56271842-56271842 on Build GRCh38
rs1049025373 Chr.12: 56271733-56271733 on Build GRCh38
rs950375568 Chr.12: 56272094-56272094 on Build GRCh38
rs1005384502 Chr.12: 56271816-56271816 on Build GRCh38
rs566330694 Chr.12: 56271948-56271948 on Build GRCh38
rs750687320 Chr.12: 56271722-56271722 on Build GRCh38
rs879096532 Chr.12: 56271921-56271921 on Build GRCh38
rs941643576 Chr.12: 56271673-56271673 on Build GRCh38
rs1046362567 Chr.12: 56271718-56271718 on Build GRCh38
rs565085051 Chr.12: 56271860-56271860 on Build GRCh38
rs547684776 Chr.12: 56271890-56271890 on Build GRCh38
rs1002056867 Chr.12: 56271723-56271723 on Build GRCh38
rs929635012 Chr.12: 56271650-56271650 on Build GRCh38
rs953968700 Chr.12: 56271589-56271589 on Build GRCh38
rs187710366 Chr.12: 56271565-56271565 on Build GRCh38
rs950239206 Chr.12: 56271678-56271678 on Build GRCh38
rs913483430 Chr.12: 56272077-56272077 on Build GRCh38
rs911087587 Chr.12: 56271557-56271557 on Build GRCh38
rs936209109 Chr.12: 56271633-56271633 on Build GRCh38
rs963843629 Chr.12: 56271596-56271596 on Build GRCh38
rs887795100 Chr.12: 56271801-56271801 on Build GRCh38
rs777468623 Chr.12: 56271984-56271984 on Build GRCh38
rs758641229 Chr.12: 56271972-56271972 on Build GRCh38
rs55781726 Chr.12: 56271755-56271755 on Build GRCh38
rs905931189 Chr.12: 56271721-56271721 on Build GRCh38
rs976515938 Chr.12: 56271608-56271608 on Build GRCh38
rs375418765 Chr.12: 56271606-56271606 on Build GRCh38
rs80010441 Chr.12: 56271653-56271653 on Build GRCh38
rs561103913 Chr.12: 56271591-56271591 on Build GRCh38
rs922307217 Chr.12: 56271638-56271638 on Build GRCh38
rs1046759176 Chr.12: 56271652-56271652 on Build GRCh38
rs1045708889 Chr.12: 56271692-56271692 on Build GRCh38
rs925638194 Chr.12: 56271626-56271626 on Build GRCh38
rs768361500 Chr.12: 56271859-56271859 on Build GRCh38
rs1029933424 Chr.12: 56271895-56271895 on Build GRCh38
rs543501673 Chr.12: 56271823-56271823 on Build GRCh38
rs12314191 Chr.12: 56272076-56272076 on Build GRCh38
rs985220122 Chr.12: 56271941-56271941 on Build GRCh38
rs530350752 Chr.12: 56271996-56271996 on Build GRCh38
rs193099996 Chr.12: 56271878-56271878 on Build GRCh38
rs1015472009 Chr.12: 56271869-56271869 on Build GRCh38
rs531629089 Chr.12: 56271811-56271811 on Build GRCh38

View Full Product Details