Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 27718167-27718743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EOMES
Entrez Gene ID
8320
Gene Name
eomesodermin
Gene Aliases
TBR2
UniGene
Hs.591663
Gene Chromosome Location
Chr.3: 27715949-27722715 on Build GRCh38
Transcripts
NM_001278182.1 NM_001278183.1 NM_005442.3 XM_005265510.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3592232 c.1315G>A p.D439N Chr.3: 27718737-27718737 on Build GRCh38
4564675 c.1346_1347CC>TT p.P449L Chr.3: 27718619-27718620 on Build GRCh38
3592233 c.1315G>A p.D439N Chr.3: 27718737-27718737 on Build GRCh38
4950551 c.1372T>G p.Y458D Chr.3: 27718594-27718594 on Build GRCh38
5947974 c.1350T>G p.F450L Chr.3: 27718616-27718616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547805164 Chr.3: 27718252-27718252 on Build GRCh38
rs771819862 Chr.3: 27718587-27718587 on Build GRCh38
rs778431768 Chr.3: 27718568-27718568 on Build GRCh38
rs111754222 Chr.3: 27718402-27718402 on Build GRCh38
rs886812942 Chr.3: 27718265-27718265 on Build GRCh38
rs375610979 Chr.3: 27718714-27718714 on Build GRCh38
rs202235986 Chr.3: 27718690-27718690 on Build GRCh38
rs201135280 Chr.3: 27718589-27718589 on Build GRCh38
rs200264554 Chr.3: 27718684-27718684 on Build GRCh38
rs1003946566 Chr.3: 27718278-27718278 on Build GRCh38
rs770394055 Chr.3: 27718672-27718672 on Build GRCh38
rs927308481 Chr.3: 27718314-27718314 on Build GRCh38
rs184626668 Chr.3: 27718274-27718274 on Build GRCh38
rs917239581 Chr.3: 27718390-27718390 on Build GRCh38
rs138385500 Chr.3: 27718353-27718353 on Build GRCh38
rs547203236 Chr.3: 27718300-27718300 on Build GRCh38
rs774377658 Chr.3: 27718695-27718695 on Build GRCh38
rs192368477 Chr.3: 27718257-27718257 on Build GRCh38
rs796327719 Chr.3: 27718357-27718358 on Build GRCh38
rs527480179 Chr.3: 27718294-27718294 on Build GRCh38
rs747407947 Chr.3: 27718592-27718592 on Build GRCh38
rs1010094457 Chr.3: 27718689-27718689 on Build GRCh38
rs766497359 Chr.3: 27718542-27718542 on Build GRCh38
rs998400085 Chr.3: 27718189-27718189 on Build GRCh38
rs745344912 Chr.3: 27718570-27718570 on Build GRCh38
rs960554414 Chr.3: 27718646-27718646 on Build GRCh38
rs934239596 Chr.3: 27718529-27718529 on Build GRCh38
rs200537781 Chr.3: 27718619-27718619 on Build GRCh38
rs962503348 Chr.3: 27718280-27718280 on Build GRCh38
rs1013496613 Chr.3: 27718694-27718694 on Build GRCh38
rs980563634 Chr.3: 27718309-27718309 on Build GRCh38
rs900044303 Chr.3: 27718524-27718524 on Build GRCh38
rs754449783 Chr.3: 27718666-27718667 on Build GRCh38
rs372326238 Chr.3: 27718696-27718696 on Build GRCh38
rs372750653 Chr.3: 27718536-27718536 on Build GRCh38
rs561608875 Chr.3: 27718637-27718637 on Build GRCh38
rs779910017 Chr.3: 27718226-27718226 on Build GRCh38
rs955863695 Chr.3: 27718230-27718230 on Build GRCh38
rs930767387 Chr.3: 27718317-27718317 on Build GRCh38
rs368534964 Chr.3: 27718459-27718459 on Build GRCh38
rs556128255 Chr.3: 27718491-27718491 on Build GRCh38
rs926006009 Chr.3: 27718465-27718465 on Build GRCh38
rs375555986 Chr.3: 27718571-27718571 on Build GRCh38
rs773390866 Chr.3: 27718683-27718683 on Build GRCh38
rs752604245 Chr.3: 27718551-27718551 on Build GRCh38
rs1038044466 Chr.3: 27718565-27718565 on Build GRCh38
rs988613068 Chr.3: 27718257-27718259 on Build GRCh38
rs999826393 Chr.3: 27718595-27718595 on Build GRCh38
rs960606684 Chr.3: 27718358-27718358 on Build GRCh38
rs541599351 Chr.3: 27718555-27718555 on Build GRCh38
rs1017121386 Chr.3: 27718279-27718279 on Build GRCh38
rs201555899 Chr.3: 27718282-27718282 on Build GRCh38
rs946068097 Chr.3: 27718411-27718411 on Build GRCh38
rs753724810 Chr.3: 27718561-27718561 on Build GRCh38
rs992115967 Chr.3: 27718388-27718388 on Build GRCh38
rs753595500 Chr.3: 27718706-27718706 on Build GRCh38
rs1039812825 Chr.3: 27718514-27718514 on Build GRCh38
rs572707801 Chr.3: 27718553-27718553 on Build GRCh38
rs540714360 Chr.3: 27718229-27718230 on Build GRCh38
rs898207211 Chr.3: 27718566-27718566 on Build GRCh38
rs187408953 Chr.3: 27718438-27718438 on Build GRCh38
rs571149553 Chr.3: 27718269-27718269 on Build GRCh38
rs889458440 Chr.3: 27718557-27718557 on Build GRCh38
rs184648686 Chr.3: 27718541-27718541 on Build GRCh38
rs1049136292 Chr.3: 27718350-27718350 on Build GRCh38
rs377175155 Chr.3: 27718197-27718197 on Build GRCh38
rs765068632 Chr.3: 27718719-27718719 on Build GRCh38
rs539383569 Chr.3: 27718526-27718526 on Build GRCh38
rs777301986 Chr.3: 27718622-27718622 on Build GRCh38
rs191882980 Chr.3: 27718516-27718516 on Build GRCh38
rs761871476 Chr.3: 27718446-27718446 on Build GRCh38

View Full Product Details