Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50196308-50196877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAT1
Entrez Gene ID
2779
Gene Name
G protein subunit alpha transducin 1
Gene Aliases
CSNB1G, CSNBAD3, GBT1, GNATR
UniGene
Hs.517978
Gene Chromosome Location
Chr.3: 50191610-50197696 on Build GRCh38
Transcripts
NM_000172.3 NM_144499.2

SNP Detail

SNP ID Chromosome Location
rs183851764 Chr.3: 50196515-50196515 on Build GRCh38
rs911576803 Chr.3: 50196590-50196590 on Build GRCh38
rs978533911 Chr.3: 50196543-50196543 on Build GRCh38
rs955462068 Chr.3: 50196660-50196660 on Build GRCh38
rs146349746 Chr.3: 50196784-50196784 on Build GRCh38
rs765142440 Chr.3: 50196848-50196848 on Build GRCh38
rs942747309 Chr.3: 50196522-50196522 on Build GRCh38
rs879811738 Chr.3: 50196335-50196335 on Build GRCh38
rs1033210684 Chr.3: 50196548-50196548 on Build GRCh38
rs554401168 Chr.3: 50196409-50196409 on Build GRCh38
rs1043856138 Chr.3: 50196539-50196539 on Build GRCh38
rs1052611572 Chr.3: 50196550-50196550 on Build GRCh38
rs761974103 Chr.3: 50196521-50196521 on Build GRCh38
rs538946106 Chr.3: 50196343-50196343 on Build GRCh38
rs114828382 Chr.3: 50196440-50196440 on Build GRCh38
rs548345252 Chr.3: 50196690-50196690 on Build GRCh38
rs576063928 Chr.3: 50196484-50196484 on Build GRCh38
rs1014135554 Chr.3: 50196785-50196785 on Build GRCh38
rs886431349 Chr.3: 50196367-50196367 on Build GRCh38
rs886058687 Chr.3: 50196367-50196367 on Build GRCh38
rs74718377 Chr.3: 50196663-50196663 on Build GRCh38
rs564641622 Chr.3: 50196562-50196562 on Build GRCh38
rs549813093 Chr.3: 50196847-50196847 on Build GRCh38
rs1004914296 Chr.3: 50196400-50196400 on Build GRCh38
rs987202438 Chr.3: 50196490-50196490 on Build GRCh38
rs34877766 Chr.3: 50196693-50196693 on Build GRCh38
rs141521241 Chr.3: 50196578-50196578 on Build GRCh38
rs886545847 Chr.3: 50196689-50196689 on Build GRCh38
rs572562871 Chr.3: 50196430-50196430 on Build GRCh38
rs559557322 Chr.3: 50196652-50196652 on Build GRCh38
rs955694046 Chr.3: 50196563-50196563 on Build GRCh38
rs896498306 Chr.3: 50196702-50196702 on Build GRCh38
rs918472141 Chr.3: 50196483-50196483 on Build GRCh38
rs944930814 Chr.3: 50196596-50196596 on Build GRCh38
rs970209995 Chr.3: 50196428-50196428 on Build GRCh38
rs891084737 Chr.3: 50196540-50196540 on Build GRCh38
rs541334685 Chr.3: 50196640-50196640 on Build GRCh38
rs1022766234 Chr.3: 50196505-50196505 on Build GRCh38
rs1023977090 Chr.3: 50196422-50196422 on Build GRCh38
rs1014598066 Chr.3: 50196455-50196455 on Build GRCh38
rs1016710383 Chr.3: 50196452-50196452 on Build GRCh38
rs577257104 Chr.3: 50196584-50196584 on Build GRCh38
rs563891409 Chr.3: 50196635-50196635 on Build GRCh38
rs886058689 Chr.3: 50196645-50196645 on Build GRCh38
rs766209614 Chr.3: 50196642-50196642 on Build GRCh38
rs988460257 Chr.3: 50196585-50196585 on Build GRCh38
rs905557444 Chr.3: 50196589-50196589 on Build GRCh38
rs886058688 Chr.3: 50196471-50196471 on Build GRCh38
rs377222546 Chr.3: 50196423-50196423 on Build GRCh38
rs1045802112 Chr.3: 50196588-50196588 on Build GRCh38
rs1014302496 Chr.3: 50196379-50196379 on Build GRCh38
rs1044747667 Chr.3: 50196831-50196831 on Build GRCh38
rs754793155 Chr.3: 50196796-50196796 on Build GRCh38
rs780402124 Chr.3: 50196781-50196781 on Build GRCh38
rs192468377 Chr.3: 50196453-50196453 on Build GRCh38

View Full Product Details