Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74454926-74455521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INO80B
Entrez Gene ID
83444
Gene Name
INO80 complex subunit B
Gene Aliases
HMGA1L4, HMGIYL4, IES2, PAP-1BP, PAPA-1, PAPA1, ZNHIT4, hIes2
UniGene
Hs.410786
Gene Chromosome Location
Chr.2: 74455023-74457960 on Build GRCh38
Transcripts
NM_031288.3 NR_037849.1

Gene Symbol
INO80B-WBP1
Entrez Gene ID
100532735
Gene Name
INO80B-WBP1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.410786
Gene Chromosome Location
Chr.2: 74455023-74460891 on Build GRCh38
Transcripts
NM_031288.3 NR_037849.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6457996 c.9G>T p.K3N Chr.2: 74455125-74455125 on Build GRCh38
4776256 c.95G>A p.G32D Chr.2: 74455442-74455442 on Build GRCh38
4765300 c.112_113insA p.H40fs*67 Chr.2: 74455459-74455460 on Build GRCh38
6685969 c.57C>T p.G19G Chr.2: 74455443-74455443 on Build GRCh38
1210685 c.72G>T p.K24N Chr.2: 74455458-74455458 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751838746 Chr.2: 74455225-74455247 on Build GRCh38
rs764250539 Chr.2: 74455353-74455353 on Build GRCh38
rs755950497 Chr.2: 74455267-74455267 on Build GRCh38
rs776542331 Chr.2: 74455437-74455437 on Build GRCh38
rs748955330 Chr.2: 74455163-74455163 on Build GRCh38
rs770271166 Chr.2: 74455418-74455418 on Build GRCh38
rs77721432 Chr.2: 74455132-74455132 on Build GRCh38
rs758997721 Chr.2: 74455080-74455080 on Build GRCh38
rs767467055 Chr.2: 74455144-74455144 on Build GRCh38
rs201919647 Chr.2: 74455177-74455177 on Build GRCh38
rs775763226 Chr.2: 74455226-74455226 on Build GRCh38
rs559135758 Chr.2: 74455237-74455237 on Build GRCh38
rs768864080 Chr.2: 74455080-74455080 on Build GRCh38
rs376389067 Chr.2: 74455066-74455066 on Build GRCh38
rs766203690 Chr.2: 74455272-74455272 on Build GRCh38
rs371689219 Chr.2: 74455367-74455367 on Build GRCh38
rs941951792 Chr.2: 74455023-74455023 on Build GRCh38
rs376540733 Chr.2: 74455371-74455371 on Build GRCh38
rs150048389 Chr.2: 74455164-74455164 on Build GRCh38
rs764934237 Chr.2: 74455375-74455375 on Build GRCh38
rs541624369 Chr.2: 74455343-74455343 on Build GRCh38
rs963541912 Chr.2: 74455351-74455351 on Build GRCh38
rs781072430 Chr.2: 74455109-74455109 on Build GRCh38
rs747562846 Chr.2: 74455311-74455311 on Build GRCh38
rs35633869 Chr.2: 74455004-74455005 on Build GRCh38
rs754872742 Chr.2: 74455175-74455175 on Build GRCh38
rs904394279 Chr.2: 74455183-74455183 on Build GRCh38
rs986319873 Chr.2: 74455224-74455224 on Build GRCh38
rs371385683 Chr.2: 74455125-74455125 on Build GRCh38
rs969842787 Chr.2: 74455273-74455273 on Build GRCh38
rs746865317 Chr.2: 74455049-74455049 on Build GRCh38
rs1031337178 Chr.2: 74455034-74455034 on Build GRCh38
rs767422570 Chr.2: 74455250-74455250 on Build GRCh38
rs748382891 Chr.2: 74455332-74455333 on Build GRCh38
rs781755319 Chr.2: 74455390-74455390 on Build GRCh38
rs763001891 Chr.2: 74455346-74455346 on Build GRCh38
rs1000432692 Chr.2: 74455148-74455148 on Build GRCh38
rs74319211 Chr.2: 74455005-74455005 on Build GRCh38
rs370683195 Chr.2: 74455313-74455313 on Build GRCh38
rs769795488 Chr.2: 74455474-74455485 on Build GRCh38
rs750363092 Chr.2: 74455256-74455256 on Build GRCh38
rs956306934 Chr.2: 74455165-74455165 on Build GRCh38
rs747604403 Chr.2: 74455181-74455181 on Build GRCh38
rs1007413060 Chr.2: 74454987-74454987 on Build GRCh38
rs151315032 Chr.2: 74455138-74455138 on Build GRCh38
rs115610738 Chr.2: 74455432-74455432 on Build GRCh38
rs779633364 Chr.2: 74455159-74455159 on Build GRCh38
rs762697663 Chr.2: 74455479-74455479 on Build GRCh38
rs766546659 Chr.2: 74455114-74455114 on Build GRCh38
rs761075509 Chr.2: 74455103-74455103 on Build GRCh38
rs532502859 Chr.2: 74455431-74455431 on Build GRCh38
rs748865388 Chr.2: 74455073-74455073 on Build GRCh38
rs201289341 Chr.2: 74455083-74455083 on Build GRCh38
rs762316017 Chr.2: 74455086-74455086 on Build GRCh38
rs540670284 Chr.2: 74455223-74455223 on Build GRCh38
rs763604336 Chr.2: 74455482-74455482 on Build GRCh38
rs770181215 Chr.2: 74455325-74455325 on Build GRCh38
rs780660561 Chr.2: 74455112-74455112 on Build GRCh38
rs76202642 Chr.2: 74455004-74455004 on Build GRCh38
rs955727655 Chr.2: 74455045-74455045 on Build GRCh38
rs777165379 Chr.2: 74455319-74455319 on Build GRCh38
rs374413390 Chr.2: 74455184-74455184 on Build GRCh38
rs757804415 Chr.2: 74455387-74455387 on Build GRCh38
rs1013941038 Chr.2: 74455249-74455249 on Build GRCh38
rs948826986 Chr.2: 74455361-74455361 on Build GRCh38
rs759843018 Chr.2: 74455215-74455215 on Build GRCh38
rs200645639 Chr.2: 74455411-74455411 on Build GRCh38
rs910384914 Chr.2: 74455296-74455296 on Build GRCh38
rs370473011 Chr.2: 74455072-74455072 on Build GRCh38
rs571304264 Chr.2: 74455234-74455234 on Build GRCh38
rs563520386 Chr.2: 74455366-74455366 on Build GRCh38
rs992976764 Chr.2: 74455050-74455050 on Build GRCh38
rs780510155 Chr.2: 74455426-74455426 on Build GRCh38
rs374860064 Chr.2: 74455357-74455357 on Build GRCh38
rs777858361 Chr.2: 74455477-74455479 on Build GRCh38
rs773467904 Chr.2: 74455100-74455100 on Build GRCh38
rs2268417 Chr.2: 74455385-74455385 on Build GRCh38
rs746213420 Chr.2: 74455323-74455323 on Build GRCh38
rs773797901 Chr.2: 74455085-74455085 on Build GRCh38
rs922997446 Chr.2: 74454970-74454970 on Build GRCh38
rs755933772 Chr.2: 74455152-74455152 on Build GRCh38
rs150909895 Chr.2: 74454995-74454995 on Build GRCh38
rs929210769 Chr.2: 74455330-74455330 on Build GRCh38
rs757928470 Chr.2: 74455383-74455383 on Build GRCh38
rs754644248 Chr.2: 74455295-74455295 on Build GRCh38
rs763057950 Chr.2: 74455247-74455247 on Build GRCh38
rs911991703 Chr.2: 74455153-74455153 on Build GRCh38
rs892010575 Chr.2: 74455386-74455386 on Build GRCh38
rs891451847 Chr.2: 74455225-74455225 on Build GRCh38
rs757918662 Chr.2: 74455304-74455304 on Build GRCh38
rs113886180 Chr.2: 74455404-74455404 on Build GRCh38
rs755630698 Chr.2: 74455375-74455378 on Build GRCh38
rs772336300 Chr.2: 74455488-74455488 on Build GRCh38
rs370700489 Chr.2: 74455206-74455206 on Build GRCh38
rs752366861 Chr.2: 74455458-74455458 on Build GRCh38
rs543075139 Chr.2: 74455322-74455323 on Build GRCh38
rs1057120587 Chr.2: 74455332-74455332 on Build GRCh38
rs755435841 Chr.2: 74455269-74455270 on Build GRCh38
rs768723005 Chr.2: 74455430-74455430 on Build GRCh38
rs752215837 Chr.2: 74454961-74454961 on Build GRCh38
rs770891288 Chr.2: 74455433-74455433 on Build GRCh38
rs750226808 Chr.2: 74455145-74455145 on Build GRCh38
rs897811432 Chr.2: 74455074-74455074 on Build GRCh38
rs368300180 Chr.2: 74455197-74455197 on Build GRCh38
rs753593358 Chr.2: 74455288-74455288 on Build GRCh38
rs765027562 Chr.2: 74455443-74455443 on Build GRCh38
rs564421723 Chr.2: 74455428-74455428 on Build GRCh38
rs1024083724 Chr.2: 74455054-74455054 on Build GRCh38
rs943462717 Chr.2: 74455497-74455497 on Build GRCh38
rs778199534 Chr.2: 74455307-74455307 on Build GRCh38
rs747693837 Chr.2: 74455303-74455303 on Build GRCh38
rs200566124 Chr.2: 74455116-74455116 on Build GRCh38
rs769381161 Chr.2: 74455071-74455071 on Build GRCh38
rs759382243 Chr.2: 74455439-74455439 on Build GRCh38
rs772587995 Chr.2: 74455096-74455096 on Build GRCh38
rs1044561340 Chr.2: 74455140-74455140 on Build GRCh38
rs929346174 Chr.2: 74455143-74455143 on Build GRCh38
rs752379431 Chr.2: 74455376-74455376 on Build GRCh38

View Full Product Details