Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 189665211-189665583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASNSD1
Entrez Gene ID
54529
Gene Name
asparagine synthetase domain containing 1
Gene Aliases
NBLA00058, NS3TP1
UniGene
Hs.101364
Gene Chromosome Location
Chr.2: 189661399-189670831 on Build GRCh38
Transcripts
NM_019048.2 XM_017004381.1 XM_017004382.1

SNP Detail

SNP ID Chromosome Location
rs1006886838 Chr.2: 189665359-189665359 on Build GRCh38
rs559909960 Chr.2: 189665248-189665248 on Build GRCh38
rs181567607 Chr.2: 189665312-189665312 on Build GRCh38
rs899140721 Chr.2: 189665273-189665273 on Build GRCh38
rs777429322 Chr.2: 189665266-189665266 on Build GRCh38
rs1029289553 Chr.2: 189665295-189665295 on Build GRCh38
rs148109339 Chr.2: 189665403-189665403 on Build GRCh38
rs914931619 Chr.2: 189665393-189665393 on Build GRCh38
rs528355056 Chr.2: 189665449-189665449 on Build GRCh38
rs993574199 Chr.2: 189665335-189665335 on Build GRCh38
rs989160454 Chr.2: 189665392-189665392 on Build GRCh38
rs1017908883 Chr.2: 189665389-189665389 on Build GRCh38
rs987876725 Chr.2: 189665519-189665519 on Build GRCh38
rs996112699 Chr.2: 189665285-189665285 on Build GRCh38
rs980413996 Chr.2: 189665430-189665430 on Build GRCh38
rs954599626 Chr.2: 189665308-189665308 on Build GRCh38
rs1057277292 Chr.2: 189665235-189665235 on Build GRCh38
rs546529453 Chr.2: 189665507-189665507 on Build GRCh38
rs547073070 Chr.2: 189665525-189665525 on Build GRCh38
rs1030484839 Chr.2: 189665425-189665425 on Build GRCh38
rs528589370 Chr.2: 189665332-189665332 on Build GRCh38
rs1004917582 Chr.2: 189665264-189665264 on Build GRCh38
rs886505286 Chr.2: 189665238-189665238 on Build GRCh38
rs887750498 Chr.2: 189665358-189665358 on Build GRCh38
rs956443198 Chr.2: 189665381-189665381 on Build GRCh38
rs748753805 Chr.2: 189665356-189665356 on Build GRCh38
rs777249803 Chr.2: 189665385-189665385 on Build GRCh38

View Full Product Details