Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 186753700-186754187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM171B
Entrez Gene ID
165215
Gene Name
family with sequence similarity 171 member B
Gene Aliases
KIAA1946
UniGene
Hs.28872
Gene Chromosome Location
Chr.2: 186694062-186763787 on Build GRCh38
Transcripts
NM_177454.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3668904 c.951C>G p.I317M Chr.2: 186753988-186753988 on Build GRCh38
5058510 c.912T>C p.H304H Chr.2: 186753949-186753949 on Build GRCh38
5805872 c.982T>A p.W328R Chr.2: 186754019-186754019 on Build GRCh38
5422250 c.929A>T p.K310M Chr.2: 186753966-186753966 on Build GRCh38
2711586 c.988G>T p.A330S Chr.2: 186754025-186754025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745607684 Chr.2: 186754023-186754023 on Build GRCh38
rs200326195 Chr.2: 186753922-186753922 on Build GRCh38
rs1044759867 Chr.2: 186753755-186753755 on Build GRCh38
rs761669247 Chr.2: 186753844-186753844 on Build GRCh38
rs763459201 Chr.2: 186753916-186753916 on Build GRCh38
rs756135130 Chr.2: 186753985-186753985 on Build GRCh38
rs770285097 Chr.2: 186754064-186754065 on Build GRCh38
rs763585240 Chr.2: 186754089-186754089 on Build GRCh38
rs766941224 Chr.2: 186754097-186754097 on Build GRCh38
rs768467623 Chr.2: 186753753-186753753 on Build GRCh38
rs767571448 Chr.2: 186753957-186753957 on Build GRCh38
rs986459102 Chr.2: 186754095-186754095 on Build GRCh38
rs773604753 Chr.2: 186754074-186754074 on Build GRCh38
rs971818114 Chr.2: 186753804-186753804 on Build GRCh38
rs377737238 Chr.2: 186753978-186753978 on Build GRCh38
rs778705698 Chr.2: 186754022-186754022 on Build GRCh38
rs141548972 Chr.2: 186753859-186753859 on Build GRCh38
rs201265058 Chr.2: 186753974-186753974 on Build GRCh38
rs539944333 Chr.2: 186754122-186754122 on Build GRCh38
rs752890555 Chr.2: 186753979-186753979 on Build GRCh38
rs574171188 Chr.2: 186754163-186754163 on Build GRCh38
rs1002936702 Chr.2: 186753826-186753826 on Build GRCh38
rs139536114 Chr.2: 186754135-186754135 on Build GRCh38
rs746583881 Chr.2: 186754066-186754066 on Build GRCh38
rs771758543 Chr.2: 186754038-186754038 on Build GRCh38
rs777816494 Chr.2: 186754003-186754003 on Build GRCh38
rs565160598 Chr.2: 186753834-186753834 on Build GRCh38
rs914888409 Chr.2: 186753745-186753745 on Build GRCh38
rs556990370 Chr.2: 186753837-186753837 on Build GRCh38
rs188512373 Chr.2: 186753746-186753746 on Build GRCh38
rs759976098 Chr.2: 186754100-186754100 on Build GRCh38
rs749197417 Chr.2: 186754007-186754007 on Build GRCh38
rs765836118 Chr.2: 186753884-186753884 on Build GRCh38
rs944387449 Chr.2: 186754114-186754114 on Build GRCh38
rs531687623 Chr.2: 186754107-186754107 on Build GRCh38
rs373024377 Chr.2: 186753921-186753921 on Build GRCh38
rs955589290 Chr.2: 186754112-186754112 on Build GRCh38
rs73035680 Chr.2: 186754105-186754105 on Build GRCh38
rs371412229 Chr.2: 186753993-186753993 on Build GRCh38
rs947810052 Chr.2: 186753752-186753752 on Build GRCh38
rs200396345 Chr.2: 186754041-186754041 on Build GRCh38
rs539088302 Chr.2: 186753880-186753880 on Build GRCh38
rs149747896 Chr.2: 186753953-186753953 on Build GRCh38
rs748542435 Chr.2: 186754062-186754063 on Build GRCh38
rs368873477 Chr.2: 186753903-186753903 on Build GRCh38
rs1040102975 Chr.2: 186754118-186754118 on Build GRCh38
rs768025760 Chr.2: 186754074-186754074 on Build GRCh38
rs951926765 Chr.2: 186754033-186754033 on Build GRCh38
rs927854824 Chr.2: 186753806-186753806 on Build GRCh38
rs774861760 Chr.2: 186754098-186754098 on Build GRCh38
rs755271794 Chr.2: 186753954-186753954 on Build GRCh38
rs893425473 Chr.2: 186753728-186753728 on Build GRCh38
rs773896536 Chr.2: 186754080-186754080 on Build GRCh38
rs148902381 Chr.2: 186754014-186754014 on Build GRCh38
rs17855085 Chr.2: 186753941-186753941 on Build GRCh38
rs192843701 Chr.2: 186753822-186753822 on Build GRCh38

View Full Product Details