Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20952309-20952865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH3
Entrez Gene ID
55567
Gene Name
dynein axonemal heavy chain 3
Gene Aliases
DLP3, DNAHC3B, HEL-36, Hsadhc3, dnahc3-b
UniGene
Hs.526500
Gene Chromosome Location
Chr.16: 20933111-21159537 on Build GRCh38
Transcripts
NM_017539.2 XM_006721062.1 XM_011545882.2 XM_011545883.1 XM_011545885.2 XM_011545886.2 XM_011545888.2 XM_011545889.2 XM_017023426.1 XM_017023427.1 XM_017023428.1 XM_017023430.1 XM_017023432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5937681 c.11084C>T p.P3695L Chr.16: 20952537-20952537 on Build GRCh38
6646601 c.11181C>A p.Y3727* Chr.16: 20952440-20952440 on Build GRCh38
6249090 c.11186C>A p.T3729K Chr.16: 20952435-20952435 on Build GRCh38
7364917 c.11111G>A p.R3704K Chr.16: 20952510-20952510 on Build GRCh38
4459001 c.11083C>T p.P3695S Chr.16: 20952538-20952538 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747008999 Chr.16: 20952487-20952487 on Build GRCh38
rs991472733 Chr.16: 20952676-20952676 on Build GRCh38
rs757246014 Chr.16: 20952810-20952810 on Build GRCh38
rs774718288 Chr.16: 20952508-20952508 on Build GRCh38
rs771143519 Chr.16: 20952496-20952496 on Build GRCh38
rs1007202662 Chr.16: 20952794-20952795 on Build GRCh38
rs958943775 Chr.16: 20952670-20952670 on Build GRCh38
rs994929251 Chr.16: 20952643-20952643 on Build GRCh38
rs370235264 Chr.16: 20952411-20952411 on Build GRCh38
rs778318607 Chr.16: 20952566-20952567 on Build GRCh38
rs749323164 Chr.16: 20952553-20952553 on Build GRCh38
rs772532168 Chr.16: 20952576-20952576 on Build GRCh38
rs375343796 Chr.16: 20952425-20952425 on Build GRCh38
rs146352384 Chr.16: 20952538-20952538 on Build GRCh38
rs917369969 Chr.16: 20952346-20952346 on Build GRCh38
rs1009078791 Chr.16: 20952565-20952565 on Build GRCh38
rs897989648 Chr.16: 20952620-20952620 on Build GRCh38
rs758197286 Chr.16: 20952471-20952471 on Build GRCh38
rs573791190 Chr.16: 20952548-20952548 on Build GRCh38
rs181218008 Chr.16: 20952724-20952724 on Build GRCh38
rs775203165 Chr.16: 20952416-20952416 on Build GRCh38
rs555528222 Chr.16: 20952539-20952539 on Build GRCh38
rs779263235 Chr.16: 20952564-20952564 on Build GRCh38
rs754992503 Chr.16: 20952550-20952550 on Build GRCh38
rs376889706 Chr.16: 20952400-20952400 on Build GRCh38
rs781021745 Chr.16: 20952552-20952552 on Build GRCh38
rs763723451 Chr.16: 20952445-20952445 on Build GRCh38
rs915186169 Chr.16: 20952768-20952768 on Build GRCh38
rs753397200 Chr.16: 20952451-20952451 on Build GRCh38
rs375064486 Chr.16: 20952618-20952618 on Build GRCh38
rs762735814 Chr.16: 20952694-20952694 on Build GRCh38
rs367972754 Chr.16: 20952466-20952466 on Build GRCh38
rs568605345 Chr.16: 20952372-20952372 on Build GRCh38
rs532924294 Chr.16: 20952678-20952678 on Build GRCh38
rs989311654 Chr.16: 20952756-20952756 on Build GRCh38
rs535933939 Chr.16: 20952394-20952394 on Build GRCh38
rs1047921975 Chr.16: 20952595-20952595 on Build GRCh38
rs758177094 Chr.16: 20952540-20952540 on Build GRCh38
rs372926484 Chr.16: 20952343-20952343 on Build GRCh38
rs564721645 Chr.16: 20952811-20952811 on Build GRCh38
rs761281498 Chr.16: 20952524-20952524 on Build GRCh38
rs971393823 Chr.16: 20952702-20952702 on Build GRCh38
rs202213770 Chr.16: 20952578-20952578 on Build GRCh38
rs147732992 Chr.16: 20952510-20952510 on Build GRCh38
rs7199676 Chr.16: 20952443-20952443 on Build GRCh38
rs185222427 Chr.16: 20952517-20952517 on Build GRCh38
rs7199127 Chr.16: 20952345-20952345 on Build GRCh38
rs761263884 Chr.16: 20952536-20952536 on Build GRCh38
rs950333004 Chr.16: 20952630-20952631 on Build GRCh38
rs964765756 Chr.16: 20952562-20952562 on Build GRCh38
rs756979507 Chr.16: 20952458-20952458 on Build GRCh38
rs370131157 Chr.16: 20952584-20952584 on Build GRCh38
rs929430472 Chr.16: 20952357-20952357 on Build GRCh38
rs749449495 Chr.16: 20952492-20952492 on Build GRCh38
rs1032736919 Chr.16: 20952454-20952454 on Build GRCh38
rs372537271 Chr.16: 20952477-20952477 on Build GRCh38
rs771870160 Chr.16: 20952409-20952410 on Build GRCh38
rs777950411 Chr.16: 20952491-20952491 on Build GRCh38
rs773294333 Chr.16: 20952396-20952396 on Build GRCh38
rs778456968 Chr.16: 20952459-20952459 on Build GRCh38
rs761220984 Chr.16: 20952574-20952575 on Build GRCh38
rs372703654 Chr.16: 20952571-20952571 on Build GRCh38
rs924685363 Chr.16: 20952741-20952741 on Build GRCh38
rs773372645 Chr.16: 20952522-20952522 on Build GRCh38
rs766605380 Chr.16: 20952649-20952649 on Build GRCh38
rs769760299 Chr.16: 20952490-20952490 on Build GRCh38
rs751419215 Chr.16: 20952743-20952743 on Build GRCh38
rs776133393 Chr.16: 20952519-20952519 on Build GRCh38
rs771169474 Chr.16: 20952401-20952401 on Build GRCh38
rs950158977 Chr.16: 20952369-20952369 on Build GRCh38
rs768069677 Chr.16: 20952432-20952432 on Build GRCh38
rs776162817 Chr.16: 20952534-20952534 on Build GRCh38
rs557208445 Chr.16: 20952424-20952424 on Build GRCh38
rs188851172 Chr.16: 20952554-20952554 on Build GRCh38
rs1027894734 Chr.16: 20952662-20952662 on Build GRCh38
rs151204400 Chr.16: 20952846-20952847 on Build GRCh38
rs890988372 Chr.16: 20952792-20952792 on Build GRCh38
rs747353244 Chr.16: 20952587-20952587 on Build GRCh38

View Full Product Details