Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 8768069-8768325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABAT
Entrez Gene ID
18
Gene Name
4-aminobutyrate aminotransferase
Gene Aliases
GABA-AT, GABAT, NPD009
UniGene
Hs.336768
Gene Chromosome Location
Chr.16: 8674587-8784575 on Build GRCh38
Transcripts
NM_000663.4 NM_001127448.1 NM_020686.5 XM_011522400.2 XM_011522401.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7196520 c.623A>G p.Y208C Chr.16: 8768212-8768212 on Build GRCh38
180129 c.619G>A p.D207N Chr.16: 8768208-8768208 on Build GRCh38
435910 c.645C>T p.G215G Chr.16: 8768234-8768234 on Build GRCh38
4063479 c.660G>A p.R220R Chr.16: 8768249-8768249 on Build GRCh38
4503254 c.635C>T p.S212F Chr.16: 8768224-8768224 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1024424010 Chr.16: 8768090-8768090 on Build GRCh38
rs369887885 Chr.16: 8768289-8768289 on Build GRCh38
rs756875305 Chr.16: 8768274-8768274 on Build GRCh38
rs747042892 Chr.16: 8768212-8768212 on Build GRCh38
rs770138381 Chr.16: 8768297-8768297 on Build GRCh38
rs573277423 Chr.16: 8768127-8768127 on Build GRCh38
rs563750981 Chr.16: 8768098-8768098 on Build GRCh38
rs751426054 Chr.16: 8768152-8768152 on Build GRCh38
rs376594468 Chr.16: 8768148-8768148 on Build GRCh38
rs369668077 Chr.16: 8768158-8768158 on Build GRCh38
rs886052430 Chr.16: 8768264-8768265 on Build GRCh38
rs760072765 Chr.16: 8768265-8768265 on Build GRCh38
rs752253647 Chr.16: 8768175-8768175 on Build GRCh38
rs769070363 Chr.16: 8768288-8768288 on Build GRCh38
rs753309345 Chr.16: 8768271-8768271 on Build GRCh38
rs1139522 Chr.16: 8768234-8768234 on Build GRCh38
rs773628864 Chr.16: 8768306-8768306 on Build GRCh38
rs772588617 Chr.16: 8768144-8768144 on Build GRCh38
rs767550217 Chr.16: 8768237-8768237 on Build GRCh38
rs768826504 Chr.16: 8768225-8768225 on Build GRCh38
rs947705390 Chr.16: 8768137-8768137 on Build GRCh38
rs546406644 Chr.16: 8768275-8768275 on Build GRCh38
rs1038256499 Chr.16: 8768268-8768268 on Build GRCh38
rs989725510 Chr.16: 8768099-8768099 on Build GRCh38
rs762050203 Chr.16: 8768232-8768232 on Build GRCh38
rs1033978532 Chr.16: 8768159-8768159 on Build GRCh38
rs779198838 Chr.16: 8768201-8768201 on Build GRCh38
rs752732104 Chr.16: 8768240-8768240 on Build GRCh38
rs371057736 Chr.16: 8768295-8768295 on Build GRCh38
rs369997983 Chr.16: 8768126-8768126 on Build GRCh38
rs149271402 Chr.16: 8768230-8768230 on Build GRCh38
rs121434578 Chr.16: 8768248-8768248 on Build GRCh38
rs756890572 Chr.16: 8768193-8768193 on Build GRCh38
rs373357773 Chr.16: 8768207-8768207 on Build GRCh38
rs767568755 Chr.16: 8768163-8768163 on Build GRCh38
rs763619234 Chr.16: 8768269-8768269 on Build GRCh38
rs766162227 Chr.16: 8768149-8768149 on Build GRCh38
rs724159990 Chr.16: 8768220-8768220 on Build GRCh38
rs777269086 Chr.16: 8768186-8768186 on Build GRCh38
rs376506551 Chr.16: 8768276-8768276 on Build GRCh38
rs747587543 Chr.16: 8768283-8768283 on Build GRCh38
rs373385788 Chr.16: 8768147-8768147 on Build GRCh38
rs759472888 Chr.16: 8768236-8768236 on Build GRCh38
rs1139523 Chr.16: 8768235-8768235 on Build GRCh38
rs915463954 Chr.16: 8768105-8768105 on Build GRCh38
rs772498753 Chr.16: 8768208-8768208 on Build GRCh38
rs755708488 Chr.16: 8768177-8768177 on Build GRCh38
rs775488254 Chr.16: 8768145-8768145 on Build GRCh38
rs530804959 Chr.16: 8768160-8768160 on Build GRCh38
rs760745761 Chr.16: 8768146-8768146 on Build GRCh38
rs748783252 Chr.16: 8768261-8768262 on Build GRCh38
rs542575561 Chr.16: 8768187-8768187 on Build GRCh38
rs561050474 Chr.16: 8768210-8768210 on Build GRCh38

View Full Product Details