Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67119900-67120448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf70
Entrez Gene ID
80262
Gene Name
chromosome 16 open reading frame 70
Gene Aliases
C16orf6, LIN10, lin-10
UniGene
Hs.513666
Gene Chromosome Location
Chr.16: 67109881-67148544 on Build GRCh38
Transcripts
NM_001320540.1 NM_001320541.1 NM_001320542.1 NM_001320543.1 NM_025187.4 XM_006721284.3 XM_011523350.2 XM_017023729.1 XM_017023730.1 XM_017023731.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2994147 c.110G>A p.R37H Chr.16: 67120157-67120157 on Build GRCh38
7201682 c.97C>A p.Q33K Chr.16: 67120144-67120144 on Build GRCh38
972280 c.121A>T p.N41Y Chr.16: 67120168-67120168 on Build GRCh38
6079537 c.65-1G>T p.? Chr.16: 67120111-67120111 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs531283269 Chr.16: 67119974-67119974 on Build GRCh38
rs912034762 Chr.16: 67120275-67120275 on Build GRCh38
rs762048342 Chr.16: 67120117-67120117 on Build GRCh38
rs759398936 Chr.16: 67120134-67120134 on Build GRCh38
rs932305356 Chr.16: 67120299-67120299 on Build GRCh38
rs1031441827 Chr.16: 67120368-67120368 on Build GRCh38
rs756869101 Chr.16: 67120175-67120175 on Build GRCh38
rs762410433 Chr.16: 67120169-67120169 on Build GRCh38
rs959873661 Chr.16: 67120044-67120044 on Build GRCh38
rs1042689147 Chr.16: 67120309-67120309 on Build GRCh38
rs761991655 Chr.16: 67120369-67120369 on Build GRCh38
rs920674696 Chr.16: 67120252-67120252 on Build GRCh38
rs1012442757 Chr.16: 67120399-67120399 on Build GRCh38
rs551401384 Chr.16: 67119996-67119996 on Build GRCh38
rs750845517 Chr.16: 67120218-67120218 on Build GRCh38
rs774472222 Chr.16: 67120116-67120116 on Build GRCh38
rs778996914 Chr.16: 67120239-67120239 on Build GRCh38
rs970111804 Chr.16: 67119995-67119995 on Build GRCh38
rs998586908 Chr.16: 67120364-67120364 on Build GRCh38
rs895631491 Chr.16: 67120371-67120371 on Build GRCh38
rs928362304 Chr.16: 67120025-67120025 on Build GRCh38
rs78593984 Chr.16: 67120377-67120377 on Build GRCh38
rs749759258 Chr.16: 67120235-67120235 on Build GRCh38
rs1016316690 Chr.16: 67120365-67120365 on Build GRCh38
rs1039244395 Chr.16: 67120292-67120292 on Build GRCh38
rs200715975 Chr.16: 67120180-67120180 on Build GRCh38
rs934357945 Chr.16: 67120158-67120158 on Build GRCh38
rs983951789 Chr.16: 67120272-67120272 on Build GRCh38
rs904194098 Chr.16: 67120346-67120346 on Build GRCh38
rs554121700 Chr.16: 67120422-67120422 on Build GRCh38
rs867893630 Chr.16: 67119969-67119969 on Build GRCh38
rs780583070 Chr.16: 67120234-67120234 on Build GRCh38
rs569365112 Chr.16: 67120298-67120298 on Build GRCh38
rs763618795 Chr.16: 67120170-67120170 on Build GRCh38
rs944894847 Chr.16: 67120281-67120281 on Build GRCh38
rs537952558 Chr.16: 67120324-67120324 on Build GRCh38
rs75694020 Chr.16: 67120236-67120236 on Build GRCh38
rs947269430 Chr.16: 67120231-67120231 on Build GRCh38
rs748320235 Chr.16: 67120244-67120244 on Build GRCh38
rs368936324 Chr.16: 67120156-67120156 on Build GRCh38
rs761914392 Chr.16: 67120305-67120305 on Build GRCh38
rs1031474699 Chr.16: 67120010-67120010 on Build GRCh38
rs540602489 Chr.16: 67120407-67120407 on Build GRCh38
rs149407168 Chr.16: 67120157-67120157 on Build GRCh38
rs555668680 Chr.16: 67120276-67120276 on Build GRCh38
rs566796872 Chr.16: 67120233-67120233 on Build GRCh38
rs768753247 Chr.16: 67120113-67120113 on Build GRCh38
rs775964035 Chr.16: 67120103-67120103 on Build GRCh38
rs770355313 Chr.16: 67120100-67120100 on Build GRCh38
rs188455461 Chr.16: 67120122-67120122 on Build GRCh38
rs776498444 Chr.16: 67120132-67120132 on Build GRCh38
rs770648324 Chr.16: 67120131-67120131 on Build GRCh38
rs578070997 Chr.16: 67120398-67120398 on Build GRCh38
rs750203409 Chr.16: 67120428-67120428 on Build GRCh38
rs774599041 Chr.16: 67120258-67120258 on Build GRCh38
rs751192450 Chr.16: 67120171-67120171 on Build GRCh38
rs973732227 Chr.16: 67120425-67120425 on Build GRCh38
rs991451177 Chr.16: 67119944-67119944 on Build GRCh38
rs570900207 Chr.16: 67120094-67120094 on Build GRCh38
rs749419820 Chr.16: 67120105-67120105 on Build GRCh38
rs774243417 Chr.16: 67120280-67120280 on Build GRCh38

View Full Product Details