Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 36437722-36438301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNA1
Entrez Gene ID
8900
Gene Name
cyclin A1
Gene Aliases
CT146
UniGene
Hs.417050
Gene Chromosome Location
Chr.13: 36430488-36442882 on Build GRCh38
Transcripts
NM_001111045.1 NM_001111046.1 NM_001111047.1 NM_003914.3 XM_011535294.2 XM_011535295.2 XM_011535296.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4752916 c.421G>T p.E141* Chr.13: 36437752-36437752 on Build GRCh38
6622912 c.469G>A p.A157T Chr.13: 36437800-36437800 on Build GRCh38
5920581 c.596C>T p.S199F Chr.13: 36438118-36438118 on Build GRCh38
4766571 c.649C>A p.Q217K Chr.13: 36438171-36438171 on Build GRCh38
5933187 c.544+1G>A p.? Chr.13: 36437876-36437876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753412232 Chr.13: 36437820-36437820 on Build GRCh38
rs765912844 Chr.13: 36437957-36437957 on Build GRCh38
rs1005676747 Chr.13: 36437940-36437940 on Build GRCh38
rs185033021 Chr.13: 36438264-36438264 on Build GRCh38
rs1034837171 Chr.13: 36437969-36437969 on Build GRCh38
rs868177414 Chr.13: 36437752-36437752 on Build GRCh38
rs967616239 Chr.13: 36438113-36438113 on Build GRCh38
rs1046224448 Chr.13: 36438004-36438004 on Build GRCh38
rs757524421 Chr.13: 36438203-36438203 on Build GRCh38
rs539238131 Chr.13: 36438269-36438269 on Build GRCh38
rs763296052 Chr.13: 36437794-36437794 on Build GRCh38
rs35404413 Chr.13: 36438131-36438131 on Build GRCh38
rs373454867 Chr.13: 36438098-36438098 on Build GRCh38
rs377540850 Chr.13: 36438211-36438211 on Build GRCh38
rs758365529 Chr.13: 36437850-36437850 on Build GRCh38
rs758653417 Chr.13: 36438220-36438220 on Build GRCh38
rs751371810 Chr.13: 36438173-36438173 on Build GRCh38
rs529627995 Chr.13: 36437891-36437891 on Build GRCh38
rs374907767 Chr.13: 36437813-36437813 on Build GRCh38
rs778313716 Chr.13: 36438231-36438231 on Build GRCh38
rs775344684 Chr.13: 36438159-36438159 on Build GRCh38
rs778026294 Chr.13: 36438070-36438070 on Build GRCh38
rs753405934 Chr.13: 36438033-36438033 on Build GRCh38
rs773640359 Chr.13: 36438101-36438101 on Build GRCh38
rs748507767 Chr.13: 36438095-36438095 on Build GRCh38
rs772143355 Chr.13: 36437775-36437775 on Build GRCh38
rs758740150 Chr.13: 36438054-36438054 on Build GRCh38
rs747420286 Chr.13: 36438233-36438233 on Build GRCh38
rs767463764 Chr.13: 36438200-36438200 on Build GRCh38
rs777961827 Chr.13: 36438229-36438229 on Build GRCh38
rs200724447 Chr.13: 36437870-36437870 on Build GRCh38
rs764957102 Chr.13: 36438146-36438146 on Build GRCh38
rs751505490 Chr.13: 36438064-36438064 on Build GRCh38
rs199835821 Chr.13: 36437806-36437806 on Build GRCh38
rs779154163 Chr.13: 36438094-36438094 on Build GRCh38
rs895264872 Chr.13: 36437974-36437974 on Build GRCh38
rs753604464 Chr.13: 36437830-36437830 on Build GRCh38
rs749452494 Chr.13: 36437874-36437874 on Build GRCh38
rs61733784 Chr.13: 36437756-36437756 on Build GRCh38
rs763735177 Chr.13: 36438172-36438172 on Build GRCh38
rs368185225 Chr.13: 36437839-36437839 on Build GRCh38
rs985602077 Chr.13: 36437816-36437816 on Build GRCh38
rs777624140 Chr.13: 36437852-36437852 on Build GRCh38
rs936708044 Chr.13: 36437916-36437916 on Build GRCh38
rs113260257 Chr.13: 36437798-36437798 on Build GRCh38
rs143100244 Chr.13: 36437783-36437783 on Build GRCh38
rs34193801 Chr.13: 36437914-36437915 on Build GRCh38
rs755143850 Chr.13: 36438083-36438083 on Build GRCh38
rs115775515 Chr.13: 36438050-36438050 on Build GRCh38
rs759227358 Chr.13: 36438045-36438045 on Build GRCh38
rs753784627 Chr.13: 36437788-36437789 on Build GRCh38
rs756715582 Chr.13: 36438026-36438030 on Build GRCh38
rs769827681 Chr.13: 36437871-36437871 on Build GRCh38
rs371431411 Chr.13: 36437764-36437764 on Build GRCh38
rs139038414 Chr.13: 36438130-36438130 on Build GRCh38
rs765937307 Chr.13: 36437786-36437786 on Build GRCh38
rs879082940 Chr.13: 36438024-36438024 on Build GRCh38
rs1030435374 Chr.13: 36438205-36438205 on Build GRCh38
rs753892533 Chr.13: 36438202-36438202 on Build GRCh38
rs985781492 Chr.13: 36438259-36438259 on Build GRCh38
rs760220718 Chr.13: 36437920-36437920 on Build GRCh38
rs1017971659 Chr.13: 36438111-36438111 on Build GRCh38
rs745479639 Chr.13: 36438239-36438240 on Build GRCh38
rs935793918 Chr.13: 36437771-36437771 on Build GRCh38
rs780039232 Chr.13: 36438079-36438079 on Build GRCh38
rs757101069 Chr.13: 36438180-36438180 on Build GRCh38
rs748042719 Chr.13: 36437767-36437767 on Build GRCh38
rs772610332 Chr.13: 36437894-36437894 on Build GRCh38
rs766051820 Chr.13: 36437829-36437829 on Build GRCh38
rs779094220 Chr.13: 36438210-36438210 on Build GRCh38
rs540179877 Chr.13: 36437789-36437789 on Build GRCh38
rs754694176 Chr.13: 36437836-36437836 on Build GRCh38
rs199819328 Chr.13: 36438230-36438230 on Build GRCh38
rs757229441 Chr.13: 36438075-36438075 on Build GRCh38
rs776235924 Chr.13: 36438136-36438136 on Build GRCh38
rs944415916 Chr.13: 36438273-36438273 on Build GRCh38
rs747163107 Chr.13: 36438100-36438100 on Build GRCh38
rs769065045 Chr.13: 36437795-36437795 on Build GRCh38
rs770566281 Chr.13: 36438245-36438245 on Build GRCh38
rs992957639 Chr.13: 36438093-36438093 on Build GRCh38
rs373351278 Chr.13: 36438028-36438028 on Build GRCh38
rs774657616 Chr.13: 36437890-36437890 on Build GRCh38
rs368505689 Chr.13: 36437784-36437784 on Build GRCh38
rs111296887 Chr.13: 36438126-36438126 on Build GRCh38
rs780306675 Chr.13: 36437872-36437872 on Build GRCh38
rs759169379 Chr.13: 36438137-36438137 on Build GRCh38
rs373472360 Chr.13: 36438084-36438084 on Build GRCh38
rs765006569 Chr.13: 36438046-36438046 on Build GRCh38
rs764084059 Chr.13: 36438060-36438060 on Build GRCh38
rs61755284 Chr.13: 36437880-36437880 on Build GRCh38
rs201364984 Chr.13: 36437847-36437847 on Build GRCh38
rs760144028 Chr.13: 36437823-36437823 on Build GRCh38
rs754888442 Chr.13: 36437746-36437746 on Build GRCh38
rs949455527 Chr.13: 36437997-36437997 on Build GRCh38
rs767940034 Chr.13: 36437814-36437814 on Build GRCh38
rs976256337 Chr.13: 36437863-36437863 on Build GRCh38
rs923452017 Chr.13: 36437875-36437875 on Build GRCh38
rs749399552 Chr.13: 36438080-36438080 on Build GRCh38
rs771316805 Chr.13: 36438235-36438235 on Build GRCh38
rs947317 Chr.13: 36437907-36437907 on Build GRCh38
rs997896856 Chr.13: 36438144-36438144 on Build GRCh38
rs762791797 Chr.13: 36438164-36438164 on Build GRCh38

View Full Product Details