Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18175203-18175774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFI30
Entrez Gene ID
10437
Gene Name
IFI30, lysosomal thiol reductase
Gene Aliases
GILT, IFI-30, IP-30, IP30
UniGene
Hs.14623
Gene Chromosome Location
Chr.19: 18173780-18178124 on Build GRCh38
Transcripts
NM_006332.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6230820 c.478C>T p.P160S Chr.19: 18175692-18175692 on Build GRCh38
6867410 c.409C>T p.L137F Chr.19: 18175623-18175623 on Build GRCh38
1480787 c.453G>C p.E151D Chr.19: 18175667-18175667 on Build GRCh38
474360 c.369G>A p.E123E Chr.19: 18175364-18175364 on Build GRCh38
993047 c.462C>T p.D154D Chr.19: 18175676-18175676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759037664 Chr.19: 18175739-18175740 on Build GRCh38
rs201952382 Chr.19: 18175409-18175409 on Build GRCh38
rs886775610 Chr.19: 18175584-18175584 on Build GRCh38
rs370041369 Chr.19: 18175699-18175699 on Build GRCh38
rs765389900 Chr.19: 18175724-18175724 on Build GRCh38
rs764749715 Chr.19: 18175311-18175311 on Build GRCh38
rs780969560 Chr.19: 18175628-18175628 on Build GRCh38
rs546234177 Chr.19: 18175480-18175480 on Build GRCh38
rs770734901 Chr.19: 18175412-18175412 on Build GRCh38
rs760502900 Chr.19: 18175265-18175265 on Build GRCh38
rs752652915 Chr.19: 18175731-18175731 on Build GRCh38
rs540952519 Chr.19: 18175586-18175586 on Build GRCh38
rs776598587 Chr.19: 18175269-18175269 on Build GRCh38
rs747340681 Chr.19: 18175242-18175242 on Build GRCh38
rs867891145 Chr.19: 18175623-18175623 on Build GRCh38
rs368484244 Chr.19: 18175499-18175499 on Build GRCh38
rs961607804 Chr.19: 18175678-18175678 on Build GRCh38
rs371344389 Chr.19: 18175639-18175639 on Build GRCh38
rs749320488 Chr.19: 18175390-18175390 on Build GRCh38
rs375989024 Chr.19: 18175697-18175697 on Build GRCh38
rs746681575 Chr.19: 18175397-18175401 on Build GRCh38
rs45514694 Chr.19: 18175434-18175434 on Build GRCh38
rs780289383 Chr.19: 18175381-18175381 on Build GRCh38
rs35554546 Chr.19: 18175409-18175409 on Build GRCh38
rs761325297 Chr.19: 18175575-18175575 on Build GRCh38
rs762609098 Chr.19: 18175320-18175320 on Build GRCh38
rs773697080 Chr.19: 18175667-18175667 on Build GRCh38
rs891632495 Chr.19: 18175578-18175578 on Build GRCh38
rs1038437558 Chr.19: 18175599-18175599 on Build GRCh38
rs769661323 Chr.19: 18175433-18175433 on Build GRCh38
rs781720797 Chr.19: 18175248-18175248 on Build GRCh38
rs957361532 Chr.19: 18175477-18175477 on Build GRCh38
rs868043166 Chr.19: 18175261-18175261 on Build GRCh38
rs942607033 Chr.19: 18175500-18175500 on Build GRCh38
rs773765965 Chr.19: 18175684-18175684 on Build GRCh38
rs770838260 Chr.19: 18175267-18175267 on Build GRCh38
rs1045686375 Chr.19: 18175606-18175606 on Build GRCh38
rs767225140 Chr.19: 18175680-18175680 on Build GRCh38
rs775375851 Chr.19: 18175649-18175649 on Build GRCh38
rs563441583 Chr.19: 18175677-18175677 on Build GRCh38
rs759094134 Chr.19: 18175302-18175302 on Build GRCh38
rs189639826 Chr.19: 18175299-18175299 on Build GRCh38
rs759500684 Chr.19: 18175430-18175430 on Build GRCh38
rs369889024 Chr.19: 18175225-18175225 on Build GRCh38
rs768985984 Chr.19: 18175252-18175252 on Build GRCh38
rs8102682 Chr.19: 18175512-18175512 on Build GRCh38
rs781601256 Chr.19: 18175348-18175348 on Build GRCh38
rs376719198 Chr.19: 18175610-18175610 on Build GRCh38
rs750993043 Chr.19: 18175359-18175359 on Build GRCh38
rs377701391 Chr.19: 18175410-18175410 on Build GRCh38
rs989807648 Chr.19: 18175478-18175478 on Build GRCh38
rs373290769 Chr.19: 18175392-18175392 on Build GRCh38
rs112345548 Chr.19: 18175698-18175698 on Build GRCh38
rs963945597 Chr.19: 18175721-18175721 on Build GRCh38
rs866117127 Chr.19: 18175346-18175346 on Build GRCh38
rs760623241 Chr.19: 18175622-18175622 on Build GRCh38
rs957676056 Chr.19: 18175681-18175681 on Build GRCh38
rs779188416 Chr.19: 18175403-18175403 on Build GRCh38
rs147477761 Chr.19: 18175375-18175375 on Build GRCh38
rs1049410931 Chr.19: 18175509-18175509 on Build GRCh38
rs371677129 Chr.19: 18175232-18175232 on Build GRCh38
rs770506274 Chr.19: 18175401-18175402 on Build GRCh38
rs368679591 Chr.19: 18175331-18175331 on Build GRCh38
rs564510290 Chr.19: 18175486-18175486 on Build GRCh38
rs751204472 Chr.19: 18175566-18175566 on Build GRCh38
rs756698222 Chr.19: 18175582-18175582 on Build GRCh38
rs746818002 Chr.19: 18175406-18175406 on Build GRCh38
rs1044862 Chr.19: 18175652-18175652 on Build GRCh38
rs535343184 Chr.19: 18175233-18175233 on Build GRCh38
rs935754659 Chr.19: 18175524-18175524 on Build GRCh38
rs761995295 Chr.19: 18175391-18175391 on Build GRCh38
rs574328173 Chr.19: 18175601-18175601 on Build GRCh38
rs376296479 Chr.19: 18175251-18175251 on Build GRCh38
rs959329124 Chr.19: 18175653-18175653 on Build GRCh38
rs763696008 Chr.19: 18175555-18175555 on Build GRCh38
rs200373 Chr.19: 18175736-18175736 on Build GRCh38
rs940153143 Chr.19: 18175450-18175450 on Build GRCh38
rs113234467 Chr.19: 18175602-18175602 on Build GRCh38
rs767960035 Chr.19: 18175577-18175577 on Build GRCh38
rs768814819 Chr.19: 18175401-18175401 on Build GRCh38
rs922844654 Chr.19: 18175493-18175493 on Build GRCh38
rs752161587 Chr.19: 18175314-18175314 on Build GRCh38
rs772958305 Chr.19: 18175244-18175245 on Build GRCh38
rs76227216 Chr.19: 18175336-18175336 on Build GRCh38
rs369316180 Chr.19: 18175467-18175467 on Build GRCh38
rs775667220 Chr.19: 18175259-18175259 on Build GRCh38
rs748800457 Chr.19: 18175650-18175650 on Build GRCh38
rs774279559 Chr.19: 18175264-18175264 on Build GRCh38
rs149723974 Chr.19: 18175706-18175706 on Build GRCh38
rs374019360 Chr.19: 18175567-18175567 on Build GRCh38
rs555267237 Chr.19: 18175313-18175313 on Build GRCh38
rs868475246 Chr.19: 18175339-18175339 on Build GRCh38
rs924621902 Chr.19: 18175440-18175440 on Build GRCh38
rs200448181 Chr.19: 18175332-18175332 on Build GRCh38
rs764907828 Chr.19: 18175223-18175223 on Build GRCh38
rs577264122 Chr.19: 18175439-18175439 on Build GRCh38
rs776208108 Chr.19: 18175630-18175630 on Build GRCh38
rs757764865 Chr.19: 18175243-18175243 on Build GRCh38
rs560830984 Chr.19: 18175590-18175590 on Build GRCh38
rs749556308 Chr.19: 18175260-18175260 on Build GRCh38
rs201205841 Chr.19: 18175662-18175662 on Build GRCh38
rs747203127 Chr.19: 18175444-18175445 on Build GRCh38
rs750753249 Chr.19: 18175580-18175580 on Build GRCh38
rs772144480 Chr.19: 18175611-18175611 on Build GRCh38
rs768853099 Chr.19: 18175262-18175262 on Build GRCh38
rs776360467 Chr.19: 18175425-18175425 on Build GRCh38

View Full Product Details