Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 77006895-77007462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL3
Entrez Gene ID
26224
Gene Name
F-box and leucine rich repeat protein 3
Gene Aliases
FBL3, FBL3A, FBXL3A
UniGene
Hs.508284
Gene Chromosome Location
Chr.13: 76992597-77027196 on Build GRCh38
Transcripts
NM_012158.2 XM_005266336.1 XM_005266337.2 XM_017020538.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5559498 c.1190T>C p.L397P Chr.13: 77007242-77007242 on Build GRCh38
948484 c.1129G>A p.V377I Chr.13: 77007303-77007303 on Build GRCh38
6661956 c.1050G>A p.E350E Chr.13: 77007382-77007382 on Build GRCh38
6661955 c.1241A>G p.K414R Chr.13: 77007191-77007191 on Build GRCh38
948483 c.1157G>A p.R386H Chr.13: 77007275-77007275 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78684417 Chr.13: 77007196-77007196 on Build GRCh38
rs558475664 Chr.13: 77006971-77006971 on Build GRCh38
rs763008112 Chr.13: 77007095-77007100 on Build GRCh38
rs1039635264 Chr.13: 77007069-77007069 on Build GRCh38
rs764434376 Chr.13: 77007403-77007403 on Build GRCh38
rs769422396 Chr.13: 77007115-77007115 on Build GRCh38
rs960311139 Chr.13: 77006977-77006977 on Build GRCh38
rs775587973 Chr.13: 77007397-77007397 on Build GRCh38
rs1003137044 Chr.13: 77006965-77006965 on Build GRCh38
rs372087180 Chr.13: 77007256-77007256 on Build GRCh38
rs755174479 Chr.13: 77007379-77007381 on Build GRCh38
rs777828187 Chr.13: 77007245-77007245 on Build GRCh38
rs772342610 Chr.13: 77007165-77007165 on Build GRCh38
rs906070088 Chr.13: 77006949-77006950 on Build GRCh38
rs200610714 Chr.13: 77007166-77007166 on Build GRCh38
rs200042699 Chr.13: 77007103-77007103 on Build GRCh38
rs570586287 Chr.13: 77007002-77007002 on Build GRCh38
rs144603004 Chr.13: 77006968-77006968 on Build GRCh38
rs775283266 Chr.13: 77007133-77007133 on Build GRCh38
rs756988737 Chr.13: 77007275-77007275 on Build GRCh38
rs991758091 Chr.13: 77007001-77007001 on Build GRCh38
rs766450941 Chr.13: 77007095-77007097 on Build GRCh38
rs545836683 Chr.13: 77007134-77007134 on Build GRCh38
rs967480573 Chr.13: 77006928-77006928 on Build GRCh38
rs929761365 Chr.13: 77007013-77007013 on Build GRCh38
rs201346194 Chr.13: 77007114-77007114 on Build GRCh38
rs752940172 Chr.13: 77007234-77007234 on Build GRCh38
rs765244259 Chr.13: 77007223-77007223 on Build GRCh38
rs779659265 Chr.13: 77007324-77007324 on Build GRCh38
rs201176626 Chr.13: 77007316-77007316 on Build GRCh38
rs781255853 Chr.13: 77007277-77007277 on Build GRCh38
rs897174506 Chr.13: 77006980-77006980 on Build GRCh38
rs973564068 Chr.13: 77007005-77007005 on Build GRCh38
rs745444479 Chr.13: 77007375-77007375 on Build GRCh38
rs77759197 Chr.13: 77007033-77007033 on Build GRCh38
rs559604554 Chr.13: 77007351-77007351 on Build GRCh38
rs745731884 Chr.13: 77007289-77007289 on Build GRCh38
rs751773661 Chr.13: 77007250-77007265 on Build GRCh38
rs762998083 Chr.13: 77007398-77007398 on Build GRCh38
rs541827768 Chr.13: 77007044-77007044 on Build GRCh38
rs76487761 Chr.13: 77007336-77007336 on Build GRCh38
rs375666314 Chr.13: 77007326-77007326 on Build GRCh38
rs117178661 Chr.13: 77007010-77007010 on Build GRCh38
rs770294696 Chr.13: 77007068-77007068 on Build GRCh38
rs767543712 Chr.13: 77007159-77007159 on Build GRCh38
rs771136982 Chr.13: 77007342-77007342 on Build GRCh38
rs759989436 Chr.13: 77007363-77007363 on Build GRCh38
rs758580860 Chr.13: 77007241-77007241 on Build GRCh38
rs534629250 Chr.13: 77007004-77007004 on Build GRCh38
rs1044626323 Chr.13: 77006947-77006947 on Build GRCh38
rs760929977 Chr.13: 77007173-77007173 on Build GRCh38
rs914723333 Chr.13: 77006918-77006918 on Build GRCh38
rs766971971 Chr.13: 77007184-77007184 on Build GRCh38
rs759619363 Chr.13: 77007208-77007208 on Build GRCh38
rs751724560 Chr.13: 77007439-77007439 on Build GRCh38
rs761173135 Chr.13: 77007340-77007340 on Build GRCh38
rs148273762 Chr.13: 77007337-77007337 on Build GRCh38
rs890797220 Chr.13: 77007034-77007034 on Build GRCh38
rs367644997 Chr.13: 77006931-77006931 on Build GRCh38
rs769440607 Chr.13: 77007319-77007319 on Build GRCh38
rs1017439287 Chr.13: 77007058-77007060 on Build GRCh38
rs869051475 Chr.13: 77007404-77007404 on Build GRCh38
rs942217430 Chr.13: 77007003-77007003 on Build GRCh38

View Full Product Details