Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 108353190-108353781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HHLA2
Entrez Gene ID
11148
Gene Name
HERV-H LTR-associating 2
Gene Aliases
B7-H5, B7-H7, B7H7, B7y
UniGene
Hs.225968
Gene Chromosome Location
Chr.3: 108296490-108378285 on Build GRCh38
Transcripts
NM_001282556.1 NM_001282557.1 NM_001282558.1 NM_001282559.1 NM_007072.3 XM_005247080.3 XM_011512362.1 XM_011512363.1 XM_011512364.1 XM_011512367.2 XM_017005642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4979608 c.266C>A p.A89E Chr.3: 108353628-108353628 on Build GRCh38
2916728 c.311C>T p.A104V Chr.3: 108353673-108353673 on Build GRCh38
2916732 c.346G>A p.E116K Chr.3: 108353708-108353708 on Build GRCh38
2916725 c.291T>C p.N97N Chr.3: 108353653-108353653 on Build GRCh38
7327393 c.200_201insTAGC p.Y69fs*1 Chr.3: 108353562-108353563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778652084 Chr.3: 108353462-108353462 on Build GRCh38
rs140364028 Chr.3: 108353540-108353540 on Build GRCh38
rs6779254 Chr.3: 108353451-108353451 on Build GRCh38
rs746832212 Chr.3: 108353476-108353476 on Build GRCh38
rs1055612507 Chr.3: 108353534-108353534 on Build GRCh38
rs975600863 Chr.3: 108353411-108353411 on Build GRCh38
rs752349830 Chr.3: 108353568-108353568 on Build GRCh38
rs774696499 Chr.3: 108353751-108353751 on Build GRCh38
rs371274078 Chr.3: 108353707-108353707 on Build GRCh38
rs375560061 Chr.3: 108353634-108353634 on Build GRCh38
rs915734735 Chr.3: 108353373-108353373 on Build GRCh38
rs367817011 Chr.3: 108353708-108353708 on Build GRCh38
rs1006203038 Chr.3: 108353500-108353500 on Build GRCh38
rs761161559 Chr.3: 108353212-108353212 on Build GRCh38
rs768562499 Chr.3: 108353296-108353296 on Build GRCh38
rs750264144 Chr.3: 108353720-108353720 on Build GRCh38
rs58233893 Chr.3: 108353482-108353482 on Build GRCh38
rs529096683 Chr.3: 108353510-108353510 on Build GRCh38
rs763792651 Chr.3: 108353704-108353704 on Build GRCh38
rs531586600 Chr.3: 108353579-108353579 on Build GRCh38
rs554087388 Chr.3: 108353331-108353331 on Build GRCh38
rs770873611 Chr.3: 108353477-108353477 on Build GRCh38
rs945683591 Chr.3: 108353484-108353484 on Build GRCh38
rs984797711 Chr.3: 108353222-108353222 on Build GRCh38
rs771206254 Chr.3: 108353673-108353673 on Build GRCh38
rs748266514 Chr.3: 108353641-108353641 on Build GRCh38
rs773595188 Chr.3: 108353512-108353512 on Build GRCh38
rs374382407 Chr.3: 108353470-108353470 on Build GRCh38
rs781060461 Chr.3: 108353481-108353481 on Build GRCh38
rs748243757 Chr.3: 108353739-108353739 on Build GRCh38
rs200786656 Chr.3: 108353387-108353387 on Build GRCh38
rs557647742 Chr.3: 108353511-108353513 on Build GRCh38
rs777238594 Chr.3: 108353674-108353674 on Build GRCh38
rs767952028 Chr.3: 108353442-108353442 on Build GRCh38
rs778938592 Chr.3: 108353636-108353636 on Build GRCh38
rs531245052 Chr.3: 108353227-108353227 on Build GRCh38
rs762041589 Chr.3: 108353506-108353506 on Build GRCh38
rs375854823 Chr.3: 108353721-108353721 on Build GRCh38
rs2124740 Chr.3: 108353677-108353677 on Build GRCh38
rs761010314 Chr.3: 108353520-108353520 on Build GRCh38
rs773361909 Chr.3: 108353659-108353659 on Build GRCh38
rs745834506 Chr.3: 108353625-108353625 on Build GRCh38
rs761222441 Chr.3: 108353448-108353448 on Build GRCh38
rs766894660 Chr.3: 108353522-108353522 on Build GRCh38
rs536836404 Chr.3: 108353389-108353389 on Build GRCh38
rs571255744 Chr.3: 108353297-108353297 on Build GRCh38
rs762439322 Chr.3: 108353703-108353703 on Build GRCh38
rs753383127 Chr.3: 108353456-108353456 on Build GRCh38
rs997718374 Chr.3: 108353480-108353480 on Build GRCh38
rs35920413 Chr.3: 108353322-108353323 on Build GRCh38
rs2124741 Chr.3: 108353578-108353578 on Build GRCh38
rs772333265 Chr.3: 108353644-108353644 on Build GRCh38
rs1014692894 Chr.3: 108353254-108353254 on Build GRCh38
rs533857825 Chr.3: 108353324-108353324 on Build GRCh38
rs777649283 Chr.3: 108353474-108353474 on Build GRCh38
rs763063658 Chr.3: 108353409-108353409 on Build GRCh38
rs772232203 Chr.3: 108353507-108353507 on Build GRCh38
rs200858373 Chr.3: 108353754-108353754 on Build GRCh38
rs372159295 Chr.3: 108353632-108353632 on Build GRCh38
rs541551489 Chr.3: 108353471-108353471 on Build GRCh38
rs759052527 Chr.3: 108353539-108353539 on Build GRCh38
rs781114195 Chr.3: 108353615-108353615 on Build GRCh38
rs558884475 Chr.3: 108353441-108353441 on Build GRCh38
rs758419765 Chr.3: 108353750-108353750 on Build GRCh38
rs34201681 Chr.3: 108353646-108353646 on Build GRCh38
rs777334460 Chr.3: 108353603-108353603 on Build GRCh38
rs887108990 Chr.3: 108353640-108353640 on Build GRCh38
rs1039217168 Chr.3: 108353466-108353466 on Build GRCh38
rs780032909 Chr.3: 108353733-108353733 on Build GRCh38
rs6766283 Chr.3: 108353396-108353396 on Build GRCh38
rs771471810 Chr.3: 108353681-108353681 on Build GRCh38
rs761040628 Chr.3: 108353672-108353672 on Build GRCh38
rs78607433 Chr.3: 108353268-108353268 on Build GRCh38
rs370821029 Chr.3: 108353533-108353533 on Build GRCh38
rs371152582 Chr.3: 108353385-108353385 on Build GRCh38
rs544431628 Chr.3: 108353422-108353423 on Build GRCh38
rs750842514 Chr.3: 108353447-108353447 on Build GRCh38
rs991704235 Chr.3: 108353287-108353287 on Build GRCh38
rs774512569 Chr.3: 108353497-108353497 on Build GRCh38
rs958803105 Chr.3: 108353251-108353251 on Build GRCh38
rs778772789 Chr.3: 108353380-108353380 on Build GRCh38
rs909249342 Chr.3: 108353264-108353264 on Build GRCh38
rs751367823 Chr.3: 108353610-108353610 on Build GRCh38
rs199766070 Chr.3: 108353440-108353440 on Build GRCh38
rs1034879827 Chr.3: 108353247-108353247 on Build GRCh38
rs879624681 Chr.3: 108353518-108353518 on Build GRCh38
rs576770303 Chr.3: 108353402-108353402 on Build GRCh38
rs764298694 Chr.3: 108353424-108353424 on Build GRCh38
rs772956692 Chr.3: 108353388-108353388 on Build GRCh38
rs759459866 Chr.3: 108353463-108353463 on Build GRCh38
rs767318616 Chr.3: 108353710-108353710 on Build GRCh38
rs571187309 Chr.3: 108353698-108353698 on Build GRCh38

View Full Product Details