Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150708469-150709036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTN4
Entrez Gene ID
51164
Gene Name
dynactin subunit 4
Gene Aliases
DYN4, P62
UniGene
Hs.675564
Gene Chromosome Location
Chr.5: 150708747-150759095 on Build GRCh38
Transcripts
NM_001135643.1 NM_001135644.1 NM_016221.3 XM_005268457.1 XM_011537644.1 XM_011537645.1 XM_011537646.1 XM_011537647.1

SNP Detail

SNP ID Chromosome Location
rs972952257 Chr.5: 150708653-150708653 on Build GRCh38
rs933441561 Chr.5: 150708728-150708728 on Build GRCh38
rs1057482322 Chr.5: 150708999-150708999 on Build GRCh38
rs78349754 Chr.5: 150708647-150708647 on Build GRCh38
rs1042262542 Chr.5: 150708820-150708820 on Build GRCh38
rs987024050 Chr.5: 150708740-150708740 on Build GRCh38
rs542623985 Chr.5: 150708682-150708682 on Build GRCh38
rs997412271 Chr.5: 150708572-150708572 on Build GRCh38
rs750220087 Chr.5: 150709001-150709001 on Build GRCh38
rs951023158 Chr.5: 150708643-150708643 on Build GRCh38
rs901616602 Chr.5: 150708775-150708775 on Build GRCh38
rs1025073241 Chr.5: 150708630-150708630 on Build GRCh38
rs1022729656 Chr.5: 150708616-150708616 on Build GRCh38
rs765314740 Chr.5: 150708959-150708959 on Build GRCh38
rs538238894 Chr.5: 150708726-150708726 on Build GRCh38
rs1032346727 Chr.5: 150708593-150708593 on Build GRCh38
rs921060691 Chr.5: 150708529-150708529 on Build GRCh38
rs10072365 Chr.5: 150708557-150708557 on Build GRCh38
rs963352750 Chr.5: 150708668-150708668 on Build GRCh38
rs928950166 Chr.5: 150708505-150708505 on Build GRCh38
rs932488892 Chr.5: 150708533-150708533 on Build GRCh38
rs1000796503 Chr.5: 150708859-150708859 on Build GRCh38
rs550069664 Chr.5: 150708748-150708748 on Build GRCh38
rs1038854029 Chr.5: 150708556-150708556 on Build GRCh38
rs1012224544 Chr.5: 150708899-150708899 on Build GRCh38
rs533178913 Chr.5: 150708625-150708625 on Build GRCh38
rs905591184 Chr.5: 150708974-150708974 on Build GRCh38
rs187005028 Chr.5: 150708929-150708929 on Build GRCh38
rs745917926 Chr.5: 150708817-150708817 on Build GRCh38
rs1038822642 Chr.5: 150708731-150708731 on Build GRCh38
rs138212469 Chr.5: 150708656-150708656 on Build GRCh38
rs10476752 Chr.5: 150708578-150708578 on Build GRCh38
rs908887343 Chr.5: 150708684-150708684 on Build GRCh38
rs933136850 Chr.5: 150708793-150708793 on Build GRCh38
rs113119595 Chr.5: 150708618-150708618 on Build GRCh38
rs1035886308 Chr.5: 150708633-150708633 on Build GRCh38
rs556730682 Chr.5: 150708883-150708883 on Build GRCh38
rs1044146301 Chr.5: 150708945-150708945 on Build GRCh38
rs887524198 Chr.5: 150708527-150708527 on Build GRCh38
rs773621703 Chr.5: 150708552-150708552 on Build GRCh38
rs1021273715 Chr.5: 150708575-150708575 on Build GRCh38
rs970818458 Chr.5: 150708970-150708970 on Build GRCh38
rs559758916 Chr.5: 150708602-150708602 on Build GRCh38
rs752734850 Chr.5: 150708759-150708759 on Build GRCh38
rs990875664 Chr.5: 150708607-150708607 on Build GRCh38
rs912430497 Chr.5: 150708756-150708756 on Build GRCh38
rs547591203 Chr.5: 150708590-150708590 on Build GRCh38
rs984825793 Chr.5: 150708683-150708683 on Build GRCh38
rs763172136 Chr.5: 150708810-150708810 on Build GRCh38
rs538124691 Chr.5: 150708730-150708730 on Build GRCh38
rs10476941 Chr.5: 150708519-150708519 on Build GRCh38
rs902389427 Chr.5: 150708833-150708833 on Build GRCh38
rs12232 Chr.5: 150708983-150708983 on Build GRCh38
rs760147954 Chr.5: 150708677-150708677 on Build GRCh38
rs1032728265 Chr.5: 150708881-150708881 on Build GRCh38
rs922037202 Chr.5: 150708684-150708687 on Build GRCh38

View Full Product Details