Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122879286-122879864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIRC2
Entrez Gene ID
84925
Gene Name
disrupted in renal carcinoma 2
Gene Aliases
RCC4
UniGene
Hs.477346
Gene Chromosome Location
Chr.3: 122795054-122892157 on Build GRCh38
Transcripts
NM_032839.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3682934 c.1394A>G p.E465G Chr.3: 122879335-122879335 on Build GRCh38
4640480 c.1356G>A p.S452S Chr.3: 122879297-122879297 on Build GRCh38
3682933 c.1383G>T p.L461L Chr.3: 122879324-122879324 on Build GRCh38
3682932 c.1376T>C p.L459P Chr.3: 122879317-122879317 on Build GRCh38
6644254 c.1350C>T p.P450P Chr.3: 122879291-122879291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868355529 Chr.3: 122879468-122879468 on Build GRCh38
rs1045244789 Chr.3: 122879348-122879348 on Build GRCh38
rs1021913447 Chr.3: 122879321-122879321 on Build GRCh38
rs915478944 Chr.3: 122879750-122879750 on Build GRCh38
rs542730908 Chr.3: 122879409-122879409 on Build GRCh38
rs963043877 Chr.3: 122879582-122879582 on Build GRCh38
rs377561119 Chr.3: 122879642-122879642 on Build GRCh38
rs975245130 Chr.3: 122879476-122879476 on Build GRCh38
rs748827762 Chr.3: 122879389-122879389 on Build GRCh38
rs536481994 Chr.3: 122879822-122879822 on Build GRCh38
rs953448119 Chr.3: 122879496-122879496 on Build GRCh38
rs760030614 Chr.3: 122879654-122879654 on Build GRCh38
rs757007238 Chr.3: 122879353-122879353 on Build GRCh38
rs758581334 Chr.3: 122879335-122879335 on Build GRCh38
rs559532402 Chr.3: 122879439-122879439 on Build GRCh38
rs916461877 Chr.3: 122879600-122879600 on Build GRCh38
rs771039278 Chr.3: 122879425-122879425 on Build GRCh38
rs983204119 Chr.3: 122879535-122879535 on Build GRCh38
rs747391740 Chr.3: 122879520-122879520 on Build GRCh38
rs745669516 Chr.3: 122879391-122879393 on Build GRCh38
rs745780227 Chr.3: 122879373-122879373 on Build GRCh38
rs1029511414 Chr.3: 122879480-122879480 on Build GRCh38
rs573785087 Chr.3: 122879312-122879312 on Build GRCh38
rs947392371 Chr.3: 122879840-122879840 on Build GRCh38
rs138397591 Chr.3: 122879361-122879361 on Build GRCh38
rs996724929 Chr.3: 122879435-122879435 on Build GRCh38
rs1014536889 Chr.3: 122879575-122879575 on Build GRCh38
rs749369777 Chr.3: 122879378-122879378 on Build GRCh38
rs1053225525 Chr.3: 122879766-122879766 on Build GRCh38
rs779669415 Chr.3: 122879386-122879386 on Build GRCh38
rs962566256 Chr.3: 122879471-122879471 on Build GRCh38
rs982316715 Chr.3: 122879657-122879657 on Build GRCh38
rs954572926 Chr.3: 122879462-122879462 on Build GRCh38
rs2288677 Chr.3: 122879444-122879444 on Build GRCh38
rs777996216 Chr.3: 122879343-122879343 on Build GRCh38
rs564737609 Chr.3: 122879559-122879559 on Build GRCh38
rs935328833 Chr.3: 122879730-122879730 on Build GRCh38
rs751314698 Chr.3: 122879345-122879345 on Build GRCh38
rs868806089 Chr.3: 122879325-122879325 on Build GRCh38
rs973277384 Chr.3: 122879585-122879585 on Build GRCh38
rs904911305 Chr.3: 122879354-122879354 on Build GRCh38
rs946886862 Chr.3: 122879823-122879823 on Build GRCh38
rs778968768 Chr.3: 122879349-122879350 on Build GRCh38
rs552699063 Chr.3: 122879820-122879820 on Build GRCh38
rs189105188 Chr.3: 122879813-122879813 on Build GRCh38
rs753490023 Chr.3: 122879309-122879309 on Build GRCh38
rs551012272 Chr.3: 122879667-122879667 on Build GRCh38
rs772569290 Chr.3: 122879824-122879824 on Build GRCh38
rs144223930 Chr.3: 122879530-122879530 on Build GRCh38
rs988373686 Chr.3: 122879749-122879749 on Build GRCh38

View Full Product Details