Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 125966696-125967265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HINT3
Entrez Gene ID
135114
Gene Name
histidine triad nucleotide binding protein 3
Gene Aliases
HINT4
UniGene
Hs.72325
Gene Chromosome Location
Chr.6: 125956715-125980244 on Build GRCh38
Transcripts
NM_138571.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1440265 c.226G>T p.D76Y Chr.6: 125966911-125966911 on Build GRCh38
6679951 c.319G>A p.V107I Chr.6: 125967004-125967004 on Build GRCh38
1440264 c.208G>A p.D70N Chr.6: 125966893-125966893 on Build GRCh38
6470724 c.287G>A p.C96Y Chr.6: 125966972-125966972 on Build GRCh38
7425069 c.203A>T p.N68I Chr.6: 125966888-125966888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761844196 Chr.6: 125966738-125966738 on Build GRCh38
rs766130928 Chr.6: 125966972-125966972 on Build GRCh38
rs141800078 Chr.6: 125966767-125966767 on Build GRCh38
rs752349900 Chr.6: 125966888-125966888 on Build GRCh38
rs755863003 Chr.6: 125966892-125966892 on Build GRCh38
rs1036273899 Chr.6: 125966807-125966807 on Build GRCh38
rs778183165 Chr.6: 125967039-125967040 on Build GRCh38
rs540381080 Chr.6: 125967239-125967239 on Build GRCh38
rs772622280 Chr.6: 125966915-125966915 on Build GRCh38
rs759418363 Chr.6: 125966976-125966976 on Build GRCh38
rs3757212 Chr.6: 125966877-125966877 on Build GRCh38
rs181029766 Chr.6: 125967052-125967052 on Build GRCh38
rs758877327 Chr.6: 125967177-125967177 on Build GRCh38
rs76787419 Chr.6: 125966779-125966779 on Build GRCh38
rs1016565478 Chr.6: 125967235-125967235 on Build GRCh38
rs369239656 Chr.6: 125966933-125966933 on Build GRCh38
rs185919066 Chr.6: 125967100-125967100 on Build GRCh38
rs919043246 Chr.6: 125966878-125966878 on Build GRCh38
rs1013068175 Chr.6: 125967225-125967225 on Build GRCh38
rs903155075 Chr.6: 125967065-125967065 on Build GRCh38
rs755772437 Chr.6: 125966996-125966996 on Build GRCh38
rs756730822 Chr.6: 125967010-125967010 on Build GRCh38
rs971468743 Chr.6: 125967204-125967204 on Build GRCh38
rs538597571 Chr.6: 125966825-125966825 on Build GRCh38
rs766350656 Chr.6: 125966859-125966859 on Build GRCh38
rs530528813 Chr.6: 125966730-125966730 on Build GRCh38
rs779355453 Chr.6: 125966895-125966895 on Build GRCh38
rs1020347157 Chr.6: 125966764-125966764 on Build GRCh38
rs762807572 Chr.6: 125966850-125966850 on Build GRCh38
rs554638960 Chr.6: 125966962-125966962 on Build GRCh38
rs747499771 Chr.6: 125967056-125967056 on Build GRCh38
rs544774648 Chr.6: 125967039-125967039 on Build GRCh38
rs946238436 Chr.6: 125966884-125966884 on Build GRCh38
rs1011517979 Chr.6: 125966761-125966761 on Build GRCh38
rs750392115 Chr.6: 125967136-125967136 on Build GRCh38
rs201833844 Chr.6: 125966974-125966974 on Build GRCh38
rs950120228 Chr.6: 125966762-125966762 on Build GRCh38
rs947428106 Chr.6: 125967034-125967034 on Build GRCh38
rs909451735 Chr.6: 125966836-125966836 on Build GRCh38
rs758756024 Chr.6: 125967019-125967019 on Build GRCh38
rs780620774 Chr.6: 125966879-125966879 on Build GRCh38
rs963066053 Chr.6: 125966839-125966839 on Build GRCh38
rs190819958 Chr.6: 125967178-125967178 on Build GRCh38
rs145990439 Chr.6: 125967009-125967009 on Build GRCh38
rs548666320 Chr.6: 125966741-125966741 on Build GRCh38
rs933072922 Chr.6: 125966908-125966908 on Build GRCh38
rs767014894 Chr.6: 125966991-125966991 on Build GRCh38
rs776593581 Chr.6: 125966960-125966960 on Build GRCh38
rs200029047 Chr.6: 125967010-125967010 on Build GRCh38
rs567375048 Chr.6: 125966742-125966742 on Build GRCh38
rs753610538 Chr.6: 125967011-125967011 on Build GRCh38
rs751521535 Chr.6: 125966872-125966872 on Build GRCh38
rs200045236 Chr.6: 125966834-125966834 on Build GRCh38
rs114751741 Chr.6: 125966963-125966963 on Build GRCh38
rs746250111 Chr.6: 125966914-125966914 on Build GRCh38
rs762735911 Chr.6: 125966971-125966971 on Build GRCh38
rs752259987 Chr.6: 125966995-125966995 on Build GRCh38
rs747510663 Chr.6: 125966920-125966920 on Build GRCh38
rs571096799 Chr.6: 125966781-125966781 on Build GRCh38
rs377194909 Chr.6: 125966918-125966918 on Build GRCh38
rs1003389802 Chr.6: 125967078-125967078 on Build GRCh38

View Full Product Details