Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16697801-16698366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BZW2
Entrez Gene ID
28969
Gene Name
basic leucine zipper and W2 domains 2
Gene Aliases
HSPC028, MST017, MSTP017
UniGene
Hs.487635
Gene Chromosome Location
Chr.7: 16646134-16706523 on Build GRCh38
Transcripts
NM_001159767.1 NM_014038.2 NR_027624.1 XM_006715706.1 XM_006715707.1 XM_006715708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4445106 c.989C>T p.A330V Chr.7: 16698067-16698067 on Build GRCh38
1329914 c.1106A>T p.K369I Chr.7: 16698184-16698184 on Build GRCh38
3636915 c.1077C>T p.A359A Chr.7: 16698155-16698155 on Build GRCh38
88508 c.1096C>G p.L366V Chr.7: 16698174-16698174 on Build GRCh38
351581 c.1020G>C p.L340L Chr.7: 16698098-16698098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758867714 Chr.7: 16698028-16698028 on Build GRCh38
rs146163758 Chr.7: 16698117-16698117 on Build GRCh38
rs760929671 Chr.7: 16698042-16698042 on Build GRCh38
rs532940580 Chr.7: 16697948-16697948 on Build GRCh38
rs981451750 Chr.7: 16697892-16697892 on Build GRCh38
rs962306093 Chr.7: 16698041-16698041 on Build GRCh38
rs763593269 Chr.7: 16698196-16698196 on Build GRCh38
rs759622263 Chr.7: 16698160-16698160 on Build GRCh38
rs778750516 Chr.7: 16698137-16698137 on Build GRCh38
rs753174373 Chr.7: 16698079-16698079 on Build GRCh38
rs1040675641 Chr.7: 16698237-16698237 on Build GRCh38
rs537849237 Chr.7: 16698143-16698143 on Build GRCh38
rs377279314 Chr.7: 16698034-16698034 on Build GRCh38
rs1040705452 Chr.7: 16697989-16697989 on Build GRCh38
rs370117093 Chr.7: 16698208-16698208 on Build GRCh38
rs758390217 Chr.7: 16698255-16698255 on Build GRCh38
rs933981166 Chr.7: 16698191-16698191 on Build GRCh38
rs112938667 Chr.7: 16698031-16698031 on Build GRCh38
rs749277154 Chr.7: 16698128-16698128 on Build GRCh38
rs755577939 Chr.7: 16698000-16698000 on Build GRCh38
rs200898944 Chr.7: 16698021-16698021 on Build GRCh38
rs770995466 Chr.7: 16698044-16698044 on Build GRCh38
rs769784877 Chr.7: 16698171-16698171 on Build GRCh38
rs764599893 Chr.7: 16698082-16698082 on Build GRCh38
rs764036602 Chr.7: 16698335-16698335 on Build GRCh38
rs922695064 Chr.7: 16697928-16697928 on Build GRCh38
rs753048227 Chr.7: 16698008-16698008 on Build GRCh38
rs151037210 Chr.7: 16697880-16697880 on Build GRCh38
rs116196420 Chr.7: 16697995-16697995 on Build GRCh38
rs759549226 Chr.7: 16698062-16698062 on Build GRCh38
rs943679886 Chr.7: 16697984-16697984 on Build GRCh38
rs934036645 Chr.7: 16697933-16697933 on Build GRCh38
rs780551598 Chr.7: 16698100-16698100 on Build GRCh38
rs747219700 Chr.7: 16698141-16698141 on Build GRCh38
rs762278127 Chr.7: 16698201-16698201 on Build GRCh38
rs946607861 Chr.7: 16698321-16698321 on Build GRCh38
rs1035719877 Chr.7: 16698059-16698059 on Build GRCh38
rs767127508 Chr.7: 16698207-16698207 on Build GRCh38
rs889647658 Chr.7: 16697976-16697976 on Build GRCh38
rs541381109 Chr.7: 16697907-16697907 on Build GRCh38
rs947292559 Chr.7: 16697922-16697922 on Build GRCh38
rs200088139 Chr.7: 16698011-16698011 on Build GRCh38
rs776042480 Chr.7: 16698177-16698177 on Build GRCh38
rs531616294 Chr.7: 16697860-16697860 on Build GRCh38
rs962004378 Chr.7: 16697883-16697883 on Build GRCh38
rs780793161 Chr.7: 16698016-16698016 on Build GRCh38
rs1006363894 Chr.7: 16698317-16698317 on Build GRCh38
rs888205735 Chr.7: 16698304-16698304 on Build GRCh38
rs577663998 Chr.7: 16698301-16698301 on Build GRCh38
rs932311796 Chr.7: 16698267-16698267 on Build GRCh38
rs373283505 Chr.7: 16698068-16698068 on Build GRCh38
rs776373034 Chr.7: 16697912-16697912 on Build GRCh38
rs538394707 Chr.7: 16698341-16698341 on Build GRCh38
rs556246742 Chr.7: 16698283-16698283 on Build GRCh38
rs764548319 Chr.7: 16698198-16698198 on Build GRCh38
rs375769865 Chr.7: 16698069-16698069 on Build GRCh38
rs779473598 Chr.7: 16698228-16698228 on Build GRCh38
rs984418284 Chr.7: 16697828-16697828 on Build GRCh38
rs911296113 Chr.7: 16698303-16698303 on Build GRCh38
rs140913839 Chr.7: 16697994-16697994 on Build GRCh38
rs776722672 Chr.7: 16698146-16698146 on Build GRCh38
rs1041596850 Chr.7: 16697957-16697957 on Build GRCh38
rs757033515 Chr.7: 16698089-16698089 on Build GRCh38
rs773916035 Chr.7: 16697854-16697854 on Build GRCh38
rs768410846 Chr.7: 16698131-16698131 on Build GRCh38
rs769824877 Chr.7: 16698020-16698020 on Build GRCh38
rs746681432 Chr.7: 16698012-16698012 on Build GRCh38
rs753774025 Chr.7: 16698234-16698234 on Build GRCh38
rs977672076 Chr.7: 16698185-16698185 on Build GRCh38
rs551552541 Chr.7: 16697979-16697979 on Build GRCh38
rs755632518 Chr.7: 16698211-16698211 on Build GRCh38
rs369350638 Chr.7: 16698036-16698036 on Build GRCh38
rs144381179 Chr.7: 16698080-16698080 on Build GRCh38
rs776831816 Chr.7: 16698045-16698045 on Build GRCh38
rs937429043 Chr.7: 16697906-16697906 on Build GRCh38
rs914447584 Chr.7: 16697915-16697915 on Build GRCh38
rs765507370 Chr.7: 16698002-16698002 on Build GRCh38
rs755580498 Chr.7: 16698112-16698112 on Build GRCh38
rs751223933 Chr.7: 16698087-16698087 on Build GRCh38
rs899546986 Chr.7: 16698250-16698250 on Build GRCh38
rs879487814 Chr.7: 16697882-16697882 on Build GRCh38
rs35787234 Chr.7: 16698318-16698319 on Build GRCh38
rs150229532 Chr.7: 16698014-16698014 on Build GRCh38
rs565050965 Chr.7: 16697903-16697903 on Build GRCh38
rs981420360 Chr.7: 16698046-16698046 on Build GRCh38

View Full Product Details