Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55997279-55997530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBAS
Entrez Gene ID
2631
Gene Name
glioblastoma amplified sequence
Gene Aliases
NIPSNAP2
UniGene
Hs.591069
Gene Chromosome Location
Chr.7: 55964577-56000182 on Build GRCh38
Transcripts
NM_001202469.1 NM_001483.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1090941 c.745C>T p.R249W Chr.7: 55997398-55997398 on Build GRCh38
1090940 c.739G>T p.D247Y Chr.7: 55997392-55997392 on Build GRCh38
5611774 c.739G>C p.D247H Chr.7: 55997392-55997392 on Build GRCh38
4960972 c.734G>A p.R245K Chr.7: 55997387-55997387 on Build GRCh38
3923851 c.715T>G p.Y239D Chr.7: 55997368-55997368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565250275 Chr.7: 55997409-55997409 on Build GRCh38
rs745369632 Chr.7: 55997489-55997489 on Build GRCh38
rs951783942 Chr.7: 55997436-55997436 on Build GRCh38
rs769641960 Chr.7: 55997397-55997397 on Build GRCh38
rs758778951 Chr.7: 55997341-55997341 on Build GRCh38
rs771842081 Chr.7: 55997377-55997377 on Build GRCh38
rs770959617 Chr.7: 55997499-55997499 on Build GRCh38
rs753136830 Chr.7: 55997467-55997467 on Build GRCh38
rs1020996949 Chr.7: 55997325-55997325 on Build GRCh38
rs747130465 Chr.7: 55997338-55997341 on Build GRCh38
rs745396781 Chr.7: 55997371-55997371 on Build GRCh38
rs755460002 Chr.7: 55997358-55997358 on Build GRCh38
rs201431113 Chr.7: 55997461-55997461 on Build GRCh38
rs540734347 Chr.7: 55997362-55997362 on Build GRCh38
rs765271010 Chr.7: 55997329-55997329 on Build GRCh38
rs778633198 Chr.7: 55997482-55997482 on Build GRCh38
rs773103909 Chr.7: 55997445-55997445 on Build GRCh38
rs755370206 Chr.7: 55997465-55997465 on Build GRCh38
rs746992455 Chr.7: 55997498-55997498 on Build GRCh38
rs775475871 Chr.7: 55997398-55997398 on Build GRCh38
rs778238159 Chr.7: 55997370-55997370 on Build GRCh38
rs573332960 Chr.7: 55997310-55997310 on Build GRCh38
rs1019268339 Chr.7: 55997317-55997317 on Build GRCh38
rs768490559 Chr.7: 55997429-55997429 on Build GRCh38
rs779947090 Chr.7: 55997497-55997497 on Build GRCh38
rs111594303 Chr.7: 55997432-55997432 on Build GRCh38
rs748990956 Chr.7: 55997361-55997361 on Build GRCh38
rs766988196 Chr.7: 55997457-55997457 on Build GRCh38
rs971910165 Chr.7: 55997392-55997392 on Build GRCh38
rs765514799 Chr.7: 55997444-55997444 on Build GRCh38
rs766919038 Chr.7: 55997342-55997342 on Build GRCh38
rs767955308 Chr.7: 55997466-55997466 on Build GRCh38
rs757988427 Chr.7: 55997496-55997496 on Build GRCh38
rs750544184 Chr.7: 55997340-55997340 on Build GRCh38
rs915614266 Chr.7: 55997475-55997475 on Build GRCh38
rs761933729 Chr.7: 55997323-55997323 on Build GRCh38
rs1033601375 Chr.7: 55997407-55997407 on Build GRCh38
rs920398401 Chr.7: 55997416-55997416 on Build GRCh38
rs777553508 Chr.7: 55997360-55997360 on Build GRCh38
rs768609229 Chr.7: 55997500-55997500 on Build GRCh38
rs756802675 Chr.7: 55997474-55997474 on Build GRCh38
rs776799938 Chr.7: 55997431-55997431 on Build GRCh38

View Full Product Details