Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94489901-94490487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1429
Entrez Gene ID
25962
Gene Name
KIAA1429
Gene Aliases
MSTP054, fSAP121
UniGene
Hs.202238
Gene Chromosome Location
Chr.8: 94487777-94553518 on Build GRCh38
Transcripts
NM_015496.4 XM_017013306.1 XM_017013307.1 XM_017013308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1719288 c.5279C>T p.S1760F Chr.8: 94489944-94489944 on Build GRCh38
1624300 c.5266C>G p.R1756G Chr.8: 94489957-94489957 on Build GRCh38
373340 c.5163A>T p.G1721G Chr.8: 94490060-94490060 on Build GRCh38
3651926 c.5261C>T p.P1754L Chr.8: 94489962-94489962 on Build GRCh38
4945345 c.5201G>A p.R1734Q Chr.8: 94490022-94490022 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006563148 Chr.8: 94490343-94490343 on Build GRCh38
rs748772617 Chr.8: 94490399-94490399 on Build GRCh38
rs780944867 Chr.8: 94490056-94490056 on Build GRCh38
rs191252208 Chr.8: 94490345-94490345 on Build GRCh38
rs377290884 Chr.8: 94489935-94489935 on Build GRCh38
rs764501753 Chr.8: 94489974-94489974 on Build GRCh38
rs35018347 Chr.8: 94489928-94489929 on Build GRCh38
rs370514091 Chr.8: 94489951-94489951 on Build GRCh38
rs766400176 Chr.8: 94490130-94490130 on Build GRCh38
rs111703158 Chr.8: 94490044-94490044 on Build GRCh38
rs371467408 Chr.8: 94490091-94490091 on Build GRCh38
rs777207486 Chr.8: 94490074-94490074 on Build GRCh38
rs754348556 Chr.8: 94489980-94489980 on Build GRCh38
rs551358096 Chr.8: 94489964-94489964 on Build GRCh38
rs768686928 Chr.8: 94490068-94490068 on Build GRCh38
rs748834374 Chr.8: 94490076-94490076 on Build GRCh38
rs1003577546 Chr.8: 94490144-94490144 on Build GRCh38
rs778653930 Chr.8: 94490449-94490449 on Build GRCh38
rs3762053 Chr.8: 94490326-94490326 on Build GRCh38
rs761151690 Chr.8: 94489970-94489970 on Build GRCh38
rs149975692 Chr.8: 94490371-94490371 on Build GRCh38
rs748072056 Chr.8: 94489940-94489940 on Build GRCh38
rs79699194 Chr.8: 94490109-94490109 on Build GRCh38
rs147465632 Chr.8: 94490029-94490029 on Build GRCh38
rs765295873 Chr.8: 94490002-94490002 on Build GRCh38
rs112695657 Chr.8: 94490059-94490059 on Build GRCh38
rs370224905 Chr.8: 94490070-94490070 on Build GRCh38
rs774080306 Chr.8: 94489966-94489966 on Build GRCh38
rs751743778 Chr.8: 94490010-94490010 on Build GRCh38
rs112998318 Chr.8: 94489979-94489979 on Build GRCh38
rs572643398 Chr.8: 94490402-94490402 on Build GRCh38
rs571445933 Chr.8: 94490001-94490001 on Build GRCh38
rs765783285 Chr.8: 94490125-94490125 on Build GRCh38
rs750499013 Chr.8: 94490004-94490004 on Build GRCh38
rs372647301 Chr.8: 94490104-94490104 on Build GRCh38
rs558957661 Chr.8: 94490390-94490390 on Build GRCh38
rs763055573 Chr.8: 94490129-94490129 on Build GRCh38
rs769239806 Chr.8: 94489952-94489952 on Build GRCh38
rs1051118669 Chr.8: 94490323-94490323 on Build GRCh38
rs756074657 Chr.8: 94490071-94490071 on Build GRCh38
rs368575681 Chr.8: 94490088-94490088 on Build GRCh38
rs924072904 Chr.8: 94490276-94490276 on Build GRCh38
rs1035387686 Chr.8: 94490098-94490098 on Build GRCh38
rs780262471 Chr.8: 94490009-94490009 on Build GRCh38
rs376333024 Chr.8: 94490053-94490053 on Build GRCh38
rs752642950 Chr.8: 94490185-94490185 on Build GRCh38
rs761695318 Chr.8: 94490103-94490104 on Build GRCh38
rs569739596 Chr.8: 94489950-94489950 on Build GRCh38
rs538872899 Chr.8: 94489991-94489991 on Build GRCh38
rs1009970038 Chr.8: 94490262-94490262 on Build GRCh38
rs773529239 Chr.8: 94490127-94490127 on Build GRCh38
rs770524764 Chr.8: 94490081-94490081 on Build GRCh38
rs911151570 Chr.8: 94490007-94490007 on Build GRCh38
rs898418670 Chr.8: 94490436-94490436 on Build GRCh38
rs756060491 Chr.8: 94490213-94490213 on Build GRCh38
rs918409734 Chr.8: 94490452-94490452 on Build GRCh38
rs565594749 Chr.8: 94490226-94490226 on Build GRCh38
rs916825294 Chr.8: 94490150-94490150 on Build GRCh38
rs200609043 Chr.8: 94490006-94490006 on Build GRCh38
rs942700275 Chr.8: 94490378-94490378 on Build GRCh38
rs951131657 Chr.8: 94490045-94490045 on Build GRCh38
rs781143009 Chr.8: 94489938-94489938 on Build GRCh38
rs749569924 Chr.8: 94490434-94490436 on Build GRCh38
rs776797393 Chr.8: 94490307-94490307 on Build GRCh38
rs768377783 Chr.8: 94489932-94489932 on Build GRCh38
rs762579896 Chr.8: 94489963-94489963 on Build GRCh38
rs1006797527 Chr.8: 94489942-94489942 on Build GRCh38
rs374711961 Chr.8: 94489956-94489956 on Build GRCh38
rs767493502 Chr.8: 94490112-94490112 on Build GRCh38

View Full Product Details