Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 4847553-4847987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1E2
Entrez Gene ID
83592
Gene Name
aldo-keto reductase family 1 member E2
Gene Aliases
AKR1CL2, AKRDC1, LoopADR, hTSP, htAKR
UniGene
Hs.657944
Gene Chromosome Location
Chr.10: 4824554-4908421 on Build GRCh38
Transcripts
NM_001040177.2 NM_001271021.1 NM_001271025.1 NR_073125.1 NR_073126.1 NR_073127.1 XM_011519715.2 XM_011519718.2 XM_011519722.2 XM_011519724.2 XM_017016744.1 XM_017016745.1

SNP Detail

SNP ID Chromosome Location
rs772757333 Chr.10: 4847917-4847917 on Build GRCh38
rs866209006 Chr.10: 4847934-4847934 on Build GRCh38
rs575047640 Chr.10: 4847850-4847850 on Build GRCh38
rs910375761 Chr.10: 4847865-4847865 on Build GRCh38
rs543276362 Chr.10: 4847915-4847915 on Build GRCh38
rs543943068 Chr.10: 4847815-4847815 on Build GRCh38
rs913604917 Chr.10: 4847821-4847821 on Build GRCh38
rs554043610 Chr.10: 4847672-4847672 on Build GRCh38
rs911396238 Chr.10: 4847860-4847860 on Build GRCh38
rs942830072 Chr.10: 4847621-4847621 on Build GRCh38
rs541420450 Chr.10: 4847705-4847706 on Build GRCh38
rs577440404 Chr.10: 4847661-4847661 on Build GRCh38
rs542159688 Chr.10: 4847882-4847882 on Build GRCh38
rs1041070850 Chr.10: 4847651-4847652 on Build GRCh38
rs188927800 Chr.10: 4847822-4847822 on Build GRCh38
rs571264872 Chr.10: 4847658-4847658 on Build GRCh38
rs553102627 Chr.10: 4847586-4847586 on Build GRCh38
rs981865333 Chr.10: 4847766-4847766 on Build GRCh38
rs17133779 Chr.10: 4847782-4847782 on Build GRCh38
rs942839995 Chr.10: 4847871-4847871 on Build GRCh38
rs370252001 Chr.10: 4847573-4847573 on Build GRCh38
rs4409732 Chr.10: 4847922-4847922 on Build GRCh38
rs145929706 Chr.10: 4847883-4847883 on Build GRCh38
rs984335648 Chr.10: 4847851-4847851 on Build GRCh38
rs186774588 Chr.10: 4847732-4847732 on Build GRCh38
rs747330130 Chr.10: 4847594-4847594 on Build GRCh38
rs571973964 Chr.10: 4847698-4847698 on Build GRCh38
rs1039821173 Chr.10: 4847591-4847591 on Build GRCh38
rs1034752695 Chr.10: 4847764-4847764 on Build GRCh38
rs754035070 Chr.10: 4847958-4847958 on Build GRCh38
rs761064928 Chr.10: 4847925-4847925 on Build GRCh38
rs1869214 Chr.10: 4847578-4847578 on Build GRCh38
rs752534 Chr.10: 4847953-4847953 on Build GRCh38
rs773584514 Chr.10: 4847580-4847580 on Build GRCh38
rs987790900 Chr.10: 4847819-4847819 on Build GRCh38
rs1023509634 Chr.10: 4847730-4847730 on Build GRCh38
rs969610313 Chr.10: 4847713-4847713 on Build GRCh38
rs998174854 Chr.10: 4847613-4847613 on Build GRCh38
rs536062950 Chr.10: 4847714-4847714 on Build GRCh38
rs969172047 Chr.10: 4847748-4847748 on Build GRCh38
rs530448077 Chr.10: 4847886-4847886 on Build GRCh38
rs139817460 Chr.10: 4847887-4847887 on Build GRCh38
rs774000320 Chr.10: 4847879-4847879 on Build GRCh38
rs1034738409 Chr.10: 4847768-4847768 on Build GRCh38
rs17133780 Chr.10: 4847924-4847924 on Build GRCh38
rs1002797031 Chr.10: 4847743-4847743 on Build GRCh38
rs942761744 Chr.10: 4847587-4847587 on Build GRCh38
rs893803142 Chr.10: 4847892-4847892 on Build GRCh38
rs905039785 Chr.10: 4847949-4847949 on Build GRCh38
rs577180540 Chr.10: 4847841-4847841 on Build GRCh38
rs1049068794 Chr.10: 4847598-4847598 on Build GRCh38
rs12569410 Chr.10: 4847614-4847614 on Build GRCh38
rs541035873 Chr.10: 4847855-4847855 on Build GRCh38

View Full Product Details