Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 113730351-113730924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP7
Entrez Gene ID
840
Gene Name
caspase 7
Gene Aliases
CASP-7, CMH-1, ICE-LAP3, LICE2, MCH3
UniGene
Hs.9216
Gene Chromosome Location
Chr.10: 113679162-113730909 on Build GRCh38
Transcripts
NM_001227.4 NM_001267056.1 NM_001267057.1 NM_001267058.1 NM_001320911.1 NM_033338.5 NM_033339.4 NM_033340.3 XM_006718017.3 XM_011540260.1 XM_017016763.1 XM_017016764.1

SNP Detail

SNP ID Chromosome Location
rs12247588 Chr.10: 113730431-113730431 on Build GRCh38
rs916604825 Chr.10: 113730848-113730848 on Build GRCh38
rs961907176 Chr.10: 113730758-113730758 on Build GRCh38
rs958277735 Chr.10: 113730675-113730675 on Build GRCh38
rs920534792 Chr.10: 113730825-113730825 on Build GRCh38
rs1028951014 Chr.10: 113730416-113730416 on Build GRCh38
rs1055115409 Chr.10: 113730530-113730530 on Build GRCh38
rs991789005 Chr.10: 113730841-113730841 on Build GRCh38
rs553514657 Chr.10: 113730487-113730487 on Build GRCh38
rs973230018 Chr.10: 113730814-113730814 on Build GRCh38
rs1007324231 Chr.10: 113730463-113730463 on Build GRCh38
rs1015982354 Chr.10: 113730481-113730481 on Build GRCh38
rs1014648207 Chr.10: 113730757-113730757 on Build GRCh38
rs558317934 Chr.10: 113730629-113730629 on Build GRCh38
rs542609807 Chr.10: 113730796-113730796 on Build GRCh38
rs954466761 Chr.10: 113730380-113730380 on Build GRCh38
rs554656613 Chr.10: 113730529-113730529 on Build GRCh38
rs901453575 Chr.10: 113730405-113730405 on Build GRCh38
rs748415913 Chr.10: 113730478-113730478 on Build GRCh38
rs1028644491 Chr.10: 113730697-113730697 on Build GRCh38
rs989540398 Chr.10: 113730667-113730667 on Build GRCh38
rs895252602 Chr.10: 113730534-113730534 on Build GRCh38
rs540950787 Chr.10: 113730665-113730665 on Build GRCh38
rs762437935 Chr.10: 113730501-113730501 on Build GRCh38
rs186832916 Chr.10: 113730664-113730664 on Build GRCh38
rs758775179 Chr.10: 113730881-113730881 on Build GRCh38
rs954497513 Chr.10: 113730700-113730700 on Build GRCh38
rs901173339 Chr.10: 113730721-113730721 on Build GRCh38
rs940481181 Chr.10: 113730409-113730409 on Build GRCh38
rs963059553 Chr.10: 113730514-113730514 on Build GRCh38
rs1032589183 Chr.10: 113730666-113730666 on Build GRCh38
rs765577814 Chr.10: 113730560-113730560 on Build GRCh38
rs907721175 Chr.10: 113730398-113730398 on Build GRCh38
rs971090547 Chr.10: 113730621-113730621 on Build GRCh38
rs971752841 Chr.10: 113730524-113730524 on Build GRCh38
rs998431583 Chr.10: 113730412-113730412 on Build GRCh38
rs559351537 Chr.10: 113730687-113730687 on Build GRCh38
rs934271958 Chr.10: 113730741-113730741 on Build GRCh38
rs565689460 Chr.10: 113730490-113730490 on Build GRCh38
rs150326299 Chr.10: 113730540-113730540 on Build GRCh38
rs541386567 Chr.10: 113730461-113730461 on Build GRCh38
rs915295305 Chr.10: 113730677-113730677 on Build GRCh38
rs937262574 Chr.10: 113730833-113730833 on Build GRCh38
rs569864593 Chr.10: 113730859-113730859 on Build GRCh38
rs556922705 Chr.10: 113730388-113730388 on Build GRCh38
rs535836568 Chr.10: 113730495-113730495 on Build GRCh38
rs1037468366 Chr.10: 113730895-113730895 on Build GRCh38
rs137932518 Chr.10: 113730706-113730706 on Build GRCh38
rs987739220 Chr.10: 113730395-113730395 on Build GRCh38
rs543256940 Chr.10: 113730593-113730593 on Build GRCh38
rs190021522 Chr.10: 113730850-113730850 on Build GRCh38

View Full Product Details