Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 43327443-43328031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
API5
Entrez Gene ID
8539
Gene Name
apoptosis inhibitor 5
Gene Aliases
AAC-11, AAC11
UniGene
Hs.435771
Gene Chromosome Location
Chr.11: 43311955-43344530 on Build GRCh38
Transcripts
NM_001142930.1 NM_001142931.1 NM_001243747.1 NM_006595.3 NR_024625.1 XM_006718359.3 XM_017018464.1 XM_017018465.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6602155 c.861G>T p.L287F Chr.11: 43327794-43327794 on Build GRCh38
6602154 c.861G>T p.L287F Chr.11: 43327794-43327794 on Build GRCh38
1171781 c.910G>C p.E304Q Chr.11: 43327843-43327843 on Build GRCh38
6602156 c.861G>T p.L287F Chr.11: 43327794-43327794 on Build GRCh38
1171780 c.910G>C p.E304Q Chr.11: 43327843-43327843 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017769843 Chr.11: 43327486-43327486 on Build GRCh38
rs763015663 Chr.11: 43327917-43327917 on Build GRCh38
rs991332381 Chr.11: 43327979-43327979 on Build GRCh38
rs1022756976 Chr.11: 43328009-43328009 on Build GRCh38
rs995246217 Chr.11: 43327514-43327514 on Build GRCh38
rs28468576 Chr.11: 43327664-43327664 on Build GRCh38
rs779636648 Chr.11: 43327863-43327863 on Build GRCh38
rs771538107 Chr.11: 43327755-43327756 on Build GRCh38
rs963829018 Chr.11: 43327487-43327487 on Build GRCh38
rs147759395 Chr.11: 43327835-43327835 on Build GRCh38
rs764167463 Chr.11: 43327806-43327806 on Build GRCh38
rs543630471 Chr.11: 43327805-43327805 on Build GRCh38
rs765757971 Chr.11: 43327719-43327719 on Build GRCh38
rs747788863 Chr.11: 43327889-43327889 on Build GRCh38
rs75380901 Chr.11: 43327855-43327855 on Build GRCh38
rs978635610 Chr.11: 43327991-43327991 on Build GRCh38
rs200389272 Chr.11: 43327903-43327903 on Build GRCh38
rs1026657552 Chr.11: 43327516-43327516 on Build GRCh38
rs1039250829 Chr.11: 43327773-43327773 on Build GRCh38
rs762960842 Chr.11: 43327761-43327761 on Build GRCh38
rs776211232 Chr.11: 43327887-43327887 on Build GRCh38
rs766180167 Chr.11: 43327927-43327927 on Build GRCh38
rs1035343537 Chr.11: 43327913-43327913 on Build GRCh38
rs5743239 Chr.11: 43327729-43327729 on Build GRCh38
rs746443788 Chr.11: 43327870-43327870 on Build GRCh38
rs1007083361 Chr.11: 43327711-43327711 on Build GRCh38
rs960268551 Chr.11: 43327963-43327963 on Build GRCh38
rs150499929 Chr.11: 43327571-43327571 on Build GRCh38
rs1035987271 Chr.11: 43327552-43327552 on Build GRCh38
rs779239797 Chr.11: 43327830-43327830 on Build GRCh38
rs867440615 Chr.11: 43327811-43327811 on Build GRCh38
rs1010055999 Chr.11: 43327764-43327769 on Build GRCh38
rs558053443 Chr.11: 43327714-43327714 on Build GRCh38
rs896011620 Chr.11: 43327496-43327496 on Build GRCh38
rs568186516 Chr.11: 43327523-43327523 on Build GRCh38
rs890727959 Chr.11: 43327733-43327733 on Build GRCh38
rs982735573 Chr.11: 43327543-43327543 on Build GRCh38
rs1020099395 Chr.11: 43327794-43327794 on Build GRCh38
rs756383574 Chr.11: 43327776-43327776 on Build GRCh38
rs574633866 Chr.11: 43327891-43327891 on Build GRCh38
rs924654265 Chr.11: 43327689-43327689 on Build GRCh38
rs12292698 Chr.11: 43327585-43327585 on Build GRCh38
rs889427328 Chr.11: 43327703-43327703 on Build GRCh38
rs773111426 Chr.11: 43327893-43327893 on Build GRCh38
rs535539474 Chr.11: 43327534-43327534 on Build GRCh38
rs951077265 Chr.11: 43327531-43327531 on Build GRCh38
rs754237490 Chr.11: 43327818-43327818 on Build GRCh38
rs539016306 Chr.11: 43327598-43327598 on Build GRCh38
rs755392369 Chr.11: 43327715-43327715 on Build GRCh38
rs767677435 Chr.11: 43327769-43327769 on Build GRCh38
rs995027994 Chr.11: 43327643-43327643 on Build GRCh38
rs5743240 Chr.11: 43327831-43327831 on Build GRCh38
rs768358465 Chr.11: 43327876-43327876 on Build GRCh38
rs978776853 Chr.11: 43327668-43327668 on Build GRCh38
rs995256015 Chr.11: 43327814-43327814 on Build GRCh38
rs144200699 Chr.11: 43327828-43327828 on Build GRCh38
rs915740947 Chr.11: 43327611-43327611 on Build GRCh38
rs947483245 Chr.11: 43327642-43327642 on Build GRCh38

View Full Product Details