Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 43323238-43323810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
API5
Entrez Gene ID
8539
Gene Name
apoptosis inhibitor 5
Gene Aliases
AAC-11, AAC11
UniGene
Hs.435771
Gene Chromosome Location
Chr.11: 43311955-43344530 on Build GRCh38
Transcripts
NM_001142930.1 NM_001142931.1 NM_001243747.1 NM_006595.3 NR_024625.1 XM_006718359.3 XM_017018464.1 XM_017018465.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
926876 c.568G>T p.E190* Chr.11: 43323454-43323454 on Build GRCh38
4191537 c.690G>A p.S230S Chr.11: 43323576-43323576 on Build GRCh38
7355950 c.733G>T p.A245S Chr.11: 43323619-43323619 on Build GRCh38
926877 c.681C>A p.F227L Chr.11: 43323567-43323567 on Build GRCh38
1146714 c.549A>T p.L183L Chr.11: 43323435-43323435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748486932 Chr.11: 43323552-43323552 on Build GRCh38
rs887723401 Chr.11: 43323391-43323391 on Build GRCh38
rs763525132 Chr.11: 43323535-43323535 on Build GRCh38
rs780256757 Chr.11: 43323505-43323505 on Build GRCh38
rs878969021 Chr.11: 43323740-43323740 on Build GRCh38
rs984497463 Chr.11: 43323764-43323764 on Build GRCh38
rs541112138 Chr.11: 43323473-43323473 on Build GRCh38
rs772611364 Chr.11: 43323380-43323380 on Build GRCh38
rs756494299 Chr.11: 43323364-43323365 on Build GRCh38
rs770326243 Chr.11: 43323260-43323260 on Build GRCh38
rs956973841 Chr.11: 43323600-43323600 on Build GRCh38
rs369837996 Chr.11: 43323569-43323569 on Build GRCh38
rs112521686 Chr.11: 43323540-43323540 on Build GRCh38
rs931644570 Chr.11: 43323389-43323389 on Build GRCh38
rs1019750406 Chr.11: 43323642-43323642 on Build GRCh38
rs183867817 Chr.11: 43323648-43323648 on Build GRCh38
rs748312088 Chr.11: 43323672-43323672 on Build GRCh38
rs766370031 Chr.11: 43323587-43323587 on Build GRCh38
rs770250344 Chr.11: 43323676-43323676 on Build GRCh38
rs765804099 Chr.11: 43323404-43323404 on Build GRCh38
rs758561011 Chr.11: 43323491-43323491 on Build GRCh38
rs777157059 Chr.11: 43323549-43323549 on Build GRCh38
rs752453501 Chr.11: 43323466-43323466 on Build GRCh38
rs777710503 Chr.11: 43323477-43323477 on Build GRCh38
rs768956427 Chr.11: 43323524-43323524 on Build GRCh38
rs1804963 Chr.11: 43323451-43323451 on Build GRCh38
rs754603433 Chr.11: 43323416-43323416 on Build GRCh38
rs780340800 Chr.11: 43323412-43323412 on Build GRCh38
rs988296181 Chr.11: 43323283-43323283 on Build GRCh38
rs1014093512 Chr.11: 43323528-43323528 on Build GRCh38
rs896311618 Chr.11: 43323478-43323478 on Build GRCh38
rs760454031 Chr.11: 43323654-43323654 on Build GRCh38
rs780307879 Chr.11: 43323661-43323661 on Build GRCh38
rs905209925 Chr.11: 43323550-43323550 on Build GRCh38
rs199998268 Chr.11: 43323392-43323392 on Build GRCh38
rs912710150 Chr.11: 43323300-43323300 on Build GRCh38
rs755872041 Chr.11: 43323399-43323400 on Build GRCh38
rs372483451 Chr.11: 43323649-43323649 on Build GRCh38
rs749676661 Chr.11: 43323763-43323763 on Build GRCh38
rs571240580 Chr.11: 43323657-43323657 on Build GRCh38
rs548816637 Chr.11: 43323315-43323315 on Build GRCh38
rs908877253 Chr.11: 43323765-43323765 on Build GRCh38
rs377763153 Chr.11: 43323483-43323483 on Build GRCh38
rs375743976 Chr.11: 43323669-43323669 on Build GRCh38
rs940623476 Chr.11: 43323783-43323783 on Build GRCh38
rs536404946 Chr.11: 43323734-43323734 on Build GRCh38
rs201696733 Chr.11: 43323621-43323621 on Build GRCh38
rs753860341 Chr.11: 43323659-43323659 on Build GRCh38
rs568948385 Chr.11: 43323731-43323731 on Build GRCh38
rs751273178 Chr.11: 43323415-43323415 on Build GRCh38
rs560599812 Chr.11: 43323441-43323441 on Build GRCh38
rs777980032 Chr.11: 43323686-43323686 on Build GRCh38
rs34890314 Chr.11: 43323710-43323711 on Build GRCh38
rs373110615 Chr.11: 43323400-43323400 on Build GRCh38
rs773834543 Chr.11: 43323565-43323565 on Build GRCh38
rs770204897 Chr.11: 43323561-43323561 on Build GRCh38
rs545188503 Chr.11: 43323711-43323711 on Build GRCh38
rs759218862 Chr.11: 43323623-43323623 on Build GRCh38
rs921600090 Chr.11: 43323371-43323371 on Build GRCh38
rs779629582 Chr.11: 43323406-43323407 on Build GRCh38
rs1040090104 Chr.11: 43323364-43323364 on Build GRCh38
rs773574203 Chr.11: 43323390-43323390 on Build GRCh38
rs541022654 Chr.11: 43323662-43323662 on Build GRCh38
rs902545698 Chr.11: 43323749-43323749 on Build GRCh38
rs1004786493 Chr.11: 43323399-43323399 on Build GRCh38
rs530673988 Chr.11: 43323271-43323271 on Build GRCh38
rs944495519 Chr.11: 43323317-43323317 on Build GRCh38
rs933255209 Chr.11: 43323689-43323689 on Build GRCh38
rs368827333 Chr.11: 43323772-43323772 on Build GRCh38
rs538534179 Chr.11: 43323690-43323690 on Build GRCh38
rs550796810 Chr.11: 43323723-43323723 on Build GRCh38
rs747270288 Chr.11: 43323515-43323515 on Build GRCh38

View Full Product Details