Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86345163-86345734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIKESHI
Entrez Gene ID
51501
Gene Name
Hikeshi, heat shock protein nuclear import factor
Gene Aliases
C11orf73, HLD13, HSPC138, HSPC179, L7RN6, OPI10
UniGene
Hs.283322
Gene Chromosome Location
Chr.11: 86302211-86345943 on Build GRCh38
Transcripts
NM_001322404.1 NM_001322407.1 NM_001322409.1 NM_016401.3 NR_024597.1 NR_024598.1 NR_136324.1 XM_017017914.1 XM_017017915.1

SNP Detail

SNP ID Chromosome Location
rs775153640 Chr.11: 86345560-86345560 on Build GRCh38
rs774141787 Chr.11: 86345497-86345497 on Build GRCh38
rs528002201 Chr.11: 86345380-86345380 on Build GRCh38
rs375875670 Chr.11: 86345680-86345680 on Build GRCh38
rs148932996 Chr.11: 86345596-86345596 on Build GRCh38
rs761079545 Chr.11: 86345598-86345598 on Build GRCh38
rs1028965362 Chr.11: 86345459-86345459 on Build GRCh38
rs922199376 Chr.11: 86345222-86345222 on Build GRCh38
rs968737173 Chr.11: 86345188-86345188 on Build GRCh38
rs530994243 Chr.11: 86345413-86345413 on Build GRCh38
rs572016024 Chr.11: 86345219-86345219 on Build GRCh38
rs746973408 Chr.11: 86345652-86345652 on Build GRCh38
rs965190482 Chr.11: 86345503-86345503 on Build GRCh38
rs186219113 Chr.11: 86345291-86345291 on Build GRCh38
rs35058357 Chr.11: 86345660-86345661 on Build GRCh38
rs550790064 Chr.11: 86345484-86345484 on Build GRCh38
rs74694718 Chr.11: 86345408-86345408 on Build GRCh38
rs997944378 Chr.11: 86345522-86345522 on Build GRCh38
rs909240852 Chr.11: 86345563-86345563 on Build GRCh38
rs1043310883 Chr.11: 86345710-86345710 on Build GRCh38
rs969186219 Chr.11: 86345421-86345421 on Build GRCh38
rs919614404 Chr.11: 86345292-86345292 on Build GRCh38
rs976639188 Chr.11: 86345274-86345274 on Build GRCh38
rs766714056 Chr.11: 86345604-86345604 on Build GRCh38
rs1036774499 Chr.11: 86345290-86345290 on Build GRCh38
rs755199077 Chr.11: 86345617-86345617 on Build GRCh38
rs563575596 Chr.11: 86345340-86345340 on Build GRCh38
rs772391972 Chr.11: 86345529-86345529 on Build GRCh38
rs577032059 Chr.11: 86345351-86345351 on Build GRCh38
rs987646847 Chr.11: 86345267-86345267 on Build GRCh38
rs1010767750 Chr.11: 86345410-86345410 on Build GRCh38
rs753865329 Chr.11: 86345576-86345576 on Build GRCh38
rs80189718 Chr.11: 86345288-86345288 on Build GRCh38
rs963410015 Chr.11: 86345616-86345616 on Build GRCh38
rs538606812 Chr.11: 86345185-86345185 on Build GRCh38
rs954913715 Chr.11: 86345246-86345246 on Build GRCh38
rs1044479777 Chr.11: 86345358-86345358 on Build GRCh38
rs773772891 Chr.11: 86345550-86345550 on Build GRCh38
rs367653131 Chr.11: 86345555-86345566 on Build GRCh38
rs528472088 Chr.11: 86345365-86345365 on Build GRCh38
rs752801062 Chr.11: 86345624-86345624 on Build GRCh38
rs377658287 Chr.11: 86345464-86345465 on Build GRCh38
rs554749942 Chr.11: 86345280-86345280 on Build GRCh38
rs987720444 Chr.11: 86345505-86345505 on Build GRCh38
rs111497820 Chr.11: 86345281-86345281 on Build GRCh38
rs1021696643 Chr.11: 86345412-86345412 on Build GRCh38
rs980597672 Chr.11: 86345623-86345623 on Build GRCh38
rs190680463 Chr.11: 86345357-86345357 on Build GRCh38
rs147733341 Chr.11: 86345215-86345215 on Build GRCh38
rs754086423 Chr.11: 86345607-86345607 on Build GRCh38
rs767077751 Chr.11: 86345535-86345535 on Build GRCh38
rs769359144 Chr.11: 86345555-86345555 on Build GRCh38
rs768168001 Chr.11: 86345576-86345576 on Build GRCh38
rs904180168 Chr.11: 86345480-86345480 on Build GRCh38
rs10792848 Chr.11: 86345364-86345364 on Build GRCh38
rs527317291 Chr.11: 86345700-86345700 on Build GRCh38
rs764156215 Chr.11: 86345571-86345575 on Build GRCh38
rs1017938916 Chr.11: 86345486-86345486 on Build GRCh38
rs759174706 Chr.11: 86345533-86345533 on Build GRCh38
rs182803417 Chr.11: 86345396-86345396 on Build GRCh38
rs780849819 Chr.11: 86345662-86345662 on Build GRCh38
rs113282403 Chr.11: 86345532-86345532 on Build GRCh38
rs201671408 Chr.11: 86345635-86345635 on Build GRCh38
rs996205802 Chr.11: 86345457-86345457 on Build GRCh38
rs913389584 Chr.11: 86345693-86345693 on Build GRCh38
rs950730444 Chr.11: 86345312-86345312 on Build GRCh38
rs892024767 Chr.11: 86345397-86345397 on Build GRCh38
rs983832169 Chr.11: 86345332-86345332 on Build GRCh38
rs547273792 Chr.11: 86345644-86345644 on Build GRCh38
rs534659008 Chr.11: 86345224-86345224 on Build GRCh38
rs965180659 Chr.11: 86345255-86345255 on Build GRCh38
rs762361741 Chr.11: 86345568-86345568 on Build GRCh38
rs182936327 Chr.11: 86345665-86345665 on Build GRCh38
rs757067977 Chr.11: 86345653-86345653 on Build GRCh38
rs187152649 Chr.11: 86345510-86345510 on Build GRCh38
rs777738757 Chr.11: 86345645-86345645 on Build GRCh38
rs754398394 Chr.11: 86345621-86345621 on Build GRCh38
rs745463188 Chr.11: 86345664-86345664 on Build GRCh38
rs775100177 Chr.11: 86345230-86345230 on Build GRCh38

View Full Product Details