Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 26398588-26399038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPR2
Entrez Gene ID
3709
Gene Name
inositol 1,4,5-trisphosphate receptor type 2
Gene Aliases
ANHD, CFAP48, INSP3R2, IP3R2
UniGene
Hs.512235
Gene Chromosome Location
Chr.12: 26335337-26833198 on Build GRCh38
Transcripts
NM_002223.3 XM_017019266.1 XM_017019267.1 XM_017019268.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
108558 c.7656G>A p.Q2552Q Chr.12: 26398916-26398916 on Build GRCh38
7275699 c.7540T>C p.F2514L Chr.12: 26399032-26399032 on Build GRCh38
1135367 c.7696+2T>C p.? Chr.12: 26398874-26398874 on Build GRCh38
3811720 c.7647C>T p.S2549S Chr.12: 26398925-26398925 on Build GRCh38
1605977 c.7607T>C p.I2536T Chr.12: 26398965-26398965 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs193145513 Chr.12: 26398949-26398949 on Build GRCh38
rs199972885 Chr.12: 26398844-26398844 on Build GRCh38
rs1011343894 Chr.12: 26398757-26398757 on Build GRCh38
rs917534428 Chr.12: 26398655-26398655 on Build GRCh38
rs900427975 Chr.12: 26398761-26398761 on Build GRCh38
rs773390113 Chr.12: 26398861-26398861 on Build GRCh38
rs748602264 Chr.12: 26398854-26398854 on Build GRCh38
rs780891302 Chr.12: 26398764-26398764 on Build GRCh38
rs555893173 Chr.12: 26398830-26398833 on Build GRCh38
rs767482355 Chr.12: 26398970-26398970 on Build GRCh38
rs916533346 Chr.12: 26398876-26398876 on Build GRCh38
rs556256417 Chr.12: 26398821-26398821 on Build GRCh38
rs745514089 Chr.12: 26398787-26398787 on Build GRCh38
rs983236999 Chr.12: 26398819-26398819 on Build GRCh38
rs146419029 Chr.12: 26398916-26398916 on Build GRCh38
rs949627241 Chr.12: 26398678-26398678 on Build GRCh38
rs892864119 Chr.12: 26398744-26398744 on Build GRCh38
rs1004209834 Chr.12: 26398823-26398823 on Build GRCh38
rs375931279 Chr.12: 26398877-26398877 on Build GRCh38
rs370237159 Chr.12: 26398973-26398973 on Build GRCh38
rs950661716 Chr.12: 26398818-26398818 on Build GRCh38
rs113803799 Chr.12: 26398666-26398666 on Build GRCh38
rs747728928 Chr.12: 26398998-26398999 on Build GRCh38
rs990921850 Chr.12: 26398846-26398846 on Build GRCh38
rs755644595 Chr.12: 26398993-26398993 on Build GRCh38
rs999691140 Chr.12: 26398726-26398726 on Build GRCh38
rs751427230 Chr.12: 26398925-26398925 on Build GRCh38
rs1030204746 Chr.12: 26398806-26398806 on Build GRCh38
rs763790368 Chr.12: 26398898-26398898 on Build GRCh38
rs779700879 Chr.12: 26399017-26399017 on Build GRCh38
rs749617306 Chr.12: 26398825-26398825 on Build GRCh38
rs977872183 Chr.12: 26398687-26398687 on Build GRCh38
rs780932029 Chr.12: 26398836-26398836 on Build GRCh38
rs936515076 Chr.12: 26398735-26398735 on Build GRCh38
rs769655558 Chr.12: 26398617-26398617 on Build GRCh38
rs760865222 Chr.12: 26398865-26398865 on Build GRCh38
rs17470625 Chr.12: 26398716-26398716 on Build GRCh38
rs566800462 Chr.12: 26398766-26398766 on Build GRCh38
rs1046688677 Chr.12: 26398711-26398711 on Build GRCh38
rs771225790 Chr.12: 26398866-26398866 on Build GRCh38
rs1053980944 Chr.12: 26398736-26398736 on Build GRCh38
rs776728857 Chr.12: 26398869-26398869 on Build GRCh38
rs529679060 Chr.12: 26398788-26398788 on Build GRCh38
rs555475314 Chr.12: 26398809-26398809 on Build GRCh38
rs372773826 Chr.12: 26398841-26398841 on Build GRCh38
rs562659596 Chr.12: 26398717-26398717 on Build GRCh38
rs772767712 Chr.12: 26398859-26398859 on Build GRCh38
rs935597820 Chr.12: 26398985-26398985 on Build GRCh38

View Full Product Details