Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51073855-51074454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRNP2
Entrez Gene ID
81566
Gene Name
cysteine and serine rich nuclear protein 2
Gene Aliases
C12orf2, C12orf22, FAM130A1, PPP1R72, TAIP-12
UniGene
Hs.524425
Gene Chromosome Location
Chr.12: 51061205-51083671 on Build GRCh38
Transcripts
NM_030809.2 NR_045072.1 XM_006719621.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6301517 c.282C>T p.G94G Chr.12: 51073952-51073952 on Build GRCh38
6838279 c.342G>C p.Q114H Chr.12: 51073892-51073892 on Build GRCh38
5051676 c.233G>T p.R78L Chr.12: 51074001-51074001 on Build GRCh38
5361895 c.236G>A p.R79H Chr.12: 51073998-51073998 on Build GRCh38
1299506 c.165G>A p.L55L Chr.12: 51074069-51074069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554177940 Chr.12: 51074313-51074313 on Build GRCh38
rs753246204 Chr.12: 51074121-51074121 on Build GRCh38
rs963212479 Chr.12: 51074263-51074263 on Build GRCh38
rs757731878 Chr.12: 51074134-51074134 on Build GRCh38
rs567119347 Chr.12: 51074149-51074149 on Build GRCh38
rs201980268 Chr.12: 51073984-51073984 on Build GRCh38
rs746608392 Chr.12: 51073992-51073992 on Build GRCh38
rs748655459 Chr.12: 51073942-51073942 on Build GRCh38
rs751622283 Chr.12: 51074359-51074359 on Build GRCh38
rs575877495 Chr.12: 51074203-51074203 on Build GRCh38
rs763280332 Chr.12: 51074123-51074123 on Build GRCh38
rs182311877 Chr.12: 51074279-51074279 on Build GRCh38
rs776237948 Chr.12: 51074005-51074005 on Build GRCh38
rs992543674 Chr.12: 51074315-51074315 on Build GRCh38
rs547306870 Chr.12: 51074088-51074088 on Build GRCh38
rs759593080 Chr.12: 51074096-51074096 on Build GRCh38
rs537917638 Chr.12: 51074305-51074305 on Build GRCh38
rs925603053 Chr.12: 51074120-51074120 on Build GRCh38
rs953029293 Chr.12: 51073877-51073877 on Build GRCh38
rs936853117 Chr.12: 51074364-51074364 on Build GRCh38
rs766390202 Chr.12: 51073922-51073922 on Build GRCh38
rs374563731 Chr.12: 51074099-51074099 on Build GRCh38
rs760629899 Chr.12: 51074366-51074366 on Build GRCh38
rs901279716 Chr.12: 51074180-51074180 on Build GRCh38
rs763852155 Chr.12: 51073910-51073910 on Build GRCh38
rs992980624 Chr.12: 51074379-51074379 on Build GRCh38
rs1008746888 Chr.12: 51074165-51074165 on Build GRCh38
rs950672027 Chr.12: 51074314-51074314 on Build GRCh38
rs777028747 Chr.12: 51073878-51073878 on Build GRCh38
rs994655419 Chr.12: 51074245-51074245 on Build GRCh38
rs954814735 Chr.12: 51074144-51074144 on Build GRCh38
rs189678870 Chr.12: 51074200-51074200 on Build GRCh38
rs370494067 Chr.12: 51073959-51073959 on Build GRCh38
rs372257216 Chr.12: 51074210-51074210 on Build GRCh38
rs745336139 Chr.12: 51074015-51074015 on Build GRCh38
rs1040216914 Chr.12: 51074178-51074178 on Build GRCh38
rs769389914 Chr.12: 51074121-51074122 on Build GRCh38
rs751898592 Chr.12: 51074133-51074133 on Build GRCh38
rs979823377 Chr.12: 51074337-51074337 on Build GRCh38
rs143649719 Chr.12: 51074064-51074064 on Build GRCh38
rs538865093 Chr.12: 51074199-51074199 on Build GRCh38
rs766619608 Chr.12: 51074079-51074079 on Build GRCh38
rs958934219 Chr.12: 51074369-51074369 on Build GRCh38
rs750651991 Chr.12: 51073915-51073915 on Build GRCh38
rs770726961 Chr.12: 51073998-51073998 on Build GRCh38
rs796899055 Chr.12: 51074360-51074360 on Build GRCh38
rs372606177 Chr.12: 51074050-51074050 on Build GRCh38
rs778148557 Chr.12: 51074132-51074132 on Build GRCh38
rs762864126 Chr.12: 51074049-51074049 on Build GRCh38
rs753937787 Chr.12: 51073929-51073929 on Build GRCh38
rs574084148 Chr.12: 51074408-51074408 on Build GRCh38
rs1025694494 Chr.12: 51074251-51074251 on Build GRCh38
rs374095986 Chr.12: 51074001-51074001 on Build GRCh38
rs368286037 Chr.12: 51074046-51074046 on Build GRCh38
rs948356479 Chr.12: 51074328-51074328 on Build GRCh38
rs747823551 Chr.12: 51073955-51073956 on Build GRCh38
rs1000949262 Chr.12: 51073950-51073950 on Build GRCh38
rs757923867 Chr.12: 51074385-51074385 on Build GRCh38
rs1030729264 Chr.12: 51074115-51074115 on Build GRCh38
rs750389420 Chr.12: 51074320-51074320 on Build GRCh38
rs140071191 Chr.12: 51074039-51074039 on Build GRCh38
rs772820425 Chr.12: 51074122-51074125 on Build GRCh38
rs758836245 Chr.12: 51074131-51074131 on Build GRCh38
rs760839840 Chr.12: 51074072-51074072 on Build GRCh38
rs368577384 Chr.12: 51073941-51073941 on Build GRCh38
rs1054350882 Chr.12: 51074380-51074380 on Build GRCh38
rs973622195 Chr.12: 51074275-51074275 on Build GRCh38

View Full Product Details