Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 10050100-10050630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKEF1
Entrez Gene ID
63926
Gene Name
ankyrin repeat and EF-hand domain containing 1
Gene Aliases
ANKRD5
UniGene
Hs.70903
Gene Chromosome Location
Chr.20: 10034999-10056764 on Build GRCh38
Transcripts
NM_001303472.1 NM_022096.5 NM_198798.2 XM_005260792.2 NR_040710.1

Gene Symbol
SNAP25-AS1
Entrez Gene ID
100131208
Gene Name
SNAP25 antisense RNA 1
Gene Aliases
-
UniGene
Hs.658178
Gene Chromosome Location
Chr.20: 10023812-10219506 on Build GRCh38
Transcripts
NM_001303472.1 NM_022096.5 NM_198798.2 XM_005260792.2 NR_040710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
370058 c.1643+1G>T p.? Chr.20: 10050213-10050213 on Build GRCh38
2702525 c.1563G>A p.K521K Chr.20: 10050132-10050132 on Build GRCh38
6840002 c.1601C>G p.A534G Chr.20: 10050170-10050170 on Build GRCh38
5495485 c.1615G>C p.D539H Chr.20: 10050184-10050184 on Build GRCh38
5526684 c.1544G>A p.G515E Chr.20: 10050113-10050113 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746768755 Chr.20: 10050558-10050558 on Build GRCh38
rs966403560 Chr.20: 10050258-10050258 on Build GRCh38
rs771857928 Chr.20: 10050259-10050259 on Build GRCh38
rs954548609 Chr.20: 10050587-10050587 on Build GRCh38
rs142915673 Chr.20: 10050128-10050128 on Build GRCh38
rs764711759 Chr.20: 10050175-10050175 on Build GRCh38
rs771117764 Chr.20: 10050181-10050182 on Build GRCh38
rs999231177 Chr.20: 10050493-10050493 on Build GRCh38
rs398121167 Chr.20: 10050607-10050608 on Build GRCh38
rs895322393 Chr.20: 10050468-10050468 on Build GRCh38
rs545444897 Chr.20: 10050228-10050228 on Build GRCh38
rs372553098 Chr.20: 10050219-10050219 on Build GRCh38
rs779463782 Chr.20: 10050225-10050225 on Build GRCh38
rs549184082 Chr.20: 10050310-10050310 on Build GRCh38
rs757764280 Chr.20: 10050206-10050206 on Build GRCh38
rs759544054 Chr.20: 10050153-10050153 on Build GRCh38
rs929919640 Chr.20: 10050494-10050494 on Build GRCh38
rs1013301047 Chr.20: 10050483-10050483 on Build GRCh38
rs771319596 Chr.20: 10050179-10050179 on Build GRCh38
rs201546867 Chr.20: 10050604-10050605 on Build GRCh38
rs556020583 Chr.20: 10050389-10050390 on Build GRCh38
rs555159995 Chr.20: 10050590-10050590 on Build GRCh38
rs556849872 Chr.20: 10050377-10050377 on Build GRCh38
rs747929655 Chr.20: 10050237-10050237 on Build GRCh38
rs778962491 Chr.20: 10050236-10050236 on Build GRCh38
rs768454811 Chr.20: 10050134-10050134 on Build GRCh38
rs984032338 Chr.20: 10050496-10050496 on Build GRCh38
rs958099693 Chr.20: 10050307-10050307 on Build GRCh38
rs958021921 Chr.20: 10050545-10050545 on Build GRCh38
rs995717856 Chr.20: 10050188-10050188 on Build GRCh38
rs202176618 Chr.20: 10050229-10050229 on Build GRCh38
rs1037288385 Chr.20: 10050132-10050132 on Build GRCh38
rs139125477 Chr.20: 10050606-10050607 on Build GRCh38
rs549424834 Chr.20: 10050484-10050484 on Build GRCh38
rs575804390 Chr.20: 10050599-10050599 on Build GRCh38
rs985638247 Chr.20: 10050432-10050432 on Build GRCh38
rs557874749 Chr.20: 10050598-10050599 on Build GRCh38
rs373175030 Chr.20: 10050367-10050371 on Build GRCh38
rs568289410 Chr.20: 10050325-10050325 on Build GRCh38
rs145395317 Chr.20: 10050152-10050152 on Build GRCh38
rs898771683 Chr.20: 10050428-10050428 on Build GRCh38
rs1020504283 Chr.20: 10050368-10050368 on Build GRCh38
rs937337814 Chr.20: 10050604-10050604 on Build GRCh38
rs1024611532 Chr.20: 10050257-10050257 on Build GRCh38
rs371916981 Chr.20: 10050165-10050165 on Build GRCh38
rs895995077 Chr.20: 10050330-10050332 on Build GRCh38
rs577562486 Chr.20: 10050600-10050601 on Build GRCh38
rs767216875 Chr.20: 10050216-10050216 on Build GRCh38
rs201872448 Chr.20: 10050187-10050187 on Build GRCh38
rs369775375 Chr.20: 10050527-10050527 on Build GRCh38
rs1048389260 Chr.20: 10050278-10050278 on Build GRCh38
rs974541846 Chr.20: 10050554-10050554 on Build GRCh38
rs986824062 Chr.20: 10050589-10050589 on Build GRCh38
rs185527241 Chr.20: 10050431-10050431 on Build GRCh38
rs541937288 Chr.20: 10050565-10050565 on Build GRCh38
rs750117443 Chr.20: 10050217-10050217 on Build GRCh38
rs770691699 Chr.20: 10050585-10050585 on Build GRCh38
rs190134265 Chr.20: 10050363-10050363 on Build GRCh38
rs761641099 Chr.20: 10050141-10050141 on Build GRCh38
rs909905205 Chr.20: 10050561-10050564 on Build GRCh38
rs111717543 Chr.20: 10050323-10050323 on Build GRCh38
rs753197232 Chr.20: 10050161-10050161 on Build GRCh38
rs6141102 Chr.20: 10050490-10050490 on Build GRCh38
rs563813876 Chr.20: 10050269-10050269 on Build GRCh38
rs1055334402 Chr.20: 10050459-10050459 on Build GRCh38
rs141133661 Chr.20: 10050364-10050368 on Build GRCh38
rs1037676335 Chr.20: 10050415-10050415 on Build GRCh38
rs776755496 Chr.20: 10050197-10050199 on Build GRCh38
rs546987220 Chr.20: 10050241-10050241 on Build GRCh38
rs765244490 Chr.20: 10050156-10050156 on Build GRCh38
rs267606072 Chr.20: 10050163-10050163 on Build GRCh38
rs544281992 Chr.20: 10050603-10050603 on Build GRCh38
rs761565888 Chr.20: 10050363-10050364 on Build GRCh38
rs746131889 Chr.20: 10050211-10050211 on Build GRCh38
rs564729872 Chr.20: 10050135-10050135 on Build GRCh38
rs182141058 Chr.20: 10050402-10050402 on Build GRCh38
rs656111 Chr.20: 10050283-10050283 on Build GRCh38
rs999149451 Chr.20: 10050369-10050370 on Build GRCh38
rs1032656464 Chr.20: 10050540-10050540 on Build GRCh38
rs932773260 Chr.20: 10050359-10050359 on Build GRCh38
rs751857877 Chr.20: 10050182-10050182 on Build GRCh38
rs937457691 Chr.20: 10050299-10050299 on Build GRCh38

View Full Product Details