Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 53729257-53729811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB3
Entrez Gene ID
51130
Gene Name
ankyrin repeat and SOCS box containing 3
Gene Aliases
ASB-3
UniGene
Hs.40763
Gene Chromosome Location
Chr.2: 53669980-53787009 on Build GRCh38
Transcripts
NM_001201965.1 NM_016115.4 NM_145863.2 NM_001164165.1

Gene Symbol
GPR75-ASB3
Entrez Gene ID
100302652
Gene Name
GPR75-ASB3 readthrough
Gene Aliases
-
UniGene
Hs.40763
Gene Chromosome Location
Chr.2: 53670293-53860033 on Build GRCh38
Transcripts
NM_001201965.1 NM_016115.4 NM_145863.2 NM_001164165.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7196614 c.582+8C>A p.? Chr.2: 53729450-53729450 on Build GRCh38
6880285 c.358G>T p.V120F Chr.2: 53729568-53729568 on Build GRCh38
82011 c.388C>T p.L130L Chr.2: 53729538-53729538 on Build GRCh38
4797739 c.468+1G>A p.? Chr.2: 53729457-53729457 on Build GRCh38
1021567 c.456G>T p.Q152H Chr.2: 53729470-53729470 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1012381052 Chr.2: 53729628-53729628 on Build GRCh38
rs1013736674 Chr.2: 53729377-53729377 on Build GRCh38
rs764341205 Chr.2: 53729416-53729416 on Build GRCh38
rs748789196 Chr.2: 53729384-53729384 on Build GRCh38
rs985927186 Chr.2: 53729600-53729600 on Build GRCh38
rs796298265 Chr.2: 53729532-53729532 on Build GRCh38
rs754479168 Chr.2: 53729428-53729428 on Build GRCh38
rs758200256 Chr.2: 53729557-53729557 on Build GRCh38
rs764137718 Chr.2: 53729566-53729566 on Build GRCh38
rs769480407 Chr.2: 53729351-53729351 on Build GRCh38
rs1002647818 Chr.2: 53729605-53729605 on Build GRCh38
rs2302797 Chr.2: 53729746-53729746 on Build GRCh38
rs930091971 Chr.2: 53729495-53729495 on Build GRCh38
rs565548762 Chr.2: 53729518-53729518 on Build GRCh38
rs776120466 Chr.2: 53729546-53729546 on Build GRCh38
rs771411958 Chr.2: 53729678-53729678 on Build GRCh38
rs991082894 Chr.2: 53729335-53729335 on Build GRCh38
rs537288593 Chr.2: 53729658-53729658 on Build GRCh38
rs562195641 Chr.2: 53729753-53729753 on Build GRCh38
rs1002760383 Chr.2: 53729682-53729682 on Build GRCh38
rs912615174 Chr.2: 53729779-53729779 on Build GRCh38
rs1053971995 Chr.2: 53729685-53729685 on Build GRCh38
rs771217585 Chr.2: 53729492-53729492 on Build GRCh38
rs1041808209 Chr.2: 53729588-53729588 on Build GRCh38
rs770967538 Chr.2: 53729573-53729574 on Build GRCh38
rs781172687 Chr.2: 53729577-53729577 on Build GRCh38
rs769102259 Chr.2: 53729586-53729586 on Build GRCh38
rs769216648 Chr.2: 53729521-53729521 on Build GRCh38
rs774641511 Chr.2: 53729523-53729523 on Build GRCh38
rs918921792 Chr.2: 53729635-53729635 on Build GRCh38
rs746055408 Chr.2: 53729579-53729579 on Build GRCh38
rs573180732 Chr.2: 53729731-53729731 on Build GRCh38
rs574668719 Chr.2: 53729657-53729657 on Build GRCh38
rs750237566 Chr.2: 53729507-53729507 on Build GRCh38
rs759285484 Chr.2: 53729547-53729547 on Build GRCh38
rs117470288 Chr.2: 53729710-53729710 on Build GRCh38
rs200919329 Chr.2: 53729582-53729582 on Build GRCh38
rs762570538 Chr.2: 53729614-53729614 on Build GRCh38
rs375527218 Chr.2: 53729427-53729427 on Build GRCh38
rs942541416 Chr.2: 53729784-53729784 on Build GRCh38
rs779504828 Chr.2: 53729380-53729380 on Build GRCh38
rs941658987 Chr.2: 53729340-53729340 on Build GRCh38
rs1050763861 Chr.2: 53729287-53729287 on Build GRCh38
rs570004562 Chr.2: 53729283-53729283 on Build GRCh38
rs759800954 Chr.2: 53729705-53729705 on Build GRCh38
rs529904293 Chr.2: 53729776-53729776 on Build GRCh38
rs960846645 Chr.2: 53729304-53729304 on Build GRCh38
rs768070876 Chr.2: 53729474-53729474 on Build GRCh38
rs538249714 Chr.2: 53729330-53729330 on Build GRCh38
rs909141829 Chr.2: 53729290-53729290 on Build GRCh38
rs1039602794 Chr.2: 53729528-53729528 on Build GRCh38
rs760580086 Chr.2: 53729508-53729508 on Build GRCh38
rs772407091 Chr.2: 53729598-53729598 on Build GRCh38
rs149162759 Chr.2: 53729551-53729551 on Build GRCh38
rs751346352 Chr.2: 53729571-53729571 on Build GRCh38
rs559667658 Chr.2: 53729721-53729721 on Build GRCh38
rs953300053 Chr.2: 53729599-53729599 on Build GRCh38
rs774872223 Chr.2: 53729594-53729594 on Build GRCh38
rs373062855 Chr.2: 53729620-53729620 on Build GRCh38
rs767257466 Chr.2: 53729355-53729355 on Build GRCh38
rs749400075 Chr.2: 53729584-53729584 on Build GRCh38
rs1002386658 Chr.2: 53729405-53729405 on Build GRCh38
rs758422060 Chr.2: 53729473-53729473 on Build GRCh38
rs748695690 Chr.2: 53729596-53729596 on Build GRCh38
rs10490467 Chr.2: 53729701-53729701 on Build GRCh38
rs113219714 Chr.2: 53729531-53729531 on Build GRCh38
rs781583301 Chr.2: 53729493-53729493 on Build GRCh38
rs897549307 Chr.2: 53729367-53729367 on Build GRCh38
rs905353739 Chr.2: 53729391-53729391 on Build GRCh38
rs941963940 Chr.2: 53729619-53729619 on Build GRCh38
rs764080512 Chr.2: 53729616-53729616 on Build GRCh38
rs201274554 Chr.2: 53729497-53729497 on Build GRCh38
rs777709018 Chr.2: 53729475-53729475 on Build GRCh38
rs751976992 Chr.2: 53729417-53729417 on Build GRCh38
rs981179375 Chr.2: 53729301-53729301 on Build GRCh38
rs759543410 Chr.2: 53729408-53729408 on Build GRCh38
rs752559053 Chr.2: 53729455-53729455 on Build GRCh38
rs552102685 Chr.2: 53729390-53729390 on Build GRCh38
rs568494845 Chr.2: 53729714-53729714 on Build GRCh38
rs766035887 Chr.2: 53729419-53729419 on Build GRCh38
rs756430338 Chr.2: 53729414-53729415 on Build GRCh38
rs746802009 Chr.2: 53729490-53729490 on Build GRCh38
rs139465377 Chr.2: 53729625-53729625 on Build GRCh38
rs1009543653 Chr.2: 53729451-53729451 on Build GRCh38
rs766263785 Chr.2: 53729516-53729516 on Build GRCh38
rs763353934 Chr.2: 53729520-53729520 on Build GRCh38
rs761155524 Chr.2: 53729611-53729611 on Build GRCh38
rs763555580 Chr.2: 53729409-53729409 on Build GRCh38
rs34266611 Chr.2: 53729538-53729538 on Build GRCh38
rs748199918 Chr.2: 53729640-53729640 on Build GRCh38
rs778529075 Chr.2: 53729430-53729430 on Build GRCh38
rs534680350 Chr.2: 53729524-53729524 on Build GRCh38
rs778122815 Chr.2: 53729443-53729443 on Build GRCh38
rs557230195 Chr.2: 53729699-53729699 on Build GRCh38
rs1035214806 Chr.2: 53729302-53729302 on Build GRCh38
rs965751501 Chr.2: 53729722-53729722 on Build GRCh38
rs765976804 Chr.2: 53729525-53729525 on Build GRCh38

View Full Product Details