Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 190204391-190204884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIBCH
Entrez Gene ID
26275
Gene Name
3-hydroxyisobutyryl-CoA hydrolase
Gene Aliases
HIBYLCOAH
UniGene
Hs.656685
Gene Chromosome Location
Chr.2: 190204634-190320045 on Build GRCh38
Transcripts
NM_014362.3 NM_198047.2 XM_011510953.1 XM_011510954.1

SNP Detail

SNP ID Chromosome Location
rs1026733739 Chr.2: 190204602-190204602 on Build GRCh38
rs556373861 Chr.2: 190204756-190204756 on Build GRCh38
rs749282039 Chr.2: 190204499-190204500 on Build GRCh38
rs909756768 Chr.2: 190204433-190204433 on Build GRCh38
rs946489044 Chr.2: 190204480-190204480 on Build GRCh38
rs942528184 Chr.2: 190204434-190204434 on Build GRCh38
rs796951832 Chr.2: 190204452-190204452 on Build GRCh38
rs1033970723 Chr.2: 190204697-190204697 on Build GRCh38
rs931549477 Chr.2: 190204503-190204503 on Build GRCh38
rs984872952 Chr.2: 190204653-190204656 on Build GRCh38
rs913605087 Chr.2: 190204733-190204733 on Build GRCh38
rs11546637 Chr.2: 190204858-190204858 on Build GRCh38
rs1043489785 Chr.2: 190204483-190204483 on Build GRCh38
rs12618060 Chr.2: 190204504-190204504 on Build GRCh38
rs922432723 Chr.2: 190204707-190204707 on Build GRCh38
rs36117929 Chr.2: 190204802-190204802 on Build GRCh38
rs566798526 Chr.2: 190204449-190204449 on Build GRCh38
rs1058227 Chr.2: 190204750-190204750 on Build GRCh38
rs545251841 Chr.2: 190204808-190204808 on Build GRCh38
rs1036090847 Chr.2: 190204633-190204633 on Build GRCh38
rs975383747 Chr.2: 190204437-190204437 on Build GRCh38
rs575582739 Chr.2: 190204679-190204679 on Build GRCh38
rs993728211 Chr.2: 190204580-190204580 on Build GRCh38
rs773512252 Chr.2: 190204803-190204803 on Build GRCh38
rs1011431147 Chr.2: 190204715-190204715 on Build GRCh38
rs143748300 Chr.2: 190204512-190204512 on Build GRCh38
rs922492808 Chr.2: 190204444-190204444 on Build GRCh38
rs556312087 Chr.2: 190204484-190204484 on Build GRCh38
rs768921265 Chr.2: 190204683-190204683 on Build GRCh38
rs79966744 Chr.2: 190204668-190204668 on Build GRCh38
rs778803254 Chr.2: 190204829-190204837 on Build GRCh38
rs940193749 Chr.2: 190204551-190204551 on Build GRCh38
rs1002356808 Chr.2: 190204508-190204508 on Build GRCh38
rs866777035 Chr.2: 190204706-190204706 on Build GRCh38
rs749995609 Chr.2: 190204766-190204766 on Build GRCh38
rs963868645 Chr.2: 190204659-190204659 on Build GRCh38
rs181536793 Chr.2: 190204657-190204657 on Build GRCh38
rs904963052 Chr.2: 190204785-190204785 on Build GRCh38
rs1058230 Chr.2: 190204708-190204708 on Build GRCh38
rs151170023 Chr.2: 190204832-190204832 on Build GRCh38
rs921298371 Chr.2: 190204467-190204467 on Build GRCh38
rs988074027 Chr.2: 190204722-190204722 on Build GRCh38
rs892098783 Chr.2: 190204461-190204461 on Build GRCh38
rs578151670 Chr.2: 190204796-190204796 on Build GRCh38
rs977660690 Chr.2: 190204786-190204786 on Build GRCh38
rs751126327 Chr.2: 190204422-190204422 on Build GRCh38
rs967208983 Chr.2: 190204772-190204772 on Build GRCh38
rs554061859 Chr.2: 190204429-190204429 on Build GRCh38

View Full Product Details