Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24116790-24117389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF11
Entrez Gene ID
80344
Gene Name
DDB1 and CUL4 associated factor 11
Gene Aliases
GL014, PRO2389, WDR23
UniGene
Hs.525251
Gene Chromosome Location
Chr.14: 24114697-24125242 on Build GRCh38
Transcripts
NM_001163484.1 NM_025230.4 NM_181357.2 NR_028099.1 NR_028100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6075496 c.172G>T p.V58L Chr.14: 24116933-24116933 on Build GRCh38
6640334 c.269G>A p.R90Q Chr.14: 24117030-24117030 on Build GRCh38
4751344 c.305G>A p.R102Q Chr.14: 24117287-24117287 on Build GRCh38
5382130 c.268C>T p.R90* Chr.14: 24117029-24117029 on Build GRCh38
954882 c.231A>G p.E77E Chr.14: 24116992-24116992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112788511 Chr.14: 24117054-24117054 on Build GRCh38
rs746369292 Chr.14: 24117235-24117235 on Build GRCh38
rs74040011 Chr.14: 24116842-24116842 on Build GRCh38
rs150144513 Chr.14: 24117327-24117327 on Build GRCh38
rs748755162 Chr.14: 24116985-24116985 on Build GRCh38
rs142043152 Chr.14: 24117365-24117365 on Build GRCh38
rs764252072 Chr.14: 24116886-24116886 on Build GRCh38
rs777354392 Chr.14: 24117090-24117090 on Build GRCh38
rs376365336 Chr.14: 24116903-24116903 on Build GRCh38
rs772477569 Chr.14: 24117018-24117018 on Build GRCh38
rs755234465 Chr.14: 24117146-24117146 on Build GRCh38
rs7359087 Chr.14: 24116938-24116938 on Build GRCh38
rs995104946 Chr.14: 24117186-24117186 on Build GRCh38
rs975716068 Chr.14: 24117323-24117323 on Build GRCh38
rs770449338 Chr.14: 24116993-24116993 on Build GRCh38
rs537697700 Chr.14: 24117306-24117306 on Build GRCh38
rs760869014 Chr.14: 24116884-24116884 on Build GRCh38
rs370427032 Chr.14: 24117026-24117026 on Build GRCh38
rs927851621 Chr.14: 24116833-24116833 on Build GRCh38
rs946335325 Chr.14: 24116885-24116885 on Build GRCh38
rs902247311 Chr.14: 24116916-24116916 on Build GRCh38
rs751503186 Chr.14: 24116905-24116905 on Build GRCh38
rs753535701 Chr.14: 24117095-24117095 on Build GRCh38
rs923636478 Chr.14: 24117357-24117357 on Build GRCh38
rs765449557 Chr.14: 24117066-24117066 on Build GRCh38
rs962948440 Chr.14: 24117083-24117083 on Build GRCh38
rs764045255 Chr.14: 24117266-24117267 on Build GRCh38
rs746399014 Chr.14: 24117017-24117017 on Build GRCh38
rs749935959 Chr.14: 24117159-24117159 on Build GRCh38
rs370086589 Chr.14: 24117261-24117261 on Build GRCh38
rs760544752 Chr.14: 24117261-24117262 on Build GRCh38
rs552050421 Chr.14: 24117213-24117213 on Build GRCh38
rs767818870 Chr.14: 24116880-24116880 on Build GRCh38
rs951536435 Chr.14: 24117112-24117112 on Build GRCh38
rs762194456 Chr.14: 24117277-24117277 on Build GRCh38
rs761965309 Chr.14: 24117064-24117064 on Build GRCh38
rs925217648 Chr.14: 24117325-24117325 on Build GRCh38
rs1025626486 Chr.14: 24117108-24117108 on Build GRCh38
rs377181947 Chr.14: 24117249-24117249 on Build GRCh38
rs1025281852 Chr.14: 24117210-24117210 on Build GRCh38
rs199590153 Chr.14: 24117287-24117287 on Build GRCh38
rs532253968 Chr.14: 24117134-24117134 on Build GRCh38
rs1030236641 Chr.14: 24116948-24116948 on Build GRCh38
rs905369424 Chr.14: 24117114-24117114 on Build GRCh38
rs778350751 Chr.14: 24117225-24117225 on Build GRCh38
rs756726865 Chr.14: 24117096-24117096 on Build GRCh38
rs749832638 Chr.14: 24117352-24117352 on Build GRCh38
rs781632336 Chr.14: 24117275-24117275 on Build GRCh38
rs201461302 Chr.14: 24117025-24117025 on Build GRCh38
rs752368662 Chr.14: 24117079-24117079 on Build GRCh38
rs769195482 Chr.14: 24117271-24117271 on Build GRCh38
rs560866985 Chr.14: 24116900-24116900 on Build GRCh38
rs754772732 Chr.14: 24116912-24116912 on Build GRCh38
rs61744791 Chr.14: 24117358-24117358 on Build GRCh38
rs777406049 Chr.14: 24116950-24116950 on Build GRCh38
rs141724116 Chr.14: 24117302-24117302 on Build GRCh38
rs1050829196 Chr.14: 24117362-24117362 on Build GRCh38
rs368460521 Chr.14: 24117315-24117315 on Build GRCh38
rs531190872 Chr.14: 24116823-24116823 on Build GRCh38
rs763098384 Chr.14: 24117301-24117301 on Build GRCh38
rs754929890 Chr.14: 24117075-24117075 on Build GRCh38
rs981589238 Chr.14: 24117192-24117192 on Build GRCh38
rs760439162 Chr.14: 24117312-24117312 on Build GRCh38
rs747824055 Chr.14: 24116941-24116941 on Build GRCh38
rs898147889 Chr.14: 24117135-24117135 on Build GRCh38
rs751511135 Chr.14: 24117072-24117072 on Build GRCh38
rs775199533 Chr.14: 24116870-24116871 on Build GRCh38
rs529758287 Chr.14: 24116904-24116904 on Build GRCh38
rs372889586 Chr.14: 24117237-24117237 on Build GRCh38
rs769140311 Chr.14: 24117364-24117364 on Build GRCh38
rs375616424 Chr.14: 24117321-24117321 on Build GRCh38
rs374124611 Chr.14: 24117086-24117086 on Build GRCh38
rs773109585 Chr.14: 24117288-24117288 on Build GRCh38
rs748537443 Chr.14: 24117367-24117367 on Build GRCh38
rs189242071 Chr.14: 24117120-24117120 on Build GRCh38
rs759893916 Chr.14: 24116866-24116866 on Build GRCh38
rs373096009 Chr.14: 24116881-24116881 on Build GRCh38
rs990664090 Chr.14: 24117286-24117286 on Build GRCh38
rs774815144 Chr.14: 24117000-24117000 on Build GRCh38
rs761110023 Chr.14: 24117030-24117030 on Build GRCh38
rs767503738 Chr.14: 24117078-24117078 on Build GRCh38
rs567312085 Chr.14: 24117322-24117322 on Build GRCh38
rs1055012075 Chr.14: 24116879-24116879 on Build GRCh38
rs764374946 Chr.14: 24117043-24117043 on Build GRCh38
rs112454085 Chr.14: 24117060-24117060 on Build GRCh38
rs369906769 Chr.14: 24116942-24116942 on Build GRCh38
rs368388963 Chr.14: 24117353-24117353 on Build GRCh38

View Full Product Details