Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 85624619-85625152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLRT2
Entrez Gene ID
23768
Gene Name
fibronectin leucine rich transmembrane protein 2
Gene Aliases
-
UniGene
Hs.533710
Gene Chromosome Location
Chr.14: 85530067-85628696 on Build GRCh38
Transcripts
NM_013231.4 XM_005267490.3 XM_011536611.2 XM_017021129.1 XM_017021130.1 XM_017021131.1 XM_017021132.1 XM_017021133.1 XM_017021134.1 XM_017021135.1

SNP Detail

SNP ID Chromosome Location
rs977441924 Chr.14: 85625067-85625067 on Build GRCh38
rs556280623 Chr.14: 85624818-85624818 on Build GRCh38
rs28364861 Chr.14: 85624762-85624763 on Build GRCh38
rs760549634 Chr.14: 85624786-85624786 on Build GRCh38
rs1003404780 Chr.14: 85624839-85624839 on Build GRCh38
rs1053681263 Chr.14: 85624733-85624733 on Build GRCh38
rs576171573 Chr.14: 85624850-85624850 on Build GRCh38
rs577347767 Chr.14: 85625021-85625021 on Build GRCh38
rs749960220 Chr.14: 85624931-85624931 on Build GRCh38
rs1042134667 Chr.14: 85624767-85624767 on Build GRCh38
rs577253377 Chr.14: 85624904-85624904 on Build GRCh38
rs28364858 Chr.14: 85624757-85624758 on Build GRCh38
rs910351110 Chr.14: 85624692-85624692 on Build GRCh38
rs776245690 Chr.14: 85624899-85624899 on Build GRCh38
rs890474497 Chr.14: 85625053-85625053 on Build GRCh38
rs45549637 Chr.14: 85624878-85624878 on Build GRCh38
rs1017503225 Chr.14: 85625063-85625063 on Build GRCh38
rs910323388 Chr.14: 85624963-85624963 on Build GRCh38
rs866319164 Chr.14: 85625034-85625034 on Build GRCh38
rs1007500919 Chr.14: 85624756-85624756 on Build GRCh38
rs913802701 Chr.14: 85625026-85625026 on Build GRCh38
rs28364862 Chr.14: 85624778-85624778 on Build GRCh38
rs538192281 Chr.14: 85624760-85624760 on Build GRCh38
rs572388163 Chr.14: 85624742-85624742 on Build GRCh38
rs189922752 Chr.14: 85625051-85625051 on Build GRCh38
rs571562308 Chr.14: 85624642-85624642 on Build GRCh38
rs1007595626 Chr.14: 85625061-85625061 on Build GRCh38
rs907282396 Chr.14: 85624807-85624807 on Build GRCh38
rs538945823 Chr.14: 85624694-85624694 on Build GRCh38
rs555017171 Chr.14: 85624831-85624831 on Build GRCh38
rs927650841 Chr.14: 85624863-85624863 on Build GRCh38
rs866174085 Chr.14: 85624873-85624873 on Build GRCh38
rs954652461 Chr.14: 85624676-85624676 on Build GRCh38
rs28364860 Chr.14: 85624760-85624761 on Build GRCh38
rs1016457835 Chr.14: 85624882-85624882 on Build GRCh38
rs28364863 Chr.14: 85624898-85624898 on Build GRCh38
rs761577162 Chr.14: 85624903-85624903 on Build GRCh38
rs923025356 Chr.14: 85624656-85624656 on Build GRCh38
rs957830293 Chr.14: 85624988-85624988 on Build GRCh38
rs10145152 Chr.14: 85625071-85625071 on Build GRCh38
rs764922397 Chr.14: 85624930-85624930 on Build GRCh38
rs1043653963 Chr.14: 85624937-85624937 on Build GRCh38
rs547998231 Chr.14: 85625095-85625095 on Build GRCh38
rs1055777014 Chr.14: 85625109-85625109 on Build GRCh38
rs963664282 Chr.14: 85625066-85625066 on Build GRCh38
rs950477796 Chr.14: 85624842-85624843 on Build GRCh38
rs985972280 Chr.14: 85624683-85624683 on Build GRCh38
rs775410851 Chr.14: 85624669-85624669 on Build GRCh38
rs989344449 Chr.14: 85624997-85624997 on Build GRCh38
rs572967956 Chr.14: 85624775-85624775 on Build GRCh38
rs184271318 Chr.14: 85624707-85624707 on Build GRCh38
rs973667952 Chr.14: 85624815-85624815 on Build GRCh38
rs28364859 Chr.14: 85624759-85624759 on Build GRCh38
rs768200672 Chr.14: 85625114-85625114 on Build GRCh38
rs529667662 Chr.14: 85625088-85625088 on Build GRCh38
rs554036177 Chr.14: 85624913-85624913 on Build GRCh38
rs903308471 Chr.14: 85624655-85624655 on Build GRCh38
rs998944278 Chr.14: 85625012-85625012 on Build GRCh38

View Full Product Details