Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40737543-40738049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RMDN3
Entrez Gene ID
55177
Gene Name
regulator of microtubule dynamics 3
Gene Aliases
FAM82A2, FAM82C, RMD-3, RMD3, ptpip51
UniGene
Hs.511067
Gene Chromosome Location
Chr.15: 40735884-40755336 on Build GRCh38
Transcripts
NM_001304802.1 NM_001323894.1 NM_001323895.1 NM_001323896.1 NM_001323897.1 NM_018145.2 XM_011521755.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2269782 c.1181C>A p.P394H Chr.15: 40737671-40737671 on Build GRCh38
5383226 c.1125G>A p.Q375Q Chr.15: 40737965-40737965 on Build GRCh38
961292 c.1098C>G p.H366Q Chr.15: 40737992-40737992 on Build GRCh38
6190400 c.1147_1148insA p.T386fs*7 Chr.15: 40737704-40737705 on Build GRCh38
699917 c.1185C>T p.L395L Chr.15: 40737667-40737667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1031591459 Chr.15: 40737885-40737885 on Build GRCh38
rs1056465315 Chr.15: 40737790-40737790 on Build GRCh38
rs763785943 Chr.15: 40737770-40737770 on Build GRCh38
rs758361710 Chr.15: 40737591-40737591 on Build GRCh38
rs748146910 Chr.15: 40737947-40737947 on Build GRCh38
rs370226833 Chr.15: 40737956-40737956 on Build GRCh38
rs920837853 Chr.15: 40737670-40737670 on Build GRCh38
rs778564666 Chr.15: 40737721-40737721 on Build GRCh38
rs745401087 Chr.15: 40737657-40737657 on Build GRCh38
rs190232842 Chr.15: 40737719-40737719 on Build GRCh38
rs750634533 Chr.15: 40737704-40737704 on Build GRCh38
rs867578589 Chr.15: 40737695-40737695 on Build GRCh38
rs201314462 Chr.15: 40737725-40737725 on Build GRCh38
rs754655723 Chr.15: 40738026-40738026 on Build GRCh38
rs974085696 Chr.15: 40737637-40737637 on Build GRCh38
rs761812108 Chr.15: 40737949-40737949 on Build GRCh38
rs754215096 Chr.15: 40737772-40737772 on Build GRCh38
rs773047269 Chr.15: 40737874-40737874 on Build GRCh38
rs549238967 Chr.15: 40737617-40737617 on Build GRCh38
rs754170561 Chr.15: 40738003-40738003 on Build GRCh38
rs141170560 Chr.15: 40737971-40737971 on Build GRCh38
rs759602144 Chr.15: 40737759-40737759 on Build GRCh38
rs771730317 Chr.15: 40737724-40737724 on Build GRCh38
rs571623017 Chr.15: 40737769-40737769 on Build GRCh38
rs772944423 Chr.15: 40737955-40737955 on Build GRCh38
rs770574225 Chr.15: 40737638-40737638 on Build GRCh38
rs780643768 Chr.15: 40737603-40737603 on Build GRCh38
rs182138409 Chr.15: 40737938-40737938 on Build GRCh38
rs750548655 Chr.15: 40737583-40737583 on Build GRCh38
rs368286317 Chr.15: 40737840-40737840 on Build GRCh38
rs747834784 Chr.15: 40737570-40737570 on Build GRCh38
rs752047912 Chr.15: 40737607-40737607 on Build GRCh38
rs867019950 Chr.15: 40737777-40737777 on Build GRCh38
rs751265623 Chr.15: 40738025-40738025 on Build GRCh38
rs778478991 Chr.15: 40737647-40737647 on Build GRCh38
rs767621061 Chr.15: 40737974-40737974 on Build GRCh38
rs776666230 Chr.15: 40737726-40737726 on Build GRCh38
rs774297949 Chr.15: 40737758-40737758 on Build GRCh38
rs376437590 Chr.15: 40737844-40737844 on Build GRCh38
rs769752945 Chr.15: 40737739-40737739 on Build GRCh38
rs986557588 Chr.15: 40737687-40737687 on Build GRCh38
rs771071312 Chr.15: 40737969-40737969 on Build GRCh38
rs986666973 Chr.15: 40737880-40737880 on Build GRCh38
rs77019569 Chr.15: 40737898-40737898 on Build GRCh38
rs780818826 Chr.15: 40737945-40737945 on Build GRCh38
rs765493162 Chr.15: 40737580-40737580 on Build GRCh38
rs975345183 Chr.15: 40737577-40737577 on Build GRCh38
rs951308692 Chr.15: 40737686-40737686 on Build GRCh38
rs761646686 Chr.15: 40737681-40737681 on Build GRCh38
rs975549286 Chr.15: 40737746-40737746 on Build GRCh38
rs760575607 Chr.15: 40737671-40737671 on Build GRCh38
rs759639779 Chr.15: 40737972-40737972 on Build GRCh38
rs763204761 Chr.15: 40737706-40737706 on Build GRCh38
rs779702744 Chr.15: 40737931-40737931 on Build GRCh38
rs367810649 Chr.15: 40737744-40737744 on Build GRCh38
rs202155220 Chr.15: 40737674-40737674 on Build GRCh38
rs757993967 Chr.15: 40737923-40737923 on Build GRCh38
rs1001735542 Chr.15: 40737806-40737806 on Build GRCh38
rs747987701 Chr.15: 40737736-40737736 on Build GRCh38
rs760445620 Chr.15: 40737769-40737770 on Build GRCh38
rs377218919 Chr.15: 40737697-40737697 on Build GRCh38
rs536849601 Chr.15: 40737912-40737912 on Build GRCh38
rs764521318 Chr.15: 40738002-40738002 on Build GRCh38
rs773806391 Chr.15: 40737696-40737696 on Build GRCh38
rs1025603408 Chr.15: 40737868-40737868 on Build GRCh38
rs1034846826 Chr.15: 40737825-40737825 on Build GRCh38
rs376231723 Chr.15: 40738000-40738000 on Build GRCh38
rs780801716 Chr.15: 40737843-40737844 on Build GRCh38
rs773038957 Chr.15: 40737743-40737743 on Build GRCh38
rs755502599 Chr.15: 40737716-40737716 on Build GRCh38
rs771465329 Chr.15: 40737695-40737696 on Build GRCh38
rs776576292 Chr.15: 40737679-40737679 on Build GRCh38
rs757596064 Chr.15: 40738020-40738020 on Build GRCh38
rs771657797 Chr.15: 40737718-40737719 on Build GRCh38
rs755490824 Chr.15: 40737611-40737611 on Build GRCh38
rs946301252 Chr.15: 40737770-40737772 on Build GRCh38
rs1008603063 Chr.15: 40737902-40737902 on Build GRCh38
rs781612711 Chr.15: 40737718-40737718 on Build GRCh38
rs775030407 Chr.15: 40737663-40737663 on Build GRCh38
rs753158808 Chr.15: 40737765-40737765 on Build GRCh38
rs147774712 Chr.15: 40737913-40737913 on Build GRCh38
rs144155217 Chr.15: 40737767-40737767 on Build GRCh38
rs766421128 Chr.15: 40737715-40737715 on Build GRCh38
rs771439397 Chr.15: 40737661-40737661 on Build GRCh38
rs904371733 Chr.15: 40737858-40737858 on Build GRCh38
rs1046388789 Chr.15: 40737959-40737959 on Build GRCh38
rs148669324 Chr.15: 40738022-40738022 on Build GRCh38
rs775014079 Chr.15: 40737725-40737726 on Build GRCh38
rs754498140 Chr.15: 40737941-40737941 on Build GRCh38
rs766618371 Chr.15: 40737749-40737749 on Build GRCh38
rs748505864 Chr.15: 40737621-40737621 on Build GRCh38
rs11558808 Chr.15: 40738004-40738004 on Build GRCh38
rs780908773 Chr.15: 40738028-40738028 on Build GRCh38
rs763144882 Chr.15: 40737747-40737747 on Build GRCh38
rs745408248 Chr.15: 40737696-40737697 on Build GRCh38
rs537754484 Chr.15: 40737817-40737817 on Build GRCh38
rs928943502 Chr.15: 40737977-40737977 on Build GRCh38
rs745544738 Chr.15: 40737723-40737723 on Build GRCh38
rs118074807 Chr.15: 40737843-40737843 on Build GRCh38
rs776539286 Chr.15: 40737774-40737775 on Build GRCh38
rs769842993 Chr.15: 40737950-40737950 on Build GRCh38

View Full Product Details