Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129637529-129638124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB6
Entrez Gene ID
140459
Gene Name
ankyrin repeat and SOCS box containing 6
Gene Aliases
-
UniGene
Hs.125037
Gene Chromosome Location
Chr.9: 129634604-129642169 on Build GRCh38
Transcripts
NM_001202403.1 NM_017873.3 NM_177999.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6605590 c.1234G>T p.E412* Chr.9: 129637822-129637822 on Build GRCh38
4761476 c.1217A>C p.K406T Chr.9: 129637839-129637839 on Build GRCh38
4769807 c.972C>A p.R324R Chr.9: 129638084-129638084 on Build GRCh38
6605588 c.1050T>G p.P350P Chr.9: 129638006-129638006 on Build GRCh38
1330698 c.1166C>T p.P389L Chr.9: 129637890-129637890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748986072 Chr.9: 129637761-129637761 on Build GRCh38
rs774643625 Chr.9: 129637715-129637715 on Build GRCh38
rs768263683 Chr.9: 129637768-129637768 on Build GRCh38
rs757771802 Chr.9: 129637836-129637836 on Build GRCh38
rs747416480 Chr.9: 129637759-129637759 on Build GRCh38
rs1026649921 Chr.9: 129637985-129637985 on Build GRCh38
rs765516002 Chr.9: 129637804-129637804 on Build GRCh38
rs375847126 Chr.9: 129637769-129637769 on Build GRCh38
rs952521220 Chr.9: 129638006-129638006 on Build GRCh38
rs28438775 Chr.9: 129637807-129637807 on Build GRCh38
rs767641601 Chr.9: 129638084-129638084 on Build GRCh38
rs998050722 Chr.9: 129637915-129637915 on Build GRCh38
rs780392964 Chr.9: 129637921-129637921 on Build GRCh38
rs567076408 Chr.9: 129637710-129637710 on Build GRCh38
rs374183739 Chr.9: 129637952-129637952 on Build GRCh38
rs1010617471 Chr.9: 129637554-129637555 on Build GRCh38
rs148649978 Chr.9: 129638017-129638017 on Build GRCh38
rs868422522 Chr.9: 129637596-129637596 on Build GRCh38
rs927477222 Chr.9: 129637815-129637815 on Build GRCh38
rs142387881 Chr.9: 129637963-129637963 on Build GRCh38
rs771015223 Chr.9: 129637942-129637942 on Build GRCh38
rs745862880 Chr.9: 129638052-129638052 on Build GRCh38
rs760654457 Chr.9: 129637787-129637787 on Build GRCh38
rs909589631 Chr.9: 129637655-129637655 on Build GRCh38
rs1057196838 Chr.9: 129637832-129637832 on Build GRCh38
rs55757508 Chr.9: 129637613-129637613 on Build GRCh38
rs374558502 Chr.9: 129637870-129637870 on Build GRCh38
rs761209551 Chr.9: 129638092-129638092 on Build GRCh38
rs780849303 Chr.9: 129638042-129638042 on Build GRCh38
rs768314791 Chr.9: 129637854-129637854 on Build GRCh38
rs778475042 Chr.9: 129637857-129637857 on Build GRCh38
rs752799886 Chr.9: 129637899-129637899 on Build GRCh38
rs768013886 Chr.9: 129637996-129637996 on Build GRCh38
rs1028138894 Chr.9: 129637683-129637683 on Build GRCh38
rs978879389 Chr.9: 129638097-129638097 on Build GRCh38
rs1043063527 Chr.9: 129637878-129637878 on Build GRCh38
rs781625054 Chr.9: 129637918-129637918 on Build GRCh38
rs768562696 Chr.9: 129637963-129637965 on Build GRCh38
rs985670679 Chr.9: 129637949-129637949 on Build GRCh38
rs767503499 Chr.9: 129637779-129637779 on Build GRCh38
rs1029438199 Chr.9: 129637593-129637594 on Build GRCh38
rs985223756 Chr.9: 129637625-129637625 on Build GRCh38
rs762017113 Chr.9: 129638072-129638072 on Build GRCh38
rs771732219 Chr.9: 129637934-129637934 on Build GRCh38
rs778093305 Chr.9: 129637758-129637758 on Build GRCh38
rs763136780 Chr.9: 129637970-129637970 on Build GRCh38
rs953914646 Chr.9: 129637600-129637600 on Build GRCh38
rs750994437 Chr.9: 129638004-129638004 on Build GRCh38
rs189258281 Chr.9: 129638085-129638085 on Build GRCh38
rs200707909 Chr.9: 129637889-129637889 on Build GRCh38
rs1029572669 Chr.9: 129637916-129637916 on Build GRCh38
rs764334444 Chr.9: 129637975-129637975 on Build GRCh38
rs769834879 Chr.9: 129638058-129638058 on Build GRCh38
rs927497086 Chr.9: 129638099-129638099 on Build GRCh38
rs572351581 Chr.9: 129637692-129637692 on Build GRCh38
rs918251247 Chr.9: 129638001-129638001 on Build GRCh38
rs993902398 Chr.9: 129637984-129637984 on Build GRCh38
rs754198641 Chr.9: 129638018-129638018 on Build GRCh38
rs769783503 Chr.9: 129637957-129637957 on Build GRCh38
rs573790742 Chr.9: 129638064-129638064 on Build GRCh38
rs374889902 Chr.9: 129637684-129637684 on Build GRCh38
rs772763780 Chr.9: 129637874-129637874 on Build GRCh38
rs775175943 Chr.9: 129638059-129638059 on Build GRCh38
rs776978337 Chr.9: 129637760-129637760 on Build GRCh38
rs1019329270 Chr.9: 129637700-129637700 on Build GRCh38
rs759879968 Chr.9: 129637801-129637801 on Build GRCh38
rs141198008 Chr.9: 129638026-129638026 on Build GRCh38
rs149393165 Chr.9: 129637833-129637833 on Build GRCh38
rs138458518 Chr.9: 129637931-129637931 on Build GRCh38
rs199686158 Chr.9: 129637905-129637905 on Build GRCh38
rs1010241690 Chr.9: 129637557-129637557 on Build GRCh38
rs568864328 Chr.9: 129637824-129637824 on Build GRCh38
rs776601036 Chr.9: 129637890-129637890 on Build GRCh38
rs759843312 Chr.9: 129638027-129638027 on Build GRCh38
rs369236250 Chr.9: 129637991-129637991 on Build GRCh38
rs764140909 Chr.9: 129637906-129637906 on Build GRCh38
rs1007923305 Chr.9: 129637699-129637699 on Build GRCh38
rs1042304178 Chr.9: 129637558-129637558 on Build GRCh38
rs777189353 Chr.9: 129637937-129637937 on Build GRCh38
rs756761460 Chr.9: 129638028-129638028 on Build GRCh38
rs891768702 Chr.9: 129637552-129637552 on Build GRCh38
rs145897651 Chr.9: 129637850-129637850 on Build GRCh38
rs138887880 Chr.9: 129637825-129637825 on Build GRCh38
rs774562163 Chr.9: 129638066-129638066 on Build GRCh38
rs368837136 Chr.9: 129637561-129637562 on Build GRCh38
rs371398374 Chr.9: 129637917-129637917 on Build GRCh38
rs368489586 Chr.9: 129637914-129637914 on Build GRCh38
rs762815742 Chr.9: 129637924-129637924 on Build GRCh38
rs766870193 Chr.9: 129638096-129638096 on Build GRCh38
rs533555558 Chr.9: 129637666-129637666 on Build GRCh38
rs746489802 Chr.9: 129637938-129637938 on Build GRCh38
rs774314449 Chr.9: 129637986-129637986 on Build GRCh38
rs747697267 Chr.9: 129637861-129637861 on Build GRCh38
rs1001217380 Chr.9: 129637588-129637588 on Build GRCh38
rs534325718 Chr.9: 129637892-129637892 on Build GRCh38
rs759356227 Chr.9: 129637891-129637891 on Build GRCh38
rs906029847 Chr.9: 129637810-129637810 on Build GRCh38
rs750796506 Chr.9: 129637919-129637919 on Build GRCh38
rs776652325 Chr.9: 129637944-129637944 on Build GRCh38
rs749059356 Chr.9: 129638062-129638062 on Build GRCh38
rs529525585 Chr.9: 129637791-129637791 on Build GRCh38
rs1019294578 Chr.9: 129637964-129637964 on Build GRCh38
rs772956445 Chr.9: 129637786-129637786 on Build GRCh38
rs1558871 Chr.9: 129637574-129637574 on Build GRCh38
rs545122687 Chr.9: 129637562-129637562 on Build GRCh38
rs781573949 Chr.9: 129637838-129637838 on Build GRCh38
rs760577422 Chr.9: 129637880-129637880 on Build GRCh38
rs145932765 Chr.9: 129637992-129637992 on Build GRCh38

View Full Product Details