Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 31400976-31401537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DROSHA
Entrez Gene ID
29102
Gene Name
drosha ribonuclease III
Gene Aliases
ETOHI2, HSA242976, RANSE3L, RN3, RNASE3L, RNASEN
UniGene
Hs.97997
Gene Chromosome Location
Chr.5: 31400494-31532175 on Build GRCh38
Transcripts
NM_001100412.1 NM_013235.4 XM_005248291.3 XM_005248292.3 XM_005248293.3 XM_005248294.3 XM_017009399.1 XM_017009400.1 XM_017009401.1 XM_017009402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3827928 c.4070G>T p.W1357L Chr.5: 31401487-31401487 on Build GRCh38
1634185 c.4030A>G p.I1344V Chr.5: 31401527-31401527 on Build GRCh38
3854663 c.4037G>A p.R1346Q Chr.5: 31401520-31401520 on Build GRCh38
280610 c.4024C>T p.R1342W Chr.5: 31401533-31401533 on Build GRCh38
6649328 c.4094_4095delAG p.E1365fs*3 Chr.5: 31401462-31401463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760456686 Chr.5: 31401331-31401331 on Build GRCh38
rs61751196 Chr.5: 31401467-31401467 on Build GRCh38
rs780966031 Chr.5: 31401345-31401345 on Build GRCh38
rs759311633 Chr.5: 31401318-31401318 on Build GRCh38
rs775237795 Chr.5: 31401328-31401328 on Build GRCh38
rs896572470 Chr.5: 31401082-31401082 on Build GRCh38
rs150603393 Chr.5: 31401285-31401285 on Build GRCh38
rs536092006 Chr.5: 31401295-31401295 on Build GRCh38
rs72547276 Chr.5: 31401393-31401393 on Build GRCh38
rs187777012 Chr.5: 31401322-31401322 on Build GRCh38
rs116086102 Chr.5: 31401040-31401040 on Build GRCh38
rs769842401 Chr.5: 31401299-31401299 on Build GRCh38
rs777815779 Chr.5: 31401433-31401433 on Build GRCh38
rs774167018 Chr.5: 31401308-31401308 on Build GRCh38
rs896515701 Chr.5: 31401504-31401504 on Build GRCh38
rs769605158 Chr.5: 31401337-31401344 on Build GRCh38
rs1002580624 Chr.5: 31401024-31401024 on Build GRCh38
rs770922289 Chr.5: 31401463-31401463 on Build GRCh38
rs1055527452 Chr.5: 31401452-31401452 on Build GRCh38
rs139722849 Chr.5: 31401303-31401303 on Build GRCh38
rs1024564515 Chr.5: 31401005-31401005 on Build GRCh38
rs941060593 Chr.5: 31401370-31401370 on Build GRCh38
rs774306960 Chr.5: 31401481-31401481 on Build GRCh38
rs550965893 Chr.5: 31401011-31401011 on Build GRCh38
rs1013653152 Chr.5: 31401108-31401108 on Build GRCh38
rs199506308 Chr.5: 31401507-31401507 on Build GRCh38
rs751916206 Chr.5: 31401338-31401338 on Build GRCh38
rs761935330 Chr.5: 31401417-31401417 on Build GRCh38
rs766661064 Chr.5: 31401170-31401170 on Build GRCh38
rs376216811 Chr.5: 31401457-31401457 on Build GRCh38
rs771971658 Chr.5: 31401510-31401510 on Build GRCh38
rs766853644 Chr.5: 31401359-31401359 on Build GRCh38
rs114594866 Chr.5: 31401143-31401143 on Build GRCh38
rs559000514 Chr.5: 31401459-31401459 on Build GRCh38
rs1032974920 Chr.5: 31401227-31401227 on Build GRCh38
rs547345067 Chr.5: 31401233-31401233 on Build GRCh38
rs909568038 Chr.5: 31401368-31401368 on Build GRCh38
rs772793274 Chr.5: 31401471-31401472 on Build GRCh38
rs1014491371 Chr.5: 31400995-31400995 on Build GRCh38
rs10719 Chr.5: 31401340-31401340 on Build GRCh38
rs951414007 Chr.5: 31401325-31401325 on Build GRCh38
rs755317391 Chr.5: 31401377-31401377 on Build GRCh38
rs750919146 Chr.5: 31401056-31401056 on Build GRCh38
rs764335536 Chr.5: 31401272-31401272 on Build GRCh38
rs768904826 Chr.5: 31401283-31401283 on Build GRCh38
rs1007629334 Chr.5: 31401172-31401172 on Build GRCh38
rs758819754 Chr.5: 31401347-31401347 on Build GRCh38
rs905793180 Chr.5: 31401023-31401023 on Build GRCh38
rs1001001871 Chr.5: 31401209-31401209 on Build GRCh38
rs72547275 Chr.5: 31401335-31401335 on Build GRCh38
rs752035593 Chr.5: 31401373-31401373 on Build GRCh38
rs781582130 Chr.5: 31401392-31401392 on Build GRCh38
rs1042424366 Chr.5: 31401516-31401516 on Build GRCh38
rs758884859 Chr.5: 31401074-31401074 on Build GRCh38
rs747946391 Chr.5: 31401313-31401314 on Build GRCh38
rs773315864 Chr.5: 31401302-31401302 on Build GRCh38
rs756394925 Chr.5: 31401427-31401427 on Build GRCh38
rs533297896 Chr.5: 31401472-31401472 on Build GRCh38

View Full Product Details