Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31730248-31730742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC1
Entrez Gene ID
1192
Gene Name
chloride intracellular channel 1
Gene Aliases
G6, NCC27
UniGene
Hs.414565
Gene Chromosome Location
Chr.6: 31730581-31737318 on Build GRCh38
Transcripts
NM_001287593.1 NM_001287594.1 NM_001288.4 NM_001303007.1 NM_013974.2

Gene Symbol
DDAH2
Entrez Gene ID
23564
Gene Name
dimethylarginine dimethylaminohydrolase 2
Gene Aliases
DDAH, DDAHII, G6a, HEL-S-277, NG30
UniGene
Hs.247362
Gene Chromosome Location
Chr.6: 31727037-31730265 on Build GRCh38
Transcripts
NM_001287593.1 NM_001287594.1 NM_001288.4 NM_001303007.1 NM_013974.2

SNP Detail

SNP ID Chromosome Location
rs532450245 Chr.6: 31730295-31730295 on Build GRCh38
rs907589613 Chr.6: 31730716-31730716 on Build GRCh38
rs909318950 Chr.6: 31730408-31730408 on Build GRCh38
rs978725586 Chr.6: 31730462-31730462 on Build GRCh38
rs904320476 Chr.6: 31730717-31730717 on Build GRCh38
rs1036395858 Chr.6: 31730419-31730419 on Build GRCh38
rs2272592 Chr.6: 31730575-31730575 on Build GRCh38
rs1037582784 Chr.6: 31730361-31730361 on Build GRCh38
rs894616685 Chr.6: 31730518-31730518 on Build GRCh38
rs374730014 Chr.6: 31730320-31730320 on Build GRCh38
rs1004479295 Chr.6: 31730355-31730355 on Build GRCh38
rs960143053 Chr.6: 31730274-31730274 on Build GRCh38
rs942871522 Chr.6: 31730303-31730303 on Build GRCh38
rs1031547519 Chr.6: 31730323-31730323 on Build GRCh38
rs1021357678 Chr.6: 31730581-31730581 on Build GRCh38
rs1804846 Chr.6: 31730675-31730675 on Build GRCh38
rs1014361989 Chr.6: 31730431-31730431 on Build GRCh38
rs938878068 Chr.6: 31730461-31730461 on Build GRCh38
rs1034183920 Chr.6: 31730400-31730400 on Build GRCh38
rs895911821 Chr.6: 31730412-31730412 on Build GRCh38
rs926188235 Chr.6: 31730301-31730301 on Build GRCh38
rs1001808204 Chr.6: 31730391-31730391 on Build GRCh38
rs977076544 Chr.6: 31730335-31730335 on Build GRCh38
rs1057509665 Chr.6: 31730472-31730472 on Build GRCh38
rs987197411 Chr.6: 31730624-31730624 on Build GRCh38
rs549802415 Chr.6: 31730279-31730279 on Build GRCh38
rs558538356 Chr.6: 31730698-31730698 on Build GRCh38
rs566512762 Chr.6: 31730338-31730338 on Build GRCh38
rs965299483 Chr.6: 31730334-31730334 on Build GRCh38
rs805304 Chr.6: 31730311-31730311 on Build GRCh38
rs897839342 Chr.6: 31730458-31730458 on Build GRCh38
rs571524624 Chr.6: 31730280-31730280 on Build GRCh38
rs964327461 Chr.6: 31730480-31730480 on Build GRCh38
rs753709294 Chr.6: 31730545-31730545 on Build GRCh38
rs112304772 Chr.6: 31730604-31730604 on Build GRCh38
rs1033715363 Chr.6: 31730467-31730467 on Build GRCh38
rs967343599 Chr.6: 31730281-31730281 on Build GRCh38
rs942005585 Chr.6: 31730411-31730411 on Build GRCh38
rs11549122 Chr.6: 31730682-31730682 on Build GRCh38
rs934129872 Chr.6: 31730598-31730598 on Build GRCh38
rs1042297155 Chr.6: 31730446-31730453 on Build GRCh38
rs886118015 Chr.6: 31730346-31730346 on Build GRCh38
rs533659033 Chr.6: 31730364-31730364 on Build GRCh38
rs536094065 Chr.6: 31730328-31730328 on Build GRCh38
rs986217663 Chr.6: 31730375-31730375 on Build GRCh38
rs976012252 Chr.6: 31730540-31730541 on Build GRCh38

View Full Product Details