Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 90415167-90415692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN12
Entrez Gene ID
9069
Gene Name
claudin 12
Gene Aliases
-
UniGene
Hs.731917
Gene Chromosome Location
Chr.7: 90403334-90415954 on Build GRCh38
Transcripts
NM_001185072.2 NM_001185073.2 NM_012129.4 XM_017012892.1 XM_017012893.1

Gene Symbol
LOC102723899
Entrez Gene ID
102723899
Gene Name
pituitary tumor-transforming gene 1 protein-interacting protein
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.7: 90403379-90513397 on Build GRCh38
Transcripts
NM_001185072.2 NM_001185073.2 NM_012129.4 XM_017012892.1 XM_017012893.1

SNP Detail

SNP ID Chromosome Location
rs1041890726 Chr.7: 90415630-90415630 on Build GRCh38
rs186714887 Chr.7: 90415496-90415496 on Build GRCh38
rs774762433 Chr.7: 90415417-90415417 on Build GRCh38
rs893323586 Chr.7: 90415338-90415338 on Build GRCh38
rs767405026 Chr.7: 90415416-90415416 on Build GRCh38
rs1047849387 Chr.7: 90415577-90415577 on Build GRCh38
rs751187006 Chr.7: 90415290-90415290 on Build GRCh38
rs901114812 Chr.7: 90415640-90415640 on Build GRCh38
rs745517196 Chr.7: 90415580-90415580 on Build GRCh38
rs1035394292 Chr.7: 90415432-90415432 on Build GRCh38
rs558116406 Chr.7: 90415508-90415508 on Build GRCh38
rs572884216 Chr.7: 90415511-90415511 on Build GRCh38
rs923308201 Chr.7: 90415575-90415575 on Build GRCh38
rs1050526265 Chr.7: 90415321-90415321 on Build GRCh38
rs1048883 Chr.7: 90415328-90415328 on Build GRCh38
rs769109092 Chr.7: 90415287-90415287 on Build GRCh38
rs1035319829 Chr.7: 90415202-90415202 on Build GRCh38
rs556800887 Chr.7: 90415374-90415374 on Build GRCh38
rs943930679 Chr.7: 90415286-90415286 on Build GRCh38
rs397829267 Chr.7: 90415244-90415244 on Build GRCh38
rs754831345 Chr.7: 90415395-90415395 on Build GRCh38
rs1026715065 Chr.7: 90415482-90415482 on Build GRCh38
rs952560771 Chr.7: 90415230-90415230 on Build GRCh38
rs947424244 Chr.7: 90415627-90415627 on Build GRCh38
rs976635967 Chr.7: 90415567-90415567 on Build GRCh38
rs184613376 Chr.7: 90415285-90415285 on Build GRCh38
rs901086369 Chr.7: 90415267-90415267 on Build GRCh38
rs931973835 Chr.7: 90415591-90415591 on Build GRCh38
rs781641197 Chr.7: 90415458-90415458 on Build GRCh38
rs1012698606 Chr.7: 90415473-90415473 on Build GRCh38
rs986645515 Chr.7: 90415605-90415605 on Build GRCh38
rs576737071 Chr.7: 90415652-90415652 on Build GRCh38
rs397759219 Chr.7: 90415243-90415243 on Build GRCh38
rs9731 Chr.7: 90415631-90415631 on Build GRCh38
rs568718729 Chr.7: 90415477-90415477 on Build GRCh38
rs544649141 Chr.7: 90415609-90415609 on Build GRCh38
rs1002551520 Chr.7: 90415190-90415190 on Build GRCh38
rs938947137 Chr.7: 90415633-90415633 on Build GRCh38
rs894138443 Chr.7: 90415449-90415449 on Build GRCh38
rs1008866447 Chr.7: 90415203-90415203 on Build GRCh38
rs970325178 Chr.7: 90415430-90415430 on Build GRCh38
rs1027265526 Chr.7: 90415212-90415212 on Build GRCh38
rs755973015 Chr.7: 90415576-90415576 on Build GRCh38
rs577566452 Chr.7: 90415273-90415273 on Build GRCh38
rs546965228 Chr.7: 90415207-90415207 on Build GRCh38
rs930764716 Chr.7: 90415521-90415521 on Build GRCh38
rs770889312 Chr.7: 90415351-90415351 on Build GRCh38
rs999951759 Chr.7: 90415315-90415315 on Build GRCh38
rs932109832 Chr.7: 90415304-90415304 on Build GRCh38
rs571487034 Chr.7: 90415237-90415237 on Build GRCh38
rs958446477 Chr.7: 90415204-90415204 on Build GRCh38
rs370567033 Chr.7: 90415549-90415549 on Build GRCh38
rs992452555 Chr.7: 90415402-90415402 on Build GRCh38
rs1042255724 Chr.7: 90415354-90415354 on Build GRCh38
rs982647638 Chr.7: 90415498-90415498 on Build GRCh38
rs191175070 Chr.7: 90415524-90415524 on Build GRCh38
rs1031349699 Chr.7: 90415343-90415343 on Build GRCh38

View Full Product Details