Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 107249973-107250539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPT1
Entrez Gene ID
284
Gene Name
angiopoietin 1
Gene Aliases
AGP1, AGPT, ANG1
UniGene
Hs.369675
Gene Chromosome Location
Chr.8: 107249482-107498055 on Build GRCh38
Transcripts
NM_001146.4 NM_001199859.2 NM_001314051.1

SNP Detail

SNP ID Chromosome Location
rs989979726 Chr.8: 107250117-107250117 on Build GRCh38
rs375717187 Chr.8: 107250441-107250441 on Build GRCh38
rs41343148 Chr.8: 107250256-107250256 on Build GRCh38
rs965591344 Chr.8: 107250480-107250480 on Build GRCh38
rs140916829 Chr.8: 107250065-107250065 on Build GRCh38
rs535445506 Chr.8: 107250479-107250479 on Build GRCh38
rs775305790 Chr.8: 107250261-107250261 on Build GRCh38
rs568073405 Chr.8: 107250441-107250442 on Build GRCh38
rs1002266763 Chr.8: 107250124-107250124 on Build GRCh38
rs1053386243 Chr.8: 107250287-107250287 on Build GRCh38
rs532447883 Chr.8: 107250276-107250276 on Build GRCh38
rs965770183 Chr.8: 107250067-107250067 on Build GRCh38
rs1031860699 Chr.8: 107250418-107250421 on Build GRCh38
rs200580638 Chr.8: 107250440-107250441 on Build GRCh38
rs187668099 Chr.8: 107250272-107250272 on Build GRCh38
rs957140079 Chr.8: 107250438-107250438 on Build GRCh38
rs895721706 Chr.8: 107250304-107250304 on Build GRCh38
rs983391459 Chr.8: 107250174-107250174 on Build GRCh38
rs537919914 Chr.8: 107250423-107250428 on Build GRCh38
rs183291388 Chr.8: 107250221-107250221 on Build GRCh38
rs41415945 Chr.8: 107250034-107250034 on Build GRCh38
rs1013569283 Chr.8: 107250096-107250096 on Build GRCh38
rs374681967 Chr.8: 107250179-107250179 on Build GRCh38
rs867389223 Chr.8: 107250323-107250323 on Build GRCh38
rs747345938 Chr.8: 107250042-107250042 on Build GRCh38
rs1002214624 Chr.8: 107250401-107250401 on Build GRCh38
rs1042752285 Chr.8: 107250274-107250274 on Build GRCh38
rs2507800 Chr.8: 107250441-107250441 on Build GRCh38
rs867060598 Chr.8: 107250075-107250075 on Build GRCh38
rs897897124 Chr.8: 107250229-107250229 on Build GRCh38
rs1014560352 Chr.8: 107250305-107250305 on Build GRCh38
rs924479700 Chr.8: 107250098-107250098 on Build GRCh38
rs1018583324 Chr.8: 107250285-107250285 on Build GRCh38
rs769502225 Chr.8: 107250214-107250214 on Build GRCh38
rs41421548 Chr.8: 107250146-107250146 on Build GRCh38
rs1016145936 Chr.8: 107250476-107250476 on Build GRCh38
rs1025424980 Chr.8: 107250115-107250115 on Build GRCh38
rs977042540 Chr.8: 107250231-107250231 on Build GRCh38
rs529587302 Chr.8: 107250010-107250010 on Build GRCh38
rs1007726861 Chr.8: 107250283-107250283 on Build GRCh38
rs553725542 Chr.8: 107250322-107250322 on Build GRCh38
rs1028352049 Chr.8: 107250027-107250027 on Build GRCh38
rs1045099371 Chr.8: 107250158-107250158 on Build GRCh38
rs888573451 Chr.8: 107250444-107250444 on Build GRCh38
rs764561148 Chr.8: 107250460-107250460 on Build GRCh38
rs927913397 Chr.8: 107250168-107250168 on Build GRCh38
rs76514844 Chr.8: 107250280-107250280 on Build GRCh38
rs1042804894 Chr.8: 107250002-107250002 on Build GRCh38
rs41364248 Chr.8: 107250154-107250154 on Build GRCh38
rs905155982 Chr.8: 107250324-107250324 on Build GRCh38
rs771920254 Chr.8: 107250003-107250003 on Build GRCh38
rs568972530 Chr.8: 107250252-107250252 on Build GRCh38
rs969391773 Chr.8: 107250118-107250118 on Build GRCh38
rs1004368765 Chr.8: 107250456-107250456 on Build GRCh38
rs543050785 Chr.8: 107250030-107250030 on Build GRCh38

View Full Product Details