Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 112152288-112152866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPAM
Entrez Gene ID
57678
Gene Name
glycerol-3-phosphate acyltransferase, mitochondrial
Gene Aliases
GPAT, GPAT1
UniGene
Hs.42586
Gene Chromosome Location
Chr.10: 112149864-112215395 on Build GRCh38
Transcripts
NM_001244949.1 NM_020918.5 XM_005269998.1

SNP Detail

SNP ID Chromosome Location
rs970995784 Chr.10: 112152644-112152644 on Build GRCh38
rs1004558987 Chr.10: 112152496-112152496 on Build GRCh38
rs756958398 Chr.10: 112152715-112152715 on Build GRCh38
rs1050532950 Chr.10: 112152507-112152507 on Build GRCh38
rs182129505 Chr.10: 112152718-112152718 on Build GRCh38
rs951766132 Chr.10: 112152492-112152492 on Build GRCh38
rs1048099516 Chr.10: 112152591-112152591 on Build GRCh38
rs570007655 Chr.10: 112152698-112152698 on Build GRCh38
rs765996273 Chr.10: 112152389-112152389 on Build GRCh38
rs953389582 Chr.10: 112152721-112152721 on Build GRCh38
rs1036505733 Chr.10: 112152532-112152532 on Build GRCh38
rs764283714 Chr.10: 112152645-112152645 on Build GRCh38
rs571149826 Chr.10: 112152820-112152820 on Build GRCh38
rs368181251 Chr.10: 112152646-112152646 on Build GRCh38
rs111633881 Chr.10: 112152683-112152683 on Build GRCh38
rs995049651 Chr.10: 112152566-112152566 on Build GRCh38
rs539899844 Chr.10: 112152694-112152694 on Build GRCh38
rs919391405 Chr.10: 112152794-112152794 on Build GRCh38
rs558590211 Chr.10: 112152803-112152803 on Build GRCh38
rs986650419 Chr.10: 112152724-112152724 on Build GRCh38
rs548153379 Chr.10: 112152558-112152558 on Build GRCh38
rs1023365217 Chr.10: 112152612-112152612 on Build GRCh38
rs566352411 Chr.10: 112152567-112152567 on Build GRCh38
rs970583785 Chr.10: 112152411-112152411 on Build GRCh38
rs944750997 Chr.10: 112152758-112152758 on Build GRCh38
rs969244223 Chr.10: 112152834-112152834 on Build GRCh38
rs939230892 Chr.10: 112152317-112152317 on Build GRCh38
rs960671118 Chr.10: 112152500-112152500 on Build GRCh38
rs960677166 Chr.10: 112152815-112152815 on Build GRCh38
rs956623396 Chr.10: 112152693-112152693 on Build GRCh38
rs1053496908 Chr.10: 112152369-112152369 on Build GRCh38
rs374009875 Chr.10: 112152778-112152778 on Build GRCh38
rs189862805 Chr.10: 112152515-112152515 on Build GRCh38
rs565489233 Chr.10: 112152546-112152546 on Build GRCh38
rs550948040 Chr.10: 112152652-112152652 on Build GRCh38
rs571755745 Chr.10: 112152360-112152360 on Build GRCh38
rs890644670 Chr.10: 112152516-112152516 on Build GRCh38
rs542239176 Chr.10: 112152365-112152365 on Build GRCh38
rs944881332 Chr.10: 112152524-112152524 on Build GRCh38
rs900698842 Chr.10: 112152471-112152471 on Build GRCh38
rs754092463 Chr.10: 112152685-112152685 on Build GRCh38
rs568364945 Chr.10: 112152446-112152446 on Build GRCh38
rs532743406 Chr.10: 112152549-112152549 on Build GRCh38
rs534161263 Chr.10: 112152331-112152331 on Build GRCh38
rs1005011590 Chr.10: 112152733-112152733 on Build GRCh38
rs186973964 Chr.10: 112152375-112152375 on Build GRCh38
rs1036535872 Chr.10: 112152780-112152780 on Build GRCh38
rs140409361 Chr.10: 112152499-112152499 on Build GRCh38
rs549799749 Chr.10: 112152521-112152521 on Build GRCh38
rs144174877 Chr.10: 112152824-112152824 on Build GRCh38
rs544914493 Chr.10: 112152326-112152326 on Build GRCh38
rs915260560 Chr.10: 112152716-112152716 on Build GRCh38
rs911994924 Chr.10: 112152749-112152749 on Build GRCh38
rs900751538 Chr.10: 112152673-112152673 on Build GRCh38
rs979227995 Chr.10: 112152555-112152555 on Build GRCh38
rs774597322 Chr.10: 112152665-112152665 on Build GRCh38
rs1012448858 Chr.10: 112152606-112152606 on Build GRCh38
rs1009538180 Chr.10: 112152376-112152376 on Build GRCh38
rs923846377 Chr.10: 112152456-112152456 on Build GRCh38
rs557314766 Chr.10: 112152506-112152506 on Build GRCh38
rs915204437 Chr.10: 112152503-112152503 on Build GRCh38
rs1019122468 Chr.10: 112152394-112152394 on Build GRCh38
rs561073906 Chr.10: 112152482-112152482 on Build GRCh38
rs575682228 Chr.10: 112152605-112152605 on Build GRCh38
rs947877342 Chr.10: 112152358-112152358 on Build GRCh38
rs935218310 Chr.10: 112152480-112152480 on Build GRCh38

View Full Product Details