Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55684023-55684622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA7
Entrez Gene ID
3679
Gene Name
integrin subunit alpha 7
Gene Aliases
-
UniGene
Hs.524484
Gene Chromosome Location
Chr.12: 55684568-55716037 on Build GRCh38
Transcripts
NM_001144996.1 NM_001144997.1 NM_002206.2 XM_005268839.1 XM_005268840.1 XM_005268841.1 XM_005268842.1 XM_005268844.1 XM_005268846.1 XM_005268848.1 XM_005268849.1 XM_005268850.1 XM_017019265.1 NM_152637.2

Gene Symbol
METTL7B
Entrez Gene ID
196410
Gene Name
methyltransferase like 7B
Gene Aliases
ALDI
UniGene
Hs.51483
Gene Chromosome Location
Chr.12: 55681546-55684611 on Build GRCh38
Transcripts
NM_001144996.1 NM_001144997.1 NM_002206.2 XM_005268839.1 XM_005268840.1 XM_005268841.1 XM_005268842.1 XM_005268844.1 XM_005268846.1 XM_005268848.1 XM_005268849.1 XM_005268850.1 XM_017019265.1 NM_152637.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5897043 c.718G>A p.G240R Chr.12: 55684032-55684032 on Build GRCh38
5375567 c.569G>A p.G190E Chr.12: 55684033-55684033 on Build GRCh38
5375566 c.719G>A p.G240E Chr.12: 55684033-55684033 on Build GRCh38
7193710 c.566delT p.M189fs*>6 Chr.12: 55684030-55684030 on Build GRCh38
5897044 c.568G>A p.G190R Chr.12: 55684032-55684032 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748137642 Chr.12: 55684411-55684411 on Build GRCh38
rs1000898878 Chr.12: 55684419-55684419 on Build GRCh38
rs939772650 Chr.12: 55684202-55684202 on Build GRCh38
rs993703344 Chr.12: 55684263-55684263 on Build GRCh38
rs1028167368 Chr.12: 55684519-55684519 on Build GRCh38
rs115447442 Chr.12: 55684379-55684379 on Build GRCh38
rs2293409 Chr.12: 55684180-55684180 on Build GRCh38
rs543416052 Chr.12: 55684436-55684436 on Build GRCh38
rs765230297 Chr.12: 55684094-55684094 on Build GRCh38
rs773499560 Chr.12: 55684050-55684050 on Build GRCh38
rs373374930 Chr.12: 55684119-55684119 on Build GRCh38
rs576413311 Chr.12: 55684427-55684427 on Build GRCh38
rs776815993 Chr.12: 55684067-55684067 on Build GRCh38
rs775103934 Chr.12: 55684045-55684045 on Build GRCh38
rs1055290648 Chr.12: 55684237-55684237 on Build GRCh38
rs556040535 Chr.12: 55684346-55684346 on Build GRCh38
rs1037702185 Chr.12: 55684466-55684466 on Build GRCh38
rs35563419 Chr.12: 55684271-55684272 on Build GRCh38
rs139946452 Chr.12: 55684364-55684364 on Build GRCh38
rs1038472531 Chr.12: 55684220-55684220 on Build GRCh38
rs571924384 Chr.12: 55684162-55684162 on Build GRCh38
rs957252041 Chr.12: 55684130-55684130 on Build GRCh38
rs577485987 Chr.12: 55684362-55684362 on Build GRCh38
rs372983213 Chr.12: 55684423-55684423 on Build GRCh38
rs770188094 Chr.12: 55684079-55684091 on Build GRCh38
rs143904434 Chr.12: 55684414-55684414 on Build GRCh38
rs898685500 Chr.12: 55684230-55684230 on Build GRCh38
rs369470172 Chr.12: 55684078-55684078 on Build GRCh38
rs964827029 Chr.12: 55684472-55684472 on Build GRCh38
rs986448145 Chr.12: 55684363-55684363 on Build GRCh38
rs550506831 Chr.12: 55684128-55684128 on Build GRCh38
rs954836851 Chr.12: 55684290-55684290 on Build GRCh38
rs532728267 Chr.12: 55684531-55684531 on Build GRCh38
rs1002350993 Chr.12: 55684231-55684231 on Build GRCh38
rs986964049 Chr.12: 55684313-55684313 on Build GRCh38
rs896782166 Chr.12: 55684256-55684256 on Build GRCh38
rs993714660 Chr.12: 55684510-55684510 on Build GRCh38
rs759797076 Chr.12: 55684077-55684077 on Build GRCh38
rs531325176 Chr.12: 55684120-55684120 on Build GRCh38
rs566249550 Chr.12: 55684199-55684199 on Build GRCh38
rs966791998 Chr.12: 55684288-55684288 on Build GRCh38
rs191835929 Chr.12: 55684333-55684333 on Build GRCh38
rs1001378070 Chr.12: 55684306-55684306 on Build GRCh38
rs12320702 Chr.12: 55684447-55684447 on Build GRCh38
rs907030832 Chr.12: 55684126-55684126 on Build GRCh38
rs914232346 Chr.12: 55684348-55684348 on Build GRCh38
rs374262365 Chr.12: 55684097-55684097 on Build GRCh38
rs975280851 Chr.12: 55684343-55684343 on Build GRCh38
rs938765673 Chr.12: 55684147-55684147 on Build GRCh38
rs866714037 Chr.12: 55684085-55684085 on Build GRCh38
rs964116113 Chr.12: 55684337-55684337 on Build GRCh38
rs1051239619 Chr.12: 55684177-55684177 on Build GRCh38
rs1042370340 Chr.12: 55684413-55684413 on Build GRCh38
rs748461175 Chr.12: 55684075-55684075 on Build GRCh38
rs182866415 Chr.12: 55684334-55684334 on Build GRCh38
rs548047474 Chr.12: 55684547-55684547 on Build GRCh38
rs939761175 Chr.12: 55684371-55684371 on Build GRCh38
rs996640398 Chr.12: 55684485-55684485 on Build GRCh38
rs897828960 Chr.12: 55684498-55684498 on Build GRCh38
rs1006201612 Chr.12: 55684182-55684182 on Build GRCh38
rs114876595 Chr.12: 55684273-55684273 on Build GRCh38
rs955472113 Chr.12: 55684347-55684347 on Build GRCh38
rs772649291 Chr.12: 55684046-55684046 on Build GRCh38
rs938179420 Chr.12: 55684401-55684401 on Build GRCh38
rs572205019 Chr.12: 55684387-55684387 on Build GRCh38
rs921213217 Chr.12: 55684170-55684170 on Build GRCh38
rs532764542 Chr.12: 55684171-55684171 on Build GRCh38
rs543011128 Chr.12: 55684407-55684407 on Build GRCh38
rs780325299 Chr.12: 55684116-55684116 on Build GRCh38
rs766542166 Chr.12: 55684065-55684065 on Build GRCh38
rs1048103 Chr.12: 55684405-55684405 on Build GRCh38
rs1055846926 Chr.12: 55684154-55684154 on Build GRCh38
rs754309520 Chr.12: 55684475-55684475 on Build GRCh38
rs536373912 Chr.12: 55684232-55684232 on Build GRCh38
rs967200092 Chr.12: 55684076-55684076 on Build GRCh38
rs1007749200 Chr.12: 55684546-55684546 on Build GRCh38
rs1013549376 Chr.12: 55684262-55684262 on Build GRCh38

View Full Product Details