Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 52921573-52922062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECHDC2
Entrez Gene ID
55268
Gene Name
enoyl-CoA hydratase domain containing 2
Gene Aliases
-
UniGene
Hs.476319
Gene Chromosome Location
Chr.1: 52895910-52927212 on Build GRCh38
Transcripts
NM_001198961.1 NM_001198962.1 NM_001319958.1 NM_018281.3 XM_011541709.2 XM_011541710.2 XM_011541711.2 XM_011541712.2 XM_011541715.2 XM_011541721.2 XM_011541723.2 XM_011541724.2 XM_011541725.2 XM_017001638.1 XM_017001641.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7107218 c.43C>A p.R15R Chr.1: 52921631-52921631 on Build GRCh38
7307354 c.97C>T p.R33C Chr.1: 52921577-52921577 on Build GRCh38
4852306 c.87G>A p.E29E Chr.1: 52921587-52921587 on Build GRCh38
6651348 c.37C>T p.P13S Chr.1: 52921637-52921637 on Build GRCh38
5827939 c.30delC p.W11fs*36 Chr.1: 52921644-52921644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015094171 Chr.1: 52921675-52921675 on Build GRCh38
rs759902557 Chr.1: 52921628-52921628 on Build GRCh38
rs866358771 Chr.1: 52921929-52921929 on Build GRCh38
rs189586978 Chr.1: 52921714-52921714 on Build GRCh38
rs771826453 Chr.1: 52921848-52921848 on Build GRCh38
rs184912946 Chr.1: 52921600-52921600 on Build GRCh38
rs954022311 Chr.1: 52921887-52921887 on Build GRCh38
rs905488752 Chr.1: 52921806-52921806 on Build GRCh38
rs778562647 Chr.1: 52921680-52921680 on Build GRCh38
rs746979160 Chr.1: 52921696-52921696 on Build GRCh38
rs775882035 Chr.1: 52921717-52921717 on Build GRCh38
rs146247680 Chr.1: 52921970-52921970 on Build GRCh38
rs537869643 Chr.1: 52921836-52921836 on Build GRCh38
rs765862095 Chr.1: 52921666-52921666 on Build GRCh38
rs748753422 Chr.1: 52921933-52921933 on Build GRCh38
rs1003278019 Chr.1: 52921634-52921634 on Build GRCh38
rs764412509 Chr.1: 52921718-52921718 on Build GRCh38
rs1015794664 Chr.1: 52921621-52921621 on Build GRCh38
rs986840860 Chr.1: 52921889-52921890 on Build GRCh38
rs368731634 Chr.1: 52921641-52921641 on Build GRCh38
rs939721016 Chr.1: 52921890-52921890 on Build GRCh38
rs776760601 Chr.1: 52921620-52921620 on Build GRCh38
rs772844017 Chr.1: 52921644-52921644 on Build GRCh38
rs1019005701 Chr.1: 52921982-52921982 on Build GRCh38
rs947297080 Chr.1: 52921800-52921800 on Build GRCh38
rs886780835 Chr.1: 52921876-52921876 on Build GRCh38
rs960827670 Chr.1: 52921616-52921616 on Build GRCh38
rs777603823 Chr.1: 52921692-52921692 on Build GRCh38
rs200668547 Chr.1: 52921636-52921636 on Build GRCh38
rs988065146 Chr.1: 52921775-52921775 on Build GRCh38
rs1018975993 Chr.1: 52921768-52921768 on Build GRCh38
rs915196059 Chr.1: 52921785-52921785 on Build GRCh38
rs200299177 Chr.1: 52921630-52921630 on Build GRCh38
rs995086459 Chr.1: 52921946-52921946 on Build GRCh38
rs963214844 Chr.1: 52921855-52921855 on Build GRCh38
rs781175129 Chr.1: 52921594-52921594 on Build GRCh38
rs753353382 Chr.1: 52921667-52921667 on Build GRCh38
rs776641879 Chr.1: 52921704-52921704 on Build GRCh38
rs745957302 Chr.1: 52921705-52921705 on Build GRCh38
rs752042052 Chr.1: 52921647-52921647 on Build GRCh38
rs768172492 Chr.1: 52921935-52921935 on Build GRCh38
rs914388773 Chr.1: 52921950-52921950 on Build GRCh38
rs574465490 Chr.1: 52921969-52921969 on Build GRCh38
rs1044724403 Chr.1: 52921801-52921801 on Build GRCh38
rs1056100239 Chr.1: 52921859-52921859 on Build GRCh38
rs770967138 Chr.1: 52921615-52921615 on Build GRCh38
rs181961067 Chr.1: 52921980-52921980 on Build GRCh38
rs540140070 Chr.1: 52921772-52921772 on Build GRCh38
rs754429424 Chr.1: 52921668-52921668 on Build GRCh38
rs770898476 Chr.1: 52921700-52921700 on Build GRCh38
rs934480064 Chr.1: 52921763-52921765 on Build GRCh38
rs995747401 Chr.1: 52921870-52921870 on Build GRCh38
rs757757299 Chr.1: 52921665-52921665 on Build GRCh38
rs886227762 Chr.1: 52921823-52921823 on Build GRCh38
rs141703368 Chr.1: 52921818-52921818 on Build GRCh38
rs987542904 Chr.1: 52921767-52921767 on Build GRCh38
rs953615752 Chr.1: 52921736-52921736 on Build GRCh38
rs556942931 Chr.1: 52921807-52921807 on Build GRCh38
rs7552139 Chr.1: 52921707-52921707 on Build GRCh38
rs571083734 Chr.1: 52921739-52921739 on Build GRCh38
rs138761671 Chr.1: 52921684-52921684 on Build GRCh38
rs758167745 Chr.1: 52921686-52921686 on Build GRCh38
rs116286929 Chr.1: 52921850-52921850 on Build GRCh38
rs763432611 Chr.1: 52921639-52921639 on Build GRCh38

View Full Product Details