Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157574660-157575151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRL4
Entrez Gene ID
83417
Gene Name
Fc receptor like 4
Gene Aliases
CD307d, FCRH4, IGFP2, IRTA1
UniGene
Hs.120260
Gene Chromosome Location
Chr.1: 157573749-157598080 on Build GRCh38
Transcripts
NM_031282.2 XM_011510034.1

SNP Detail

SNP ID Chromosome Location
rs919861272 Chr.1: 157575025-157575025 on Build GRCh38
rs967327820 Chr.1: 157574901-157574901 on Build GRCh38
rs928335680 Chr.1: 157575088-157575088 on Build GRCh38
rs568778227 Chr.1: 157574873-157574873 on Build GRCh38
rs906966477 Chr.1: 157574762-157574762 on Build GRCh38
rs938485621 Chr.1: 157575089-157575089 on Build GRCh38
rs1034837421 Chr.1: 157574921-157574921 on Build GRCh38
rs954818844 Chr.1: 157574920-157574920 on Build GRCh38
rs998207274 Chr.1: 157574811-157574811 on Build GRCh38
rs191313938 Chr.1: 157574859-157574859 on Build GRCh38
rs768371478 Chr.1: 157574912-157574912 on Build GRCh38
rs849829 Chr.1: 157574991-157574991 on Build GRCh38
rs748060849 Chr.1: 157575012-157575012 on Build GRCh38
rs1037155549 Chr.1: 157574753-157574753 on Build GRCh38
rs1038032456 Chr.1: 157574739-157574739 on Build GRCh38
rs149144976 Chr.1: 157574950-157574950 on Build GRCh38
rs575699734 Chr.1: 157574758-157574758 on Build GRCh38
rs536127914 Chr.1: 157574898-157574898 on Build GRCh38
rs144527505 Chr.1: 157574711-157574711 on Build GRCh38
rs758846992 Chr.1: 157575014-157575014 on Build GRCh38
rs14335 Chr.1: 157575060-157575060 on Build GRCh38
rs368927998 Chr.1: 157574699-157574699 on Build GRCh38
rs901296808 Chr.1: 157574786-157574786 on Build GRCh38
rs951804958 Chr.1: 157574969-157574969 on Build GRCh38
rs910160145 Chr.1: 157574998-157574998 on Build GRCh38
rs145600867 Chr.1: 157574720-157574720 on Build GRCh38
rs749101053 Chr.1: 157574892-157574892 on Build GRCh38
rs374917926 Chr.1: 157575078-157575078 on Build GRCh38
rs998879981 Chr.1: 157574902-157574902 on Build GRCh38
rs911089849 Chr.1: 157574962-157574962 on Build GRCh38
rs1049134107 Chr.1: 157574710-157574710 on Build GRCh38
rs1054492 Chr.1: 157574726-157574726 on Build GRCh38
rs924818920 Chr.1: 157574741-157574751 on Build GRCh38
rs562994276 Chr.1: 157574890-157574890 on Build GRCh38
rs573950795 Chr.1: 157574960-157574960 on Build GRCh38
rs1047230718 Chr.1: 157574761-157574761 on Build GRCh38
rs375310057 Chr.1: 157574879-157574879 on Build GRCh38
rs960466693 Chr.1: 157574931-157574931 on Build GRCh38
rs916302538 Chr.1: 157574955-157574955 on Build GRCh38

View Full Product Details