Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34280392-34280987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHCY
Entrez Gene ID
191
Gene Name
adenosylhomocysteinase
Gene Aliases
SAHH, adoHcyase
UniGene
Hs.388004
Gene Chromosome Location
Chr.20: 34235012-34311976 on Build GRCh38
Transcripts
NM_000687.3 NM_001161766.1 NM_001322084.1 NM_001322085.1 NM_001322086.1 XM_005260317.2 XM_011528656.2 XM_011528657.2 XM_011528658.2 XM_011528659.1 XM_017027708.1 XM_017027709.1 XM_017027710.1

SNP Detail

SNP ID Chromosome Location
rs1029827336 Chr.20: 34280696-34280696 on Build GRCh38
rs1054378702 Chr.20: 34280679-34280679 on Build GRCh38
rs927630595 Chr.20: 34280913-34280913 on Build GRCh38
rs1064500 Chr.20: 34280453-34280453 on Build GRCh38
rs1010039482 Chr.20: 34280695-34280695 on Build GRCh38
rs1058794 Chr.20: 34280463-34280463 on Build GRCh38
rs1058802 Chr.20: 34280456-34280456 on Build GRCh38
rs995010972 Chr.20: 34280860-34280860 on Build GRCh38
rs35627251 Chr.20: 34280800-34280801 on Build GRCh38
rs141075350 Chr.20: 34280908-34280908 on Build GRCh38
rs866102807 Chr.20: 34280511-34280511 on Build GRCh38
rs552401560 Chr.20: 34280662-34280694 on Build GRCh38
rs199872454 Chr.20: 34280460-34280460 on Build GRCh38
rs574438360 Chr.20: 34280607-34280607 on Build GRCh38
rs904613765 Chr.20: 34280484-34280484 on Build GRCh38
rs760409763 Chr.20: 34280650-34280650 on Build GRCh38
rs540672652 Chr.20: 34280890-34280890 on Build GRCh38
rs13245 Chr.20: 34280733-34280733 on Build GRCh38
rs948075691 Chr.20: 34280615-34280615 on Build GRCh38
rs12506 Chr.20: 34280448-34280448 on Build GRCh38
rs552690185 Chr.20: 34280451-34280451 on Build GRCh38
rs930156619 Chr.20: 34280415-34280415 on Build GRCh38
rs903851493 Chr.20: 34280672-34280672 on Build GRCh38
rs916493421 Chr.20: 34280961-34280961 on Build GRCh38
rs951637646 Chr.20: 34280861-34280861 on Build GRCh38
rs148140437 Chr.20: 34280862-34280862 on Build GRCh38
rs575932572 Chr.20: 34280746-34280746 on Build GRCh38
rs535554157 Chr.20: 34280691-34280691 on Build GRCh38
rs774376599 Chr.20: 34280866-34280866 on Build GRCh38
rs890428478 Chr.20: 34280697-34280697 on Build GRCh38
rs759056638 Chr.20: 34280853-34280853 on Build GRCh38
rs546058381 Chr.20: 34280826-34280826 on Build GRCh38
rs867868609 Chr.20: 34280682-34280682 on Build GRCh38
rs60011288 Chr.20: 34280625-34280625 on Build GRCh38
rs147081471 Chr.20: 34280846-34280846 on Build GRCh38
rs950453348 Chr.20: 34280888-34280888 on Build GRCh38
rs764824508 Chr.20: 34280933-34280933 on Build GRCh38
rs3208572 Chr.20: 34280455-34280455 on Build GRCh38
rs776423361 Chr.20: 34280718-34280718 on Build GRCh38
rs374087068 Chr.20: 34280648-34280648 on Build GRCh38
rs58365582 Chr.20: 34280431-34280431 on Build GRCh38
rs918009105 Chr.20: 34280877-34280877 on Build GRCh38
rs1043568711 Chr.20: 34280470-34280470 on Build GRCh38
rs998856710 Chr.20: 34280491-34280491 on Build GRCh38
rs14941 Chr.20: 34280417-34280417 on Build GRCh38
rs770573241 Chr.20: 34280700-34280700 on Build GRCh38
rs960383594 Chr.20: 34280917-34280917 on Build GRCh38

View Full Product Details