Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 190371090-190371642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP1
Entrez Gene ID
3628
Gene Name
inositol polyphosphate-1-phosphatase
Gene Aliases
-
UniGene
Hs.32309
Gene Chromosome Location
Chr.2: 190343470-190371665 on Build GRCh38
Transcripts
NM_001128928.1 NM_002194.3 XM_005246532.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3575582 c.1025C>T p.P342L Chr.2: 190371227-190371227 on Build GRCh38
6587228 c.1091A>T p.D364V Chr.2: 190371293-190371293 on Build GRCh38
1482569 c.1110A>G p.G370G Chr.2: 190371312-190371312 on Build GRCh38
1669607 c.904G>A p.V302I Chr.2: 190371106-190371106 on Build GRCh38
6686000 c.1139G>A p.R380Q Chr.2: 190371341-190371341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951601306 Chr.2: 190371158-190371158 on Build GRCh38
rs769062327 Chr.2: 190371264-190371264 on Build GRCh38
rs753480052 Chr.2: 190371288-190371288 on Build GRCh38
rs762369111 Chr.2: 190371421-190371421 on Build GRCh38
rs1018504160 Chr.2: 190371384-190371384 on Build GRCh38
rs765586863 Chr.2: 190371135-190371135 on Build GRCh38
rs775257302 Chr.2: 190371431-190371431 on Build GRCh38
rs143948960 Chr.2: 190371349-190371349 on Build GRCh38
rs572261182 Chr.2: 190371426-190371426 on Build GRCh38
rs1025133048 Chr.2: 190371618-190371618 on Build GRCh38
rs757398200 Chr.2: 190371346-190371346 on Build GRCh38
rs201808787 Chr.2: 190371222-190371222 on Build GRCh38
rs766982624 Chr.2: 190371153-190371153 on Build GRCh38
rs144177328 Chr.2: 190371296-190371296 on Build GRCh38
rs763474037 Chr.2: 190371257-190371257 on Build GRCh38
rs140741805 Chr.2: 190371213-190371213 on Build GRCh38
rs762624803 Chr.2: 190371445-190371445 on Build GRCh38
rs199548103 Chr.2: 190371392-190371392 on Build GRCh38
rs751671281 Chr.2: 190371454-190371454 on Build GRCh38
rs945835305 Chr.2: 190371342-190371342 on Build GRCh38
rs1030786859 Chr.2: 190371509-190371509 on Build GRCh38
rs1048644390 Chr.2: 190371549-190371549 on Build GRCh38
rs550439981 Chr.2: 190371460-190371460 on Build GRCh38
rs749963269 Chr.2: 190371154-190371154 on Build GRCh38
rs777831500 Chr.2: 190371340-190371340 on Build GRCh38
rs534906375 Chr.2: 190371131-190371131 on Build GRCh38
rs191271401 Chr.2: 190371495-190371495 on Build GRCh38
rs539053461 Chr.2: 190371525-190371525 on Build GRCh38
rs1882891 Chr.2: 190371175-190371175 on Build GRCh38
rs770136066 Chr.2: 190371239-190371239 on Build GRCh38
rs746018354 Chr.2: 190371367-190371367 on Build GRCh38
rs551503426 Chr.2: 190371540-190371540 on Build GRCh38
rs765018115 Chr.2: 190371295-190371295 on Build GRCh38
rs747108674 Chr.2: 190371208-190371208 on Build GRCh38
rs770789376 Chr.2: 190371242-190371242 on Build GRCh38
rs765944185 Chr.2: 190371274-190371274 on Build GRCh38
rs566450081 Chr.2: 190371589-190371589 on Build GRCh38
rs777871601 Chr.2: 190371204-190371204 on Build GRCh38
rs746149533 Chr.2: 190371225-190371225 on Build GRCh38
rs770373809 Chr.2: 190371423-190371423 on Build GRCh38
rs981106676 Chr.2: 190371223-190371223 on Build GRCh38
rs995772736 Chr.2: 190371537-190371537 on Build GRCh38
rs764820935 Chr.2: 190371428-190371428 on Build GRCh38
rs141851582 Chr.2: 190371341-190371341 on Build GRCh38
rs759127713 Chr.2: 190371291-190371291 on Build GRCh38
rs777464427 Chr.2: 190371173-190371174 on Build GRCh38
rs901032935 Chr.2: 190371393-190371393 on Build GRCh38
rs182469485 Chr.2: 190371588-190371588 on Build GRCh38
rs373745803 Chr.2: 190371453-190371453 on Build GRCh38
rs560029686 Chr.2: 190371345-190371345 on Build GRCh38
rs780414898 Chr.2: 190371402-190371402 on Build GRCh38
rs147389085 Chr.2: 190371404-190371404 on Build GRCh38
rs186409378 Chr.2: 190371424-190371424 on Build GRCh38
rs758307366 Chr.2: 190371310-190371310 on Build GRCh38
rs763382198 Chr.2: 190371146-190371146 on Build GRCh38
rs571825692 Chr.2: 190371327-190371327 on Build GRCh38
rs775483833 Chr.2: 190371358-190371358 on Build GRCh38
rs369912593 Chr.2: 190371142-190371142 on Build GRCh38
rs867566856 Chr.2: 190371377-190371377 on Build GRCh38
rs752624961 Chr.2: 190371182-190371182 on Build GRCh38
rs778634689 Chr.2: 190371177-190371177 on Build GRCh38
rs1036482224 Chr.2: 190371237-190371237 on Build GRCh38
rs754767057 Chr.2: 190371176-190371176 on Build GRCh38
rs749204510 Chr.2: 190371237-190371238 on Build GRCh38
rs774839534 Chr.2: 190371418-190371418 on Build GRCh38
rs138606587 Chr.2: 190371252-190371252 on Build GRCh38
rs891240691 Chr.2: 190371382-190371382 on Build GRCh38
rs760267626 Chr.2: 190371167-190371167 on Build GRCh38
rs781587537 Chr.2: 190371609-190371609 on Build GRCh38
rs376339364 Chr.2: 190371417-190371417 on Build GRCh38
rs35616200 Chr.2: 190371265-190371265 on Build GRCh38
rs758274194 Chr.2: 190371187-190371187 on Build GRCh38

View Full Product Details