Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 85621381-85621936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLRT2
Entrez Gene ID
23768
Gene Name
fibronectin leucine rich transmembrane protein 2
Gene Aliases
-
UniGene
Hs.533710
Gene Chromosome Location
Chr.14: 85530067-85628696 on Build GRCh38
Transcripts
NM_013231.4 XM_005267490.3 XM_011536611.2 XM_017021129.1 XM_017021130.1 XM_017021131.1 XM_017021132.1 XM_017021133.1 XM_017021134.1 XM_017021135.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3377475 c.121C>T p.R41C Chr.14: 85621635-85621635 on Build GRCh38
4052745 c.122G>A p.R41H Chr.14: 85621636-85621636 on Build GRCh38
366158 c.145T>A p.C49S Chr.14: 85621659-85621659 on Build GRCh38
4427112 c.3G>C p.M1I Chr.14: 85621517-85621517 on Build GRCh38
1252576 c.397T>C p.L133L Chr.14: 85621911-85621911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749768193 Chr.14: 85621778-85621778 on Build GRCh38
rs1023697101 Chr.14: 85621547-85621547 on Build GRCh38
rs142516343 Chr.14: 85621911-85621911 on Build GRCh38
rs777030016 Chr.14: 85621652-85621652 on Build GRCh38
rs754401739 Chr.14: 85621502-85621502 on Build GRCh38
rs936934530 Chr.14: 85621458-85621458 on Build GRCh38
rs757913785 Chr.14: 85621735-85621735 on Build GRCh38
rs201103211 Chr.14: 85621775-85621775 on Build GRCh38
rs773181366 Chr.14: 85621771-85621771 on Build GRCh38
rs758669921 Chr.14: 85621479-85621479 on Build GRCh38
rs10141266 Chr.14: 85621712-85621712 on Build GRCh38
rs759315914 Chr.14: 85621794-85621794 on Build GRCh38
rs766170020 Chr.14: 85621636-85621636 on Build GRCh38
rs759475732 Chr.14: 85621473-85621473 on Build GRCh38
rs752639321 Chr.14: 85621808-85621808 on Build GRCh38
rs765183184 Chr.14: 85621604-85621604 on Build GRCh38
rs188407245 Chr.14: 85621484-85621484 on Build GRCh38
rs375543804 Chr.14: 85621457-85621458 on Build GRCh38
rs558564181 Chr.14: 85621504-85621504 on Build GRCh38
rs773064350 Chr.14: 85621554-85621554 on Build GRCh38
rs769115922 Chr.14: 85621565-85621565 on Build GRCh38
rs751084522 Chr.14: 85621814-85621814 on Build GRCh38
rs764945420 Chr.14: 85621802-85621802 on Build GRCh38
rs774494135 Chr.14: 85621706-85621706 on Build GRCh38
rs749697862 Chr.14: 85621894-85621894 on Build GRCh38
rs752589389 Chr.14: 85621720-85621720 on Build GRCh38
rs755973823 Chr.14: 85621488-85621488 on Build GRCh38
rs773361874 Chr.14: 85621790-85621790 on Build GRCh38
rs796880664 Chr.14: 85621457-85621458 on Build GRCh38
rs746240615 Chr.14: 85621769-85621769 on Build GRCh38
rs777455551 Chr.14: 85621736-85621736 on Build GRCh38
rs768687035 Chr.14: 85621780-85621780 on Build GRCh38
rs1014006891 Chr.14: 85621539-85621539 on Build GRCh38
rs757901958 Chr.14: 85621663-85621663 on Build GRCh38
rs777334566 Chr.14: 85621468-85621468 on Build GRCh38
rs1044081460 Chr.14: 85621444-85621444 on Build GRCh38
rs756949238 Chr.14: 85621750-85621750 on Build GRCh38
rs187058999 Chr.14: 85621827-85621827 on Build GRCh38
rs573729441 Chr.14: 85621416-85621416 on Build GRCh38
rs757225269 Chr.14: 85621491-85621491 on Build GRCh38
rs761918713 Chr.14: 85621784-85621784 on Build GRCh38
rs375392583 Chr.14: 85621492-85621492 on Build GRCh38
rs576805931 Chr.14: 85621700-85621700 on Build GRCh38
rs982960098 Chr.14: 85621621-85621621 on Build GRCh38
rs754917817 Chr.14: 85621641-85621641 on Build GRCh38
rs1042941543 Chr.14: 85621455-85621455 on Build GRCh38
rs117507441 Chr.14: 85621833-85621833 on Build GRCh38
rs746663082 Chr.14: 85621668-85621668 on Build GRCh38
rs762985355 Chr.14: 85621616-85621616 on Build GRCh38
rs771381338 Chr.14: 85621671-85621671 on Build GRCh38
rs747657938 Chr.14: 85621533-85621533 on Build GRCh38
rs77099788 Chr.14: 85621781-85621781 on Build GRCh38
rs746268081 Chr.14: 85621684-85621684 on Build GRCh38
rs369272849 Chr.14: 85621640-85621640 on Build GRCh38
rs770305443 Chr.14: 85621688-85621688 on Build GRCh38
rs199539989 Chr.14: 85621699-85621699 on Build GRCh38
rs200776611 Chr.14: 85621707-85621707 on Build GRCh38
rs779957284 Chr.14: 85621491-85621491 on Build GRCh38
rs5810265 Chr.14: 85621457-85621457 on Build GRCh38
rs917172092 Chr.14: 85621748-85621748 on Build GRCh38
rs142772729 Chr.14: 85621713-85621713 on Build GRCh38
rs201873570 Chr.14: 85621744-85621744 on Build GRCh38
rs146660099 Chr.14: 85621661-85621661 on Build GRCh38
rs755849167 Chr.14: 85621629-85621629 on Build GRCh38
rs868026714 Chr.14: 85621551-85621551 on Build GRCh38
rs374855465 Chr.14: 85621624-85621624 on Build GRCh38
rs747435487 Chr.14: 85621486-85621486 on Build GRCh38
rs905448199 Chr.14: 85621456-85621457 on Build GRCh38
rs777211965 Chr.14: 85621596-85621596 on Build GRCh38
rs778504472 Chr.14: 85621503-85621503 on Build GRCh38
rs143193106 Chr.14: 85621422-85621422 on Build GRCh38
rs371043550 Chr.14: 85621810-85621810 on Build GRCh38
rs563943260 Chr.14: 85621478-85621478 on Build GRCh38
rs778764956 Chr.14: 85621915-85621915 on Build GRCh38
rs951622502 Chr.14: 85621590-85621590 on Build GRCh38
rs764176554 Chr.14: 85621811-85621811 on Build GRCh38
rs778753174 Chr.14: 85621644-85621644 on Build GRCh38
rs144800875 Chr.14: 85621628-85621628 on Build GRCh38
rs1048468828 Chr.14: 85621796-85621796 on Build GRCh38
rs779460062 Chr.14: 85621909-85621909 on Build GRCh38
rs765363486 Chr.14: 85621474-85621474 on Build GRCh38
rs756896260 Chr.14: 85621817-85621817 on Build GRCh38
rs149395539 Chr.14: 85621582-85621582 on Build GRCh38
rs749242316 Chr.14: 85621493-85621493 on Build GRCh38
rs755442700 Chr.14: 85621899-85621899 on Build GRCh38
rs780318340 Chr.14: 85621891-85621891 on Build GRCh38
rs771434450 Chr.14: 85621588-85621588 on Build GRCh38
rs777508524 Chr.14: 85621664-85621664 on Build GRCh38
rs934605825 Chr.14: 85621458-85621458 on Build GRCh38
rs554986569 Chr.14: 85621882-85621882 on Build GRCh38
rs182285274 Chr.14: 85621758-85621758 on Build GRCh38
rs771722069 Chr.14: 85621538-85621538 on Build GRCh38
rs775930423 Chr.14: 85621691-85621691 on Build GRCh38
rs903085914 Chr.14: 85621457-85621457 on Build GRCh38
rs772274794 Chr.14: 85621789-85621789 on Build GRCh38
rs760120757 Chr.14: 85621603-85621603 on Build GRCh38
rs775619302 Chr.14: 85621613-85621613 on Build GRCh38
rs947215229 Chr.14: 85621421-85621421 on Build GRCh38
rs372473725 Chr.14: 85621759-85621759 on Build GRCh38
rs901280957 Chr.14: 85621645-85621645 on Build GRCh38
rs781707035 Chr.14: 85621677-85621677 on Build GRCh38
rs368225812 Chr.14: 85621755-85621755 on Build GRCh38

View Full Product Details