Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 32334427-32334966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf75
Entrez Gene ID
64149
Gene Name
chromosome 17 open reading frame 75
Gene Aliases
NJMU-R1, SRI2
UniGene
Hs.655257
Gene Chromosome Location
Chr.17: 32328441-32350047 on Build GRCh38
Transcripts
NM_022344.3 XM_005258022.3 XM_017024940.1 XM_017024941.1

SNP Detail

SNP ID Chromosome Location
rs767243043 Chr.17: 32334570-32334570 on Build GRCh38
rs779766504 Chr.17: 32334592-32334592 on Build GRCh38
rs887049211 Chr.17: 32334545-32334545 on Build GRCh38
rs190434891 Chr.17: 32334503-32334503 on Build GRCh38
rs746547031 Chr.17: 32334758-32334760 on Build GRCh38
rs541937909 Chr.17: 32334756-32334756 on Build GRCh38
rs1000211226 Chr.17: 32334483-32334483 on Build GRCh38
rs763673 Chr.17: 32334903-32334903 on Build GRCh38
rs1127742 Chr.17: 32334591-32334591 on Build GRCh38
rs757482204 Chr.17: 32334788-32334788 on Build GRCh38
rs1000411272 Chr.17: 32334911-32334911 on Build GRCh38
rs778502688 Chr.17: 32334604-32334604 on Build GRCh38
rs764124264 Chr.17: 32334759-32334759 on Build GRCh38
rs747676283 Chr.17: 32334613-32334613 on Build GRCh38
rs746251152 Chr.17: 32334872-32334872 on Build GRCh38
rs780965069 Chr.17: 32334850-32334851 on Build GRCh38
rs752776532 Chr.17: 32334528-32334528 on Build GRCh38
rs1028107653 Chr.17: 32334711-32334711 on Build GRCh38
rs780093269 Chr.17: 32334757-32334758 on Build GRCh38
rs904750368 Chr.17: 32334478-32334478 on Build GRCh38
rs766693751 Chr.17: 32334500-32334500 on Build GRCh38
rs528193564 Chr.17: 32334673-32334673 on Build GRCh38
rs1010936466 Chr.17: 32334456-32334456 on Build GRCh38
rs773392379 Chr.17: 32334482-32334482 on Build GRCh38
rs907196978 Chr.17: 32334488-32334488 on Build GRCh38
rs140702394 Chr.17: 32334650-32334650 on Build GRCh38
rs200381289 Chr.17: 32334825-32334825 on Build GRCh38
rs1017151874 Chr.17: 32334562-32334562 on Build GRCh38
rs754907079 Chr.17: 32334800-32334800 on Build GRCh38
rs771802023 Chr.17: 32334621-32334621 on Build GRCh38
rs749227675 Chr.17: 32334454-32334454 on Build GRCh38
rs776925626 Chr.17: 32334731-32334731 on Build GRCh38
rs955009762 Chr.17: 32334719-32334719 on Build GRCh38
rs372303554 Chr.17: 32334830-32334830 on Build GRCh38
rs564377436 Chr.17: 32334919-32334919 on Build GRCh38
rs983758326 Chr.17: 32334724-32334724 on Build GRCh38
rs914688025 Chr.17: 32334837-32334837 on Build GRCh38
rs115007276 Chr.17: 32334866-32334866 on Build GRCh38
rs369049457 Chr.17: 32334880-32334880 on Build GRCh38
rs928765140 Chr.17: 32334918-32334918 on Build GRCh38
rs758443850 Chr.17: 32334636-32334637 on Build GRCh38
rs775496021 Chr.17: 32334740-32334740 on Build GRCh38
rs980589230 Chr.17: 32334873-32334873 on Build GRCh38
rs751787545 Chr.17: 32334784-32334784 on Build GRCh38
rs1034318460 Chr.17: 32334495-32334495 on Build GRCh38
rs746667576 Chr.17: 32334843-32334843 on Build GRCh38
rs777205617 Chr.17: 32334842-32334842 on Build GRCh38
rs764395826 Chr.17: 32334543-32334543 on Build GRCh38
rs774520638 Chr.17: 32334945-32334945 on Build GRCh38
rs761554687 Chr.17: 32334462-32334462 on Build GRCh38
rs770493008 Chr.17: 32334859-32334859 on Build GRCh38
rs868423148 Chr.17: 32334668-32334668 on Build GRCh38
rs753691500 Chr.17: 32334791-32334791 on Build GRCh38
rs1007919069 Chr.17: 32334729-32334729 on Build GRCh38
rs774065743 Chr.17: 32334458-32334458 on Build GRCh38
rs779016175 Chr.17: 32334821-32334821 on Build GRCh38
rs758555466 Chr.17: 32334530-32334530 on Build GRCh38
rs368181633 Chr.17: 32334505-32334505 on Build GRCh38
rs1040787844 Chr.17: 32334769-32334769 on Build GRCh38
rs768262036 Chr.17: 32334457-32334457 on Build GRCh38
rs746689187 Chr.17: 32334649-32334649 on Build GRCh38
rs759966846 Chr.17: 32334733-32334733 on Build GRCh38
rs558393012 Chr.17: 32334492-32334492 on Build GRCh38
rs182533138 Chr.17: 32334548-32334548 on Build GRCh38
rs749239338 Chr.17: 32334596-32334596 on Build GRCh38
rs185717321 Chr.17: 32334465-32334465 on Build GRCh38
rs765295011 Chr.17: 32334506-32334506 on Build GRCh38
rs952600945 Chr.17: 32334659-32334659 on Build GRCh38
rs750423786 Chr.17: 32334609-32334610 on Build GRCh38
rs765745101 Chr.17: 32334737-32334737 on Build GRCh38
rs767987268 Chr.17: 32334788-32334791 on Build GRCh38
rs576241181 Chr.17: 32334657-32334657 on Build GRCh38
rs375251296 Chr.17: 32334636-32334636 on Build GRCh38
rs1056298082 Chr.17: 32334527-32334527 on Build GRCh38
rs565301058 Chr.17: 32334512-32334512 on Build GRCh38

View Full Product Details