Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 12344017-12344373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFG3L2
Entrez Gene ID
10939
Gene Name
AFG3 like matrix AAA peptidase subunit 2
Gene Aliases
SCA28, SPAX5
UniGene
Hs.726355
Gene Chromosome Location
Chr.18: 12328944-12377309 on Build GRCh38
Transcripts
NM_006796.2 XM_011525601.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
986609 c.1738G>A p.V580I Chr.18: 12344173-12344173 on Build GRCh38
158948 c.1679C>T p.T560M Chr.18: 12344232-12344232 on Build GRCh38
1387800 c.1680G>A p.T560T Chr.18: 12344231-12344231 on Build GRCh38
708436 c.1729G>T p.G577C Chr.18: 12344182-12344182 on Build GRCh38
6411476 c.1690C>T p.Q564* Chr.18: 12344221-12344221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760805375 Chr.18: 12344079-12344079 on Build GRCh38
rs368238350 Chr.18: 12344294-12344294 on Build GRCh38
rs56213968 Chr.18: 12344153-12344153 on Build GRCh38
rs763265106 Chr.18: 12344232-12344232 on Build GRCh38
rs777685334 Chr.18: 12344116-12344116 on Build GRCh38
rs989332053 Chr.18: 12344317-12344317 on Build GRCh38
rs771985368 Chr.18: 12344069-12344069 on Build GRCh38
rs551042055 Chr.18: 12344175-12344175 on Build GRCh38
rs765728151 Chr.18: 12344258-12344260 on Build GRCh38
rs753148089 Chr.18: 12344199-12344199 on Build GRCh38
rs903701615 Chr.18: 12344324-12344324 on Build GRCh38
rs766925748 Chr.18: 12344091-12344091 on Build GRCh38
rs755696563 Chr.18: 12344100-12344100 on Build GRCh38
rs189177049 Chr.18: 12344280-12344280 on Build GRCh38
rs918239084 Chr.18: 12344271-12344271 on Build GRCh38
rs747162830 Chr.18: 12344122-12344122 on Build GRCh38
rs75206140 Chr.18: 12344286-12344286 on Build GRCh38
rs138460154 Chr.18: 12344109-12344109 on Build GRCh38
rs552006318 Chr.18: 12344227-12344227 on Build GRCh38
rs779521871 Chr.18: 12344060-12344060 on Build GRCh38
rs750347192 Chr.18: 12344168-12344168 on Build GRCh38
rs758811358 Chr.18: 12344205-12344205 on Build GRCh38
rs201510736 Chr.18: 12344210-12344210 on Build GRCh38
rs144122718 Chr.18: 12344189-12344189 on Build GRCh38
rs562861748 Chr.18: 12344197-12344197 on Build GRCh38
rs760219294 Chr.18: 12344173-12344173 on Build GRCh38
rs769160815 Chr.18: 12344052-12344052 on Build GRCh38
rs567049273 Chr.18: 12344315-12344315 on Build GRCh38
rs1000270201 Chr.18: 12344328-12344328 on Build GRCh38
rs771790775 Chr.18: 12344281-12344281 on Build GRCh38
rs370421677 Chr.18: 12344114-12344114 on Build GRCh38
rs200476229 Chr.18: 12344256-12344256 on Build GRCh38
rs746302967 Chr.18: 12344141-12344141 on Build GRCh38
rs781266510 Chr.18: 12344231-12344231 on Build GRCh38
rs540391344 Chr.18: 12344080-12344080 on Build GRCh38
rs1056381461 Chr.18: 12344282-12344282 on Build GRCh38
rs1012169446 Chr.18: 12344313-12344313 on Build GRCh38
rs199706026 Chr.18: 12344087-12344087 on Build GRCh38
rs771385439 Chr.18: 12344086-12344086 on Build GRCh38
rs750805669 Chr.18: 12344263-12344264 on Build GRCh38
rs111394518 Chr.18: 12344068-12344068 on Build GRCh38
rs1024713723 Chr.18: 12344290-12344291 on Build GRCh38
rs374828650 Chr.18: 12344149-12344149 on Build GRCh38
rs765992493 Chr.18: 12344174-12344174 on Build GRCh38
rs367992548 Chr.18: 12344150-12344150 on Build GRCh38
rs371687745 Chr.18: 12344094-12344094 on Build GRCh38
rs748693053 Chr.18: 12344063-12344063 on Build GRCh38
rs747241831 Chr.18: 12344271-12344271 on Build GRCh38
rs746080788 Chr.18: 12344249-12344249 on Build GRCh38
rs761827264 Chr.18: 12344088-12344088 on Build GRCh38
rs773993162 Chr.18: 12344273-12344273 on Build GRCh38
rs759762398 Chr.18: 12344144-12344144 on Build GRCh38
rs1044897718 Chr.18: 12344314-12344314 on Build GRCh38
rs775244310 Chr.18: 12344082-12344082 on Build GRCh38
rs748244264 Chr.18: 12344070-12344070 on Build GRCh38
rs932744770 Chr.18: 12344118-12344118 on Build GRCh38
rs780309456 Chr.18: 12344270-12344271 on Build GRCh38
rs977894057 Chr.18: 12344307-12344307 on Build GRCh38
rs376915348 Chr.18: 12344110-12344110 on Build GRCh38
rs886053615 Chr.18: 12344252-12344252 on Build GRCh38
rs748587965 Chr.18: 12344113-12344113 on Build GRCh38
rs762687153 Chr.18: 12344083-12344083 on Build GRCh38
rs139192793 Chr.18: 12344146-12344146 on Build GRCh38
rs755123459 Chr.18: 12344273-12344274 on Build GRCh38
rs767536465 Chr.18: 12344155-12344155 on Build GRCh38
rs897402798 Chr.18: 12344222-12344222 on Build GRCh38
rs779432036 Chr.18: 12344112-12344112 on Build GRCh38

View Full Product Details