Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 47154839-47155416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IER3IP1
Entrez Gene ID
51124
Gene Name
immediate early response 3 interacting protein 1
Gene Aliases
HSPC039, MEDS, PRO2309
UniGene
Hs.130917
Gene Chromosome Location
Chr.18: 47155019-47176374 on Build GRCh38
Transcripts
NM_016097.4

SNP Detail

SNP ID Chromosome Location
rs534373290 Chr.18: 47155238-47155238 on Build GRCh38
rs977682477 Chr.18: 47154894-47154894 on Build GRCh38
rs9831 Chr.18: 47155114-47155114 on Build GRCh38
rs866099757 Chr.18: 47155040-47155040 on Build GRCh38
rs1039810735 Chr.18: 47155102-47155102 on Build GRCh38
rs796234358 Chr.18: 47155283-47155283 on Build GRCh38
rs901831163 Chr.18: 47154990-47154990 on Build GRCh38
rs943563117 Chr.18: 47155234-47155234 on Build GRCh38
rs540392061 Chr.18: 47155354-47155354 on Build GRCh38
rs993929957 Chr.18: 47154998-47154998 on Build GRCh38
rs778156562 Chr.18: 47155082-47155082 on Build GRCh38
rs141766622 Chr.18: 47155377-47155377 on Build GRCh38
rs148061497 Chr.18: 47155253-47155253 on Build GRCh38
rs977759852 Chr.18: 47155152-47155152 on Build GRCh38
rs544324119 Chr.18: 47155254-47155254 on Build GRCh38
rs887730102 Chr.18: 47155042-47155042 on Build GRCh38
rs577928958 Chr.18: 47155288-47155288 on Build GRCh38
rs976175269 Chr.18: 47155246-47155246 on Build GRCh38
rs966805981 Chr.18: 47154870-47154870 on Build GRCh38
rs763445080 Chr.18: 47154926-47154931 on Build GRCh38
rs933759744 Chr.18: 47155149-47155149 on Build GRCh38
rs14524 Chr.18: 47155174-47155174 on Build GRCh38
rs924902521 Chr.18: 47154916-47154916 on Build GRCh38
rs946385183 Chr.18: 47155100-47155100 on Build GRCh38
rs936161868 Chr.18: 47154938-47154938 on Build GRCh38
rs569156756 Chr.18: 47154979-47154979 on Build GRCh38
rs1023415511 Chr.18: 47155276-47155276 on Build GRCh38
rs535107293 Chr.18: 47155248-47155248 on Build GRCh38
rs1017200504 Chr.18: 47155222-47155222 on Build GRCh38
rs187175176 Chr.18: 47155372-47155372 on Build GRCh38
rs564142124 Chr.18: 47155271-47155271 on Build GRCh38
rs370755992 Chr.18: 47155196-47155196 on Build GRCh38
rs115994926 Chr.18: 47155020-47155020 on Build GRCh38
rs183382447 Chr.18: 47155251-47155251 on Build GRCh38
rs1055062860 Chr.18: 47155368-47155368 on Build GRCh38
rs981952550 Chr.18: 47155007-47155007 on Build GRCh38
rs935221589 Chr.18: 47155047-47155047 on Build GRCh38
rs779675989 Chr.18: 47155226-47155227 on Build GRCh38
rs991942883 Chr.18: 47154963-47154963 on Build GRCh38
rs1040871867 Chr.18: 47154988-47154988 on Build GRCh38
rs1025167120 Chr.18: 47155127-47155127 on Build GRCh38
rs577258223 Chr.18: 47155264-47155264 on Build GRCh38
rs879613514 Chr.18: 47155008-47155008 on Build GRCh38
rs553276861 Chr.18: 47155115-47155115 on Build GRCh38
rs1016915847 Chr.18: 47154909-47154909 on Build GRCh38
rs571532485 Chr.18: 47155052-47155052 on Build GRCh38
rs941904223 Chr.18: 47155356-47155356 on Build GRCh38
rs966398173 Chr.18: 47155150-47155150 on Build GRCh38
rs1013874799 Chr.18: 47155374-47155374 on Build GRCh38
rs2251948 Chr.18: 47154985-47154985 on Build GRCh38
rs1051250438 Chr.18: 47155173-47155173 on Build GRCh38
rs191518284 Chr.18: 47155179-47155179 on Build GRCh38
rs1031975679 Chr.18: 47155159-47155159 on Build GRCh38
rs962710945 Chr.18: 47154948-47154948 on Build GRCh38
rs887782500 Chr.18: 47155320-47155320 on Build GRCh38

View Full Product Details