Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15055828-15056420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP14
Entrez Gene ID
23581
Gene Name
caspase 14
Gene Aliases
-
UniGene
Hs.466057
Gene Chromosome Location
Chr.19: 15049480-15058293 on Build GRCh38
Transcripts
NM_012114.2 XM_011527861.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4518390 c.630_631CC>TT p.R211W Chr.19: 15055990-15055991 on Build GRCh38
3282801 c.634C>T p.R212W Chr.19: 15055994-15055994 on Build GRCh38
5388849 c.655G>A p.V219I Chr.19: 15056015-15056015 on Build GRCh38
1582392 c.707G>A p.R236Q Chr.19: 15056067-15056067 on Build GRCh38
3529504 c.684C>T p.N228N Chr.19: 15056044-15056044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559517962 Chr.19: 15056221-15056221 on Build GRCh38
rs200722004 Chr.19: 15055990-15055990 on Build GRCh38
rs142933956 Chr.19: 15055875-15055875 on Build GRCh38
rs144959721 Chr.19: 15056028-15056028 on Build GRCh38
rs1044215937 Chr.19: 15055925-15055925 on Build GRCh38
rs571270904 Chr.19: 15056318-15056318 on Build GRCh38
rs773375209 Chr.19: 15056101-15056101 on Build GRCh38
rs1046138551 Chr.19: 15056063-15056063 on Build GRCh38
rs1030524257 Chr.19: 15055995-15055995 on Build GRCh38
rs199893602 Chr.19: 15055992-15055992 on Build GRCh38
rs1018201640 Chr.19: 15056174-15056174 on Build GRCh38
rs200139686 Chr.19: 15055994-15055994 on Build GRCh38
rs11574686 Chr.19: 15056277-15056277 on Build GRCh38
rs954851715 Chr.19: 15056250-15056250 on Build GRCh38
rs1030416348 Chr.19: 15056111-15056111 on Build GRCh38
rs577527483 Chr.19: 15055879-15055879 on Build GRCh38
rs717309 Chr.19: 15055867-15055867 on Build GRCh38
rs142912238 Chr.19: 15056125-15056128 on Build GRCh38
rs562909688 Chr.19: 15056072-15056072 on Build GRCh38
rs371772165 Chr.19: 15055975-15055975 on Build GRCh38
rs566839877 Chr.19: 15056362-15056362 on Build GRCh38
rs745589008 Chr.19: 15055998-15055998 on Build GRCh38
rs1038685003 Chr.19: 15055893-15055893 on Build GRCh38
rs574720596 Chr.19: 15055859-15055859 on Build GRCh38
rs906331879 Chr.19: 15056065-15056065 on Build GRCh38
rs746651266 Chr.19: 15055991-15055991 on Build GRCh38
rs749353663 Chr.19: 15056012-15056012 on Build GRCh38
rs900729270 Chr.19: 15056215-15056215 on Build GRCh38
rs938481891 Chr.19: 15056345-15056345 on Build GRCh38
rs775445646 Chr.19: 15056007-15056007 on Build GRCh38
rs61637857 Chr.19: 15056317-15056317 on Build GRCh38
rs755895168 Chr.19: 15056073-15056073 on Build GRCh38
rs1021305357 Chr.19: 15056198-15056198 on Build GRCh38
rs993529082 Chr.19: 15056099-15056099 on Build GRCh38
rs117721788 Chr.19: 15055882-15055882 on Build GRCh38
rs954882652 Chr.19: 15055996-15055996 on Build GRCh38
rs534194955 Chr.19: 15056389-15056389 on Build GRCh38
rs61750056 Chr.19: 15056041-15056041 on Build GRCh38
rs979973795 Chr.19: 15056342-15056342 on Build GRCh38
rs891513106 Chr.19: 15056351-15056351 on Build GRCh38
rs202048131 Chr.19: 15056093-15056093 on Build GRCh38
rs927704903 Chr.19: 15056234-15056234 on Build GRCh38
rs11574685 Chr.19: 15056186-15056186 on Build GRCh38
rs954797166 Chr.19: 15056149-15056149 on Build GRCh38
rs990141084 Chr.19: 15056202-15056202 on Build GRCh38
rs550959298 Chr.19: 15056348-15056348 on Build GRCh38
rs756611657 Chr.19: 15055935-15055935 on Build GRCh38
rs986283786 Chr.19: 15056153-15056153 on Build GRCh38
rs958455197 Chr.19: 15056361-15056361 on Build GRCh38
rs202225537 Chr.19: 15056087-15056087 on Build GRCh38
rs1015383092 Chr.19: 15056299-15056299 on Build GRCh38
rs768754892 Chr.19: 15056016-15056016 on Build GRCh38
rs773573153 Chr.19: 15056035-15056035 on Build GRCh38
rs149050628 Chr.19: 15056040-15056040 on Build GRCh38
rs369529451 Chr.19: 15055934-15055934 on Build GRCh38
rs936946545 Chr.19: 15056027-15056027 on Build GRCh38
rs151103624 Chr.19: 15055878-15055878 on Build GRCh38
rs73518121 Chr.19: 15055951-15055951 on Build GRCh38
rs769352959 Chr.19: 15055900-15055900 on Build GRCh38
rs763645642 Chr.19: 15056061-15056061 on Build GRCh38
rs143941999 Chr.19: 15056325-15056325 on Build GRCh38
rs780931393 Chr.19: 15056068-15056068 on Build GRCh38
rs898451494 Chr.19: 15055911-15055911 on Build GRCh38
rs752164297 Chr.19: 15055953-15055953 on Build GRCh38
rs760972661 Chr.19: 15056039-15056039 on Build GRCh38
rs141434967 Chr.19: 15055915-15055915 on Build GRCh38
rs757895582 Chr.19: 15056054-15056054 on Build GRCh38
rs947176342 Chr.19: 15056357-15056357 on Build GRCh38
rs202004752 Chr.19: 15056089-15056089 on Build GRCh38
rs531445692 Chr.19: 15056308-15056308 on Build GRCh38
rs1006209850 Chr.19: 15055921-15055921 on Build GRCh38
rs761060346 Chr.19: 15056105-15056105 on Build GRCh38
rs376759468 Chr.19: 15056001-15056001 on Build GRCh38
rs937776758 Chr.19: 15056262-15056262 on Build GRCh38
rs1030617523 Chr.19: 15056236-15056236 on Build GRCh38
rs555545375 Chr.19: 15056394-15056394 on Build GRCh38
rs141111994 Chr.19: 15056079-15056079 on Build GRCh38
rs11574687 Chr.19: 15056336-15056336 on Build GRCh38
rs1043222952 Chr.19: 15056385-15056385 on Build GRCh38
rs1016693505 Chr.19: 15055933-15055933 on Build GRCh38
rs928587858 Chr.19: 15056343-15056343 on Build GRCh38
rs906931345 Chr.19: 15055930-15055930 on Build GRCh38
rs981767065 Chr.19: 15056232-15056232 on Build GRCh38
rs147759883 Chr.19: 15056067-15056067 on Build GRCh38
rs757937137 Chr.19: 15056350-15056350 on Build GRCh38
rs774489166 Chr.19: 15056022-15056022 on Build GRCh38
rs751321912 Chr.19: 15056066-15056066 on Build GRCh38
rs766735171 Chr.19: 15056119-15056119 on Build GRCh38
rs767938581 Chr.19: 15056033-15056033 on Build GRCh38
rs111682561 Chr.19: 15056177-15056177 on Build GRCh38
rs181347483 Chr.19: 15055916-15055916 on Build GRCh38

View Full Product Details