Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49069902-49070455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNA7
Entrez Gene ID
3743
Gene Name
potassium voltage-gated channel subfamily A member 7
Gene Aliases
HAK6, KV1.7
UniGene
Hs.306973
Gene Chromosome Location
Chr.19: 49067418-49072941 on Build GRCh38
Transcripts
NM_031886.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3971141 c.1175T>C p.V392A Chr.19: 49070259-49070259 on Build GRCh38
4984316 c.1000G>A p.E334K Chr.19: 49070434-49070434 on Build GRCh38
3672969 c.1303G>T p.G435W Chr.19: 49070131-49070131 on Build GRCh38
6497663 c.1231G>T p.G411W Chr.19: 49070203-49070203 on Build GRCh38
999199 c.1055C>T p.A352V Chr.19: 49070379-49070379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747993638 Chr.19: 49070021-49070021 on Build GRCh38
rs762119570 Chr.19: 49070071-49070071 on Build GRCh38
rs765895551 Chr.19: 49070088-49070088 on Build GRCh38
rs752313141 Chr.19: 49070295-49070295 on Build GRCh38
rs986410711 Chr.19: 49070145-49070145 on Build GRCh38
rs771346014 Chr.19: 49070288-49070288 on Build GRCh38
rs973030348 Chr.19: 49069941-49069941 on Build GRCh38
rs771254385 Chr.19: 49070027-49070027 on Build GRCh38
rs761821227 Chr.19: 49070427-49070427 on Build GRCh38
rs751088840 Chr.19: 49070189-49070189 on Build GRCh38
rs113408235 Chr.19: 49070348-49070348 on Build GRCh38
rs551614576 Chr.19: 49070246-49070246 on Build GRCh38
rs376020507 Chr.19: 49070043-49070043 on Build GRCh38
rs759065174 Chr.19: 49070201-49070201 on Build GRCh38
rs371776324 Chr.19: 49070420-49070420 on Build GRCh38
rs772250720 Chr.19: 49070131-49070131 on Build GRCh38
rs745502974 Chr.19: 49070397-49070397 on Build GRCh38
rs781266465 Chr.19: 49070260-49070260 on Build GRCh38
rs533444990 Chr.19: 49070217-49070217 on Build GRCh38
rs906406795 Chr.19: 49070363-49070363 on Build GRCh38
rs770141919 Chr.19: 49070174-49070174 on Build GRCh38
rs138884521 Chr.19: 49070162-49070162 on Build GRCh38
rs753399863 Chr.19: 49070279-49070282 on Build GRCh38
rs375428032 Chr.19: 49070108-49070110 on Build GRCh38
rs145070146 Chr.19: 49070132-49070132 on Build GRCh38
rs755248193 Chr.19: 49070336-49070336 on Build GRCh38
rs965697269 Chr.19: 49069933-49069933 on Build GRCh38
rs980458921 Chr.19: 49070025-49070025 on Build GRCh38
rs559662958 Chr.19: 49070214-49070214 on Build GRCh38
rs761166864 Chr.19: 49070045-49070045 on Build GRCh38
rs771452572 Chr.19: 49070041-49070041 on Build GRCh38
rs969157340 Chr.19: 49070220-49070220 on Build GRCh38
rs117399970 Chr.19: 49069988-49069988 on Build GRCh38
rs568262365 Chr.19: 49069979-49069979 on Build GRCh38
rs1003411955 Chr.19: 49070378-49070378 on Build GRCh38
rs138349875 Chr.19: 49070123-49070123 on Build GRCh38
rs920878472 Chr.19: 49069942-49069942 on Build GRCh38
rs755932407 Chr.19: 49070111-49070111 on Build GRCh38
rs986319607 Chr.19: 49069947-49069947 on Build GRCh38
rs753834131 Chr.19: 49070313-49070313 on Build GRCh38
rs541248038 Chr.19: 49070212-49070212 on Build GRCh38
rs779475690 Chr.19: 49069931-49069931 on Build GRCh38
rs751094182 Chr.19: 49070093-49070093 on Build GRCh38
rs763988814 Chr.19: 49070303-49070303 on Build GRCh38
rs756960238 Chr.19: 49070329-49070329 on Build GRCh38
rs1054383308 Chr.19: 49070006-49070006 on Build GRCh38
rs748270841 Chr.19: 49070347-49070347 on Build GRCh38
rs371100804 Chr.19: 49070226-49070226 on Build GRCh38
rs760396855 Chr.19: 49070106-49070106 on Build GRCh38
rs747141982 Chr.19: 49070141-49070141 on Build GRCh38
rs948700780 Chr.19: 49070327-49070327 on Build GRCh38
rs774869945 Chr.19: 49070396-49070396 on Build GRCh38
rs781325560 Chr.19: 49070104-49070104 on Build GRCh38
rs370367266 Chr.19: 49070317-49070317 on Build GRCh38
rs12975537 Chr.19: 49070108-49070108 on Build GRCh38
rs778095377 Chr.19: 49070368-49070368 on Build GRCh38
rs765554335 Chr.19: 49070072-49070072 on Build GRCh38
rs767165812 Chr.19: 49070208-49070208 on Build GRCh38
rs754932345 Chr.19: 49070098-49070098 on Build GRCh38
rs752701716 Chr.19: 49070107-49070107 on Build GRCh38
rs143866728 Chr.19: 49070335-49070335 on Build GRCh38
rs374770795 Chr.19: 49070342-49070342 on Build GRCh38
rs1017219 Chr.19: 49070181-49070181 on Build GRCh38
rs572360689 Chr.19: 49070035-49070035 on Build GRCh38
rs750280618 Chr.19: 49070434-49070434 on Build GRCh38
rs939634781 Chr.19: 49070037-49070037 on Build GRCh38
rs554063947 Chr.19: 49069995-49069995 on Build GRCh38
rs549871279 Chr.19: 49070388-49070388 on Build GRCh38
rs760054168 Chr.19: 49070423-49070423 on Build GRCh38
rs767077866 Chr.19: 49070292-49070292 on Build GRCh38
rs147370499 Chr.19: 49070435-49070435 on Build GRCh38
rs531173814 Chr.19: 49070291-49070291 on Build GRCh38
rs762315865 Chr.19: 49070171-49070171 on Build GRCh38
rs140322772 Chr.19: 49070177-49070177 on Build GRCh38
rs773591235 Chr.19: 49070287-49070287 on Build GRCh38
rs142738378 Chr.19: 49070430-49070430 on Build GRCh38
rs367576655 Chr.19: 49070398-49070398 on Build GRCh38
rs756625104 Chr.19: 49070351-49070351 on Build GRCh38
rs761552109 Chr.19: 49070109-49070110 on Build GRCh38
rs756018142 Chr.19: 49070213-49070213 on Build GRCh38
rs138061289 Chr.19: 49070227-49070227 on Build GRCh38
rs138686305 Chr.19: 49070425-49070425 on Build GRCh38
rs750029780 Chr.19: 49070083-49070083 on Build GRCh38
rs889215599 Chr.19: 49070070-49070070 on Build GRCh38
rs903234792 Chr.19: 49069989-49069989 on Build GRCh38
rs764627605 Chr.19: 49070055-49070055 on Build GRCh38
rs760629817 Chr.19: 49070297-49070297 on Build GRCh38
rs765018989 Chr.19: 49070334-49070334 on Build GRCh38
rs1015063959 Chr.19: 49069930-49069930 on Build GRCh38
rs749834502 Chr.19: 49070383-49070383 on Build GRCh38
rs777084632 Chr.19: 49070158-49070158 on Build GRCh38
rs372895611 Chr.19: 49070424-49070424 on Build GRCh38
rs781127194 Chr.19: 49070014-49070014 on Build GRCh38
rs3810188 Chr.19: 49070261-49070261 on Build GRCh38
rs58259114 Chr.19: 49070057-49070057 on Build GRCh38
rs746043888 Chr.19: 49070125-49070125 on Build GRCh38
rs775117076 Chr.19: 49070417-49070417 on Build GRCh38
rs562010051 Chr.19: 49070176-49070176 on Build GRCh38
rs543909984 Chr.19: 49070157-49070157 on Build GRCh38
rs11083959 Chr.19: 49069949-49069949 on Build GRCh38
rs762658825 Chr.19: 49070180-49070180 on Build GRCh38
rs947763680 Chr.19: 49070003-49070003 on Build GRCh38
rs186760468 Chr.19: 49069944-49069944 on Build GRCh38
rs983921501 Chr.19: 49069987-49069987 on Build GRCh38
rs774582553 Chr.19: 49070029-49070029 on Build GRCh38
rs546265870 Chr.19: 49070040-49070040 on Build GRCh38
rs151047920 Chr.19: 49070110-49070110 on Build GRCh38
rs10535426 Chr.19: 49070106-49070108 on Build GRCh38
rs770266312 Chr.19: 49070285-49070285 on Build GRCh38
rs1043844175 Chr.19: 49070067-49070067 on Build GRCh38

View Full Product Details